Product References
Tet-mediated DNA demethylation regulates specification of hematopoietic stem and progenitor cells during mammalian embryogenesis.
Science advances
Ma L,Tang Q,Gao X,Lee J,Lei R,Suzuki M,Zheng D,Ito K,Frenette PS,Dawlaty MM
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2022
Smc3 dosage regulates B cell transit through germinal centers and restricts their malignant transformation.
Nature immunology
Rivas MA,Meydan C,Chin CR,Challman MF,Kim D,Bhinder B,Kloetgen A,Viny AD,Teater MR,McNally DR,Doane AS,Béguelin W,Fernández MTC,Shen H,Wang X,Levine RL,Chen Z,Tsirigos A,Elemento O,Mason CE,Melnick AM
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Aged hematopoietic stem cells are refractory to bloodborne systemic rejuvenation interventions.
The Journal of experimental medicine
Ho TT,Dellorusso PV,Verovskaya EV,Bakker ST,Flach J,Smith LK,Ventura PB,Lansinger OM,Hérault A,Zhang SY,Kang YA,Mitchell CA,Villeda SA,Passegué E
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
Differentiation of fetal hematopoietic stem cells requires ARID4B to restrict autocrine KITLG/KIT-Src signaling.
Cell reports
Young IC,Wu B,Andricovich J,Chuang ST,Li R,Tzatsos A,Wu RC,Wu MY
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue Nov 23 00:00:00 EST 2021
Ddb1 Is Essential for the Expansion of CD4+ Helper T Cells by Regulating Cell Cycle Progression and Cell Death.
Frontiers in immunology
Yang L,Chen W,Li L,Xiao Y,Fan S,Zhang Q,Xia T,Li M,Hong Y,Zhao T,Li Q,Liu WH,Xiao N
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Dec 20 00:00:00 EST 2021
Development of a VRC01-class germline targeting immunogen derived from anti-idiotypic antibodies.
Cell reports
Seydoux E,Wan YH,Feng J,Wall A,Aljedani S,Homad LJ,MacCamy AJ,Weidle C,Gray MD,Brumage L,Taylor JJ,Pancera M,Stamatatos L,McGuire AT
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue May 04 00:00:00 EDT 2021
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
RIPK3 and Caspase-1/11 Are Necessary for Optimal Antigen-Specific CD8 T Cell Response Elicited by Genetically Modified Listeria monocytogenes.
Frontiers in immunology
Rana A,de Almeida FC,Paico Montero HA,Gonzales Carazas MM,Bortoluci KR,Sad S,Amarante-Mendes GP
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Fri Mar 26 00:00:00 EDT 2021
Engineering monocyte/macrophage-specific glucocerebrosidase expression in human hematopoietic stem cells using genome editing.
Nature communications
Scharenberg SG,Poletto E,Lucot KL,Colella P,Sheikali A,Montine TJ,Porteus MH,Gomez-Ospina N
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Fri Jul 03 00:00:00 EDT 2020
Attenuation of TCR-induced transcription by Bach2 controls regulatory T cell differentiation and homeostasis.
Nature communications
Sidwell T,Liao Y,Garnham AL,Vasanthakumar A,Gloury R,Blume J,Teh PP,Chisanga D,Thelemann C,de Labastida Rivera F,Engwerda CR,Corcoran L,Kometani K,Kurosaki T,Smyth GK,Shi W,Kallies A
13-0453-82 was used in Flow Cytometry to show homeostasis of thymically-derived and peripherally-derived Treg cells is maintained through the balancing of TCR signaling induced transcriptional activity by Bach2.
Tue Jan 14 00:00:00 EST 2020
TH17 cells require ongoing classic IL-6 receptor signaling to retain transcriptional and functional identity.
Science immunology
Harbour SN,DiToro DF,Witte SJ,Zindl CL,Gao M,Schoeb TR,Jones GW,Jones SA,Hatton RD,Weaver CT
13-0453-82 was used in Immunohistochemistry (Frozen) to find that IL-6 signaling is required for both induction and maintenance of mouse TH17 cells; IL-6Rα-deficient TH17 cells rapidly lost their TH17 phenotype and did not cause disease in two models of colitis.
Fri Jul 17 00:00:00 EDT 2020
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
Interleukin-21 Induces Short-Lived Effector CD8+ T Cells but Does Not Inhibit Their Exhaustion after Mycobacterium bovis BCG Infection in Mice.
Infection and immunity
Noguchi N,Nakamura R,Hatano S,Yamada H,Sun X,Ohara N,Yoshikai Y
13-0453 was used in Flow cytometry/Cell sorting to reveal the impact of IL-21 signalling has on the differentiation of short-lived effector CD8+ Tc cells (SLECs) into early effector CD8+ T cells (EECs) but does not halt T cells exhaustion after BCG infection in mice.
Wed Aug 01 00:00:00 EDT 2018
The H3K4 methyltransferase Setd1b is essential for hematopoietic stem and progenitor cell homeostasis in mice.
eLife
Schmidt K,Zhang Q,Tasdogan A,Petzold A,Dahl A,Arneth BM,Slany R,Fehling HJ,Kranz A,Stewart AF,Anastassiadis K
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue Jun 19 00:00:00 EDT 2018
An immune cell spray (ICS) formulation allows for the delivery of functional monocyte/macrophages.
Scientific reports
Beneke V,Küster F,Neehus AL,Hesse C,Lopez-Rodriguez E,Haake K,Rafiei Hashtchin A,Schott JW,Walter D,Braun A,Wolkers WF,Ackermann M,Lachmann N
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Fri Nov 02 00:00:00 EDT 2018
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Jun 05 00:00:00 EDT 2017
The Deubiquitinase OTULIN Is an Essential Negative Regulator of Inflammation and Autoimmunity.
Cell
Damgaard RB,Walker JA,Marco-Casanova P,Morgan NV,Titheradge HL,Elliott PR,McHale D,Maher ER,McKenzie ANJ,Komander D
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Thu Aug 25 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Neutrophil ageing is regulated by the microbiome.
Nature
Zhang D,Chen G,Manwani D,Mortha A,Xu C,Faith JJ,Burk RD,Kunisaki Y,Jang JE,Scheiermann C,Merad M,Frenette PS
13-0453 was used in Flow cytometry/Cell sorting to show that neutrophil pro-inflammatory activity correlates positively with their ageing whilst in circulation.
Thu Sep 24 00:00:00 EDT 2015
MHC-mismatched mixed chimerism augments thymic regulatory T-cell production and prevents relapse of EAE in mice.
Proceedings of the National Academy of Sciences of the United States of America
Wu L,Li N,Zhang M,Xue SL,Cassady K,Lin Q,Riggs AD,Zeng D
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue Dec 29 00:00:00 EST 2015
Alternatively activated macrophages promote pancreatic fibrosis in chronic pancreatitis.
Nature communications
Xue J,Sharma V,Hsieh MH,Chawla A,Murali R,Pandol SJ,Habtezion A
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon May 18 00:00:00 EDT 2015
Heme-mediated SPI-C induction promotes monocyte differentiation into iron-recycling macrophages.
Cell
Haldar M,Kohyama M,So AY,Kc W,Wu X,Briseño CG,Satpathy AT,Kretzer NM,Arase H,Rajasekaran NS,Wang L,Egawa T,Igarashi K,Baltimore D,Murphy TL,Murphy KM
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
WASH is required for the differentiation commitment of hematopoietic stem cells in a c-Myc-dependent manner.
The Journal of experimental medicine
Xia P,Wang S,Huang G,Zhu P,Li M,Ye B,Du Y,Fan Z
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Mon Sep 22 00:00:00 EDT 2014
Lymph node stromal cells constrain immunity via MHC class II self-antigen presentation.
eLife
Baptista AP,Roozendaal R,Reijmers RM,Koning JJ,Unger WW,Greuter M,Keuning ED,Molenaar R,Goverse G,Sneeboer MM,den Haan JM,Boes M,Mebius RE
13-0453 was used in Flow cytometry/Cell sorting to investigate the role of MHC-II in lymph node stromal cells, showing that stromal cells constrain immunity through MHC-II expression.
Wed Nov 19 00:00:00 EST 2014
NK cell-extrinsic IL-18 signaling is required for efficient NK-cell activation by vaccinia virus.
European journal of immunology
Brandstadter JD,Huang X,Yang Y
13-0453 was used in Flow cytometry/Cell sorting to provide insights into the design of effective NK-cell-based therapies for viral infections and cancer.
Mon Sep 01 00:00:00 EDT 2014
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
In vivo generation of transplantable human hematopoietic cells from induced pluripotent stem cells.
Blood
Amabile G,Welner RS,Nombela-Arrieta C,D'Alise AM,Di Ruscio A,Ebralidze AK,Kraytsberg Y,Ye M,Kocher O,Neuberg DS,Khrapko K,Silberstein LE,Tenen DG
13-0453 was used in Flow cytometry/Cell sorting to develop an in vivo system in which human induced pluripotent stem cells differentiate within teratomas to derive functional myeloid and lymphoid cells.
Thu Feb 21 00:00:00 EST 2013
Arrayed lentiviral barcoding for quantification analysis of hematopoietic dynamics.
Stem cells (Dayton, Ohio)
Grosselin J,Sii-Felice K,Payen E,Chretien S,Tronik-Le Roux D,Leboulch P
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
Bcl11a controls Flt3 expression in early hematopoietic progenitors and is required for pDC development in vivo.
PloS one
Wu X,Satpathy AT,Kc W,Liu P,Murphy TL,Murphy KM
13-0453 was used in Flow cytometry/Cell sorting to investigate the role of Bcl11a in the development of dendritic cells, showing that it controls Flt3 expression in early hematopoietic progenitors and is required for pDC development.
Fri Jan 10 00:00:00 EST 2014
Plasmacytoid dendritic cells transport peripheral antigens to the thymus to promote central tolerance.
Immunity
Hadeiba H,Lahl K,Edalati A,Oderup C,Habtezion A,Pachynski R,Nguyen L,Ghodsi A,Adler S,Butcher EC
13-0453 was used in Immunohistochemistry on frozen tissues to explore the mechanisms underlying the contribution to immune tolerance conducted by peripheral dendritic cells.
Fri Mar 23 00:00:00 EDT 2012
Thymus-autonomous T cell development in the absence of progenitor import.
The Journal of experimental medicine
Martins VC,Ruggiero E,Schlenner SM,Madan V,Schmidt M,Fink PJ,von Kalle C,Rodewald HR
13-0453 was used in Flow cytometry/Cell sorting to characterise the previously unrecognised capacity of the thymus to sustain T cell development during bone marrow progenitor deprivation.
Mon Jul 30 00:00:00 EDT 2012
T cell apoptosis and induction of Foxp3+ regulatory T cells underlie the therapeutic efficacy of CD4 blockade in experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Duarte J,Carrié N,Oliveira VG,Almeida C,Agua-Doce A,Rodrigues L,Simas JP,Mars LT,Graca L
13-0453 was used in Flow cytometry/Cell sorting to investigate the mechanism underlying effective MS prevention, showing that apoptosis of T cells and induction of Foxp3+ Tregs underlie efficacy of CD4 blockade in EAE.
Wed Aug 15 00:00:00 EDT 2012
Direct reduction of antigen receptor expression in polyclonal B cell populations developing in vivo results in light chain receptor editing.
Journal of immunology (Baltimore, Md. : 1950)
Shen S,Manser T
13-0453 was used in Flow cytometry/Cell sorting to test the role of BCR expression plays in receptor editing in an polyclonal, in vivo population of B cells.
Sun Jan 01 00:00:00 EST 2012
In vitro Ag Cross-presentation and in vivo Ag Cross-presentation by Dendritic Cells in the Mouse.
Bio-protocol
Ghosh M,Shapiro LH
Published figure using CD45.1 monoclonal antibody (Product # 13-0453-82) in Flow Cytometry
Thu Dec 20 00:00:00 EST 2012
IL-6 induced by Staphylococcus aureus infection prevents the induction of skin allograft acceptance in mice.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Ahmed EB,Wang T,Daniels M,Alegre ML,Chong AS
13-0453 was used in Flow cytometry/Cell sorting to reveal that infection with S. aureus, but not P. aeruginosa, at the time of transplantation prohibits successful skin allograft acceptance by inducing inflammation and IL-6 production.
Sun May 01 00:00:00 EDT 2011
Epigenetic changes at Il12rb2 and Tbx21 in relation to plasticity behavior of Th17 cells.
Journal of immunology (Baltimore, Md. : 1950)
Bending D,Newland S,Krejcí A,Phillips JM,Bray S,Cooke A
13-0453 was used in Flow cytometry/Cell sorting to examine the epigenetic mechanisms underpinning murine Th17 plasticity in vivo.
Tue Mar 15 00:00:00 EDT 2011
Nonhematopoietic antigen blocks memory programming of alloreactive CD8+ T cells and drives their eventual exhaustion in mouse models of bone marrow transplantation.
The Journal of clinical investigation
Flutter B,Edwards N,Fallah-Arani F,Henderson S,Chai JG,Sivakumaran S,Ghorashian S,Bennett CL,Freeman GJ,Sykes M,Chakraverty R
13-0453 was used in Flow cytometry/Cell sorting to explore the role of recipient antigen expression by nonhaematopoietic cells in the failure to sustain effective cytotoxic T lymphocyte immunity following blood transplantation.
Mon Nov 01 00:00:00 EDT 2010
Germinal center T follicular helper cell IL-4 production is dependent on signaling lymphocytic activation molecule receptor (CD150).
Journal of immunology (Baltimore, Md. : 1950)
Yusuf I,Kageyama R,Monticelli L,Johnston RJ,Ditoro D,Hansen K,Barnett B,Crotty S
13-0453 was used in Immunofluorescence to study the roles of SAP adaptors and SLAM family receptors in the development and function of T follicular helper cells.
Thu Jul 01 00:00:00 EDT 2010
Intermediate-term hematopoietic stem cells with extended but time-limited reconstitution potential.
Cell stem cell
Benveniste P,Frelin C,Janmohamed S,Barbara M,Herrington R,Hyam D,Iscove NN
13-0453 was used in Flow cytometry/Cell sorting to investigate the presence of "intermediate-term" multipotent hematopoietic stem cells, revealing that these cells are able to sustain clones of differentiating cells for only 4-6 weeks.
Fri Jan 08 00:00:00 EST 2010
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
13-0453 was used in Flow cytometry/Cell sorting to track epitope-specific T cells.
Tue Jun 30 00:00:00 EDT 2009
Engagement of TLR2 does not reverse the suppressor function of mouse regulatory T cells, but promotes their survival.
Journal of immunology (Baltimore, Md. : 1950)
Chen Q,Davidson TS,Huter EN,Shevach EM
13-0453 was used in Flow cytometry/Cell sorting to investigate the role of TLRs in adaptive immunity and in regulatory T cells.
Thu Oct 01 00:00:00 EDT 2009
Combined anti-CD40 conditioning and well-timed immunization prolongs CD8+ T cell accumulation and control of established brain tumors.
Journal of immunotherapy (Hagerstown, Md. : 1997)
Ryan CM,Staveley-O'Carroll K,Schell TD
13-0453 was used in Flow cytometry/Cell sorting to investigate the augmentation of CD8+ T-cell-mediated adoptive immunotherapy by targeting a weakly immunogenic epitope.
Mon Jul 13 00:00:00 EDT 2009
miR-150, a microRNA expressed in mature B and T cells, blocks early B cell development when expressed prematurely.
Proceedings of the National Academy of Sciences of the United States of America
Zhou B,Wang S,Mayr C,Bartel DP,Lodish HF
13-0453 was used in Immunofluorescence to study how miR-150 can down-regulate mRNAs important for B cell function, and how it can block further B cell development when expressed prematurely.
Tue Apr 24 00:00:00 EDT 2007
miR-150, a microRNA expressed in mature B and T cells, blocks early B cell development when expressed prematurely.
Proceedings of the National Academy of Sciences of the United States of America
Zhou B,Wang S,Mayr C,Bartel DP,Lodish HF
13-0453 was used in Immunofluorescence to study how miR-150 can down-regulate mRNAs important for B cell function, and how it can block further B cell development when expressed prematurely.
Tue Apr 24 00:00:00 EDT 2007
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
13-0453 was used in Flow cytometry/Cell sorting to investigate the control of haematopoiesis by peroxide-sensitive regulatory mechanisms.
Wed Mar 01 00:00:00 EST 2006
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
13-0453 was used in Flow cytometry/Cell sorting to investigate the control of haematopoiesis by peroxide-sensitive regulatory mechanisms.
Wed Mar 01 00:00:00 EST 2006
Enhanced engraftment of hematopoietic stem/progenitor cells by the transient inhibition of an adaptor protein, Lnk.
Blood
Takizawa H,Kubo-Akashi C,Nobuhisa I,Kwon SM,Iseki M,Taga T,Takatsu K,Takaki S
13-0453 was used in Flow cytometry/Cell sorting to investigate the potential of Lnk for augmented engraftment of HSCs.
Sat Apr 01 00:00:00 EST 2006
Endomucin, a CD34-like sialomucin, marks hematopoietic stem cells throughout development.
The Journal of experimental medicine
Matsubara A,Iwama A,Yamazaki S,Furuta C,Hirasawa R,Morita Y,Osawa M,Motohashi T,Eto K,Ema H,Kitamura T,Vestweber D,Nakauchi H
13-0453 was used in Flow cytometry/Cell sorting to identify for the first time Endomucin as a cell-surface molecule specific to hematopoietic stem cells (HSCs).
Mon Dec 05 00:00:00 EST 2005
Essential roles of CD8+CD122+ regulatory T cells in the maintenance of T cell homeostasis.
The Journal of experimental medicine
Rifa'i M,Kawamoto Y,Nakashima I,Suzuki H
13-0453 was used in Flow cytometry/Cell sorting to investigate the effect of transfer of CD8+CD122+ cells into CD122-deficient neonates, showing that CD8+CD122+ regulatory T cells are essential for T cell homeostasis.
Mon Nov 01 00:00:00 EST 2004