Product References
ATR kinase activity promotes antibody class switch recombination in B cells through cell cycle regulation without suppressing DSB resection and microhomology usage.
Journal of leukocyte biology
Sun X,Liu M,Bai J,Xu J,Zhu C,Dong J,Chen C
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Western Blot
Wed Dec 01 00:00:00 EST 2021
The 3'-flap endonuclease XPF-ERCC1 promotes alternative end joining and chromosomal translocation during B cell class switching.
Cell reports
Bai W,Zhu G,Xu J,Chen P,Meng F,Xue H,Chen C,Dong J
14-5959-82 was used in Western Blot to reveal the role of the flap endonuclease, XPF-ERCC1, in alternative end joining and oncogenic translocation in mouse B cells.
Tue Sep 28 00:00:00 EDT 2021
Activation-induced cytidine deaminase is a possible regulator of cross-talk between oocytes and granulosa cells through GDF-9 and SCF feedback system.
Scientific reports
Iizuka T,Wakae K,Ono M,Suzuki T,Mizumoto Y,Kitamura K,Horike SI,Muramatsu M,Fujiwara H
14-5959-82 was used in Immunohistochemistry to study activation-induced cytidine deaminase expression in murine ovaries.
Mon Feb 15 00:00:00 EST 2021
Immunoglobulin Class Switch Recombination Is Initiated by Rare Cytosine Deamination Events at Switch Regions.
Molecular and cellular biology
Kim A,Han L,Yu K
14-5959 was used in Western Blotting to examine AID footprints across the entire length of an engineered switch region in cells ablated for uracil repair.
Wed Jul 29 00:00:00 EDT 2020
Interplay between UNG and AID governs intratumoral heterogeneity in mature B cell lymphoma.
PLoS genetics
Delgado P,Álvarez-Prado ÁF,Marina-Zárate E,Sernandez IV,Mur SM,de la Barrera J,Sanchez-Cabo F,Cañamero M,de Molina A,Belver L,de Yébenes VG,Ramiro AR
14-5959-82 was used in Western Blot to show using a conditional knock-in strategy that AID supra-activity alone is not sufficient to generate B cell transformation.
Tue Dec 01 00:00:00 EST 2020
Interplay between UNG and AID governs intratumoral heterogeneity in mature B cell lymphoma.
PLoS genetics
Delgado P,Álvarez-Prado ÁF,Marina-Zárate E,Sernandez IV,Mur SM,de la Barrera J,Sanchez-Cabo F,Cañamero M,de Molina A,Belver L,de Yébenes VG,Ramiro AR
14-5959-82 was used in Western Blot to show using a conditional knock-in strategy that AID supra-activity alone is not sufficient to generate B cell transformation.
Tue Dec 01 00:00:00 EST 2020
Liver Is a Generative Site for the B Cell Response to Ehrlichia muris.
Immunity
Trivedi N,Weisel F,Smita S,Joachim S,Kader M,Radhakrishnan A,Clouser C,Rosenfeld AM,Chikina M,Vigneault F,Hershberg U,Ismail N,Shlomchik MJ
14-5959-82 was used in Flow Cytometry to investigate the nature of liver B cell and spleen response to Ehrlichia muris.
Tue Dec 17 00:00:00 EST 2019
A licensing step links AID to transcription elongation for mutagenesis in B cells.
Nature communications
Methot SP,Litzler LC,Subramani PG,Eranki AK,Fifield H,Patenaude AM,Gilmore JC,Santiago GE,Bagci H,Côté JF,Larijani M,Verdun RE,Di Noia JM
14-5959 was used in Immunocytochemistry-immunoflourescence to study the mechanisms underlying productive deamination compared with non-productive activation-induced deaminase targeting.
Wed Mar 28 00:00:00 EDT 2018
Phosphatidylinositol 3-kinase δ blockade increases genomic instability in B cells.
Nature
Compagno M,Wang Q,Pighi C,Cheong TC,Meng FL,Poggio T,Yeap LS,Karaca E,Blasco RB,Langellotto F,Ambrogio C,Voena C,Wiestner A,Kasar SN,Brown JR,Sun J,Wu CJ,Gostissa M,Alt FW,Chiarle R
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Western Blot
Thu Feb 23 00:00:00 EST 2017
Phosphatidylinositol 3-kinase δ blockade increases genomic instability in B cells.
Nature
Compagno M,Wang Q,Pighi C,Cheong TC,Meng FL,Poggio T,Yeap LS,Karaca E,Blasco RB,Langellotto F,Ambrogio C,Voena C,Wiestner A,Kasar SN,Brown JR,Sun J,Wu CJ,Gostissa M,Alt FW,Chiarle R
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Western Blot
Thu Feb 23 00:00:00 EST 2017
The cell cycle restricts activation-induced cytidine deaminase activity to early G1.
The Journal of experimental medicine
Wang Q,Kieffer-Kwon KR,Oliveira TY,Mayer CT,Yao K,Pai J,Cao Z,Dose M,Casellas R,Jankovic M,Nussenzweig MC,Robbiani DF
14-5959 was used in Western Blotting to show that activation-induced cytidine deaminase (AID) deaminates cytosine after cytokinesis in early G1, and that processing of these lesions continues throughout the G1 phase of the cell cycle.
Sun Jan 01 00:00:00 EST 2017
CCCTC-Binding Factor Locks Premature IgH Germline Transcription and Restrains Class Switch Recombination.
Frontiers in immunology
Marina-Zárate E,Pérez-García A,Ramiro AR
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Western Blot
Mon Jan 31 00:00:00 EST 2022
Epstein-Barr virus nuclear protein EBNA3C directly induces expression of AID and somatic mutations in B cells.
The Journal of experimental medicine
Kalchschmidt JS,Bashford-Rogers R,Paschos K,Gillman AC,Styles CT,Kellam P,Allday MJ
14595982 was used in western blot to demonstrate induction of functional AID by EBNA3C
Mon May 30 00:00:00 EDT 2016
Epstein-Barr virus nuclear protein EBNA3C directly induces expression of AID and somatic mutations in B cells.
The Journal of experimental medicine
Kalchschmidt JS,Bashford-Rogers R,Paschos K,Gillman AC,Styles CT,Kellam P,Allday MJ
14595982 was used in western blot to demonstrate induction of functional AID by EBNA3C
Mon May 30 00:00:00 EDT 2016
TGF-β suppression of HBV RNA through AID-dependent recruitment of an RNA exosome complex.
PLoS pathogens
Liang G,Liu G,Kitamura K,Wang Z,Chowdhury S,Monjurul AM,Wakae K,Koura M,Shimadu M,Kinoshita K,Muramatsu M
14-5959 was used in Western Blotting to show that reduction of HBV transcripts by TGF-β is dependent on AID expression and can inhibit viral replication.
Wed Apr 01 00:00:00 EDT 2015
TGF-β suppression of HBV RNA through AID-dependent recruitment of an RNA exosome complex.
PLoS pathogens
Liang G,Liu G,Kitamura K,Wang Z,Chowdhury S,Monjurul AM,Wakae K,Koura M,Shimadu M,Kinoshita K,Muramatsu M
14-5959 was used in Western Blotting to show that reduction of HBV transcripts by TGF-β is dependent on AID expression and can inhibit viral replication.
Wed Apr 01 00:00:00 EDT 2015
HSP90 inhibitors decrease AID levels and activity in mice and in human cells.
European journal of immunology
Montamat-Sicotte D,Litzler LC,Abreu C,Safavi S,Zahn A,Orthwein A,Müschen M,Oppezzo P,Muñoz DP,Di Noia JM
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
HSP90 inhibitors decrease AID levels and activity in mice and in human cells.
European journal of immunology
Montamat-Sicotte D,Litzler LC,Abreu C,Safavi S,Zahn A,Orthwein A,Müschen M,Oppezzo P,Muñoz DP,Di Noia JM
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
HSP90 inhibitors decrease AID levels and activity in mice and in human cells.
European journal of immunology
Montamat-Sicotte D,Litzler LC,Abreu C,Safavi S,Zahn A,Orthwein A,Müschen M,Oppezzo P,Muñoz DP,Di Noia JM
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
HSP90 inhibitors decrease AID levels and activity in mice and in human cells.
European journal of immunology
Montamat-Sicotte D,Litzler LC,Abreu C,Safavi S,Zahn A,Orthwein A,Müschen M,Oppezzo P,Muñoz DP,Di Noia JM
Published figure using AID monoclonal antibody (Product # 14-5959-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
The transcription factor Foxp1 is a critical negative regulator of the differentiation of follicular helper T cells.
Nature immunology
Wang H,Geng J,Wen X,Bi E,Kossenkov AV,Wolf AI,Tas J,Choi YS,Takata H,Day TJ,Chang LY,Sprout SL,Becker EK,Willen J,Tian L,Wang X,Xiao C,Jiang P,Crotty S,Victora GD,Showe LC,Tucker HO,Erikson J,Hu H
14-5959 was used in Immunohistochemistry to investigate the role of Foxp1 in CD4(+) cells, showing that it is a critical negative regulator of the differentiation of follicular helper T cells.
Tue Jul 01 00:00:00 EDT 2014
Generation and characterization of induced pluripotent stem cells from Aid-deficient mice.
PloS one
Shimamoto R,Amano N,Ichisaka T,Watanabe A,Yamanaka S,Okita K
14-5959 was used in Western Blotting to investigate the underlying mechanism of DNA demethylation in generation of induced pluripotent stem cells.
Mon Jun 29 00:00:00 EDT 2015
BCL6 positively regulates AID and germinal center gene expression via repression of miR-155.
The Journal of experimental medicine
Basso K,Schneider C,Shen Q,Holmes AB,Setty M,Leslie C,Dalla-Favera R
14-5959 was used in Immunofluorescence to investigate the potential role of BCL6 in regulating gene expression via microRNAs.
Mon Dec 17 00:00:00 EST 2012