Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Description: The 14-4-4S monoclonal antibody reacts with the mouse and rat major histocompatibility complex class II, I-E subregion-encoded glycoprotein in mice strains of H-2^k, H-2^d, H-2^p and H-2^r. H-2^b, H-2^f and H-2^s haplotypes do not express the I-E antigen. MHC class II is present on B cells, monocytes, macrophages, and dendritic cells. The 14-4-4S mAb has been reported to block antigen presentation in in vitro assays. This antibody cross-reacts with rat.
Applications Reported: 14-4-4S has been reported for use in flow cytometric analysis, immunoprecipitation, complement-dependent cytotoxity and immunohistochemical staining of frozen sections (Biotinylated 14-4-4S, cat. 13-5980, is recommended for IHC).
Applications Tested: The 14-4-4S antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell surface glycoprotein; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H2-IE-alpha; histocompatibility 2, class II antigen E alpha; I-E alpha MHC class II; I-E beta MHC class II; major histocompatibility complex H-2E beta chain; MGC117888; MHC class II antigen E alpha; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein
Gene Aliases: AI323765; E-alpha-f; H-2Ea; H-2Eb; H2-Ea; H2-Ea-ps; H2Eb; I-Ealpha; Ia-3; Ia-4; Ia3; Ia4
UniProt ID: (Mouse) P04224
Entrez Gene ID: (Mouse) 100504404, (Mouse) 14969
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support