Product References
Interleukin 6 regulates the expression of programmed cell death ligand 1 in thyroid cancer.
Cancer science
Zhang GQ,Jiao Q,Shen CT,Song HJ,Zhang HZ,Qiu ZL,Luo QY
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in Flow Cytometry
Mon Mar 01 00:00:00 EST 2021
PLCγ2 regulates TREM2 signalling and integrin-mediated adhesion and migration of human iPSC-derived macrophages.
Scientific reports
Obst J,Hall-Roberts HL,Smith TB,Kreuzer M,Magno L,Di Daniel E,Davis JB,Mead E
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in ELISA
Wed Oct 06 00:00:00 EDT 2021
Murine Model of Sinusitis Infection for Screening Antimicrobial and Immunomodulatory Therapies.
Frontiers in cellular and infection microbiology
Alford MA,Choi KG,Trimble MJ,Masoudi H,Kalsi P,Pletzer D,Hancock REW
14-7069 was used in an ELISA assay to describe a simple less virulent, epidemic P. aeruginosa cystic fibrosis clinical isolate LESB65 model allowing non-invasive tracking of bacterial infection, and screening of host-responses to infection and therapies.
Fri Jul 02 00:00:00 EDT 2021
PTPN2 Regulates Interactions Between Macrophages and Intestinal Epithelial Cells to Promote Intestinal Barrier Function.
Gastroenterology
Spalinger MR,Sayoc-Becerra A,Santos AN,Shawki A,Canale V,Krishnan M,Niechcial A,Obialo N,Scharl M,Li J,Nair MG,McCole DF
14-7069-81 was used in Inhibition Assays to investigate whether PTPN2 controls interactions between IECs and macrophages to maintain intestinal barrier function.
Sun Nov 01 00:00:00 EDT 2020
TREM2 Alzheimer's variant R47H causes similar transcriptional dysregulation to knockout, yet only subtle functional phenotypes in human iPSC-derived macrophages.
Alzheimer's research & therapy
Hall-Roberts H,Agarwal D,Obst J,Smith TB,Monzón-Sandoval J,Di Daniel E,Webber C,James WS,Mead E,Davis JB,Cowley SA
14-7069 was used in an ELISA assay to investigated cell phenotypes of the R47H mutant and TREM2 KO in a model of human microglia, and compared their transcriptional signatures, to determine the mechanism by which R47H TREM2 disrupts function.
Mon Nov 16 00:00:00 EST 2020
CD19+CD24hiCD38hi B Cells Are Expanded in Juvenile Dermatomyositis and Exhibit a Pro-Inflammatory Phenotype After Activation Through Toll-Like Receptor 7 and Interferon-α.
Frontiers in immunology
Piper CJM,Wilkinson MGL,Deakin CT,Otto GW,Dowle S,Duurland CL,Adams S,Marasco E,Rosser EC,Radziszewska A,Carsetti R,Ioannou Y,Beales PL,Kelberman D,Isenberg DA,Mauri C,Nistala K,Wedderburn LR
14-7069-81 was used in Flow Cytometry to identify that immature transitional B cells were significantly expanded in active JDM, actively dividing, and correlated positively with disease activity.
Wed Nov 20 00:00:00 EST 2019
Dengue virus NS1 cytokine-independent vascular leak is dependent on endothelial glycocalyx components.
PLoS pathogens
Glasner DR,Ratnasiri K,Puerta-Guardo H,Espinosa DA,Beatty PR,Harris E
14-7069 was used in an ELISA assay to evaluate the relative contributions of inflammatory mediators and endothelial cell-intrinsic pathways in Dengue virus.
Wed Nov 01 00:00:00 EDT 2017
Mesenchymal Stromal Cell-Derived Interleukin-6 Promotes Epithelial-Mesenchymal Transition and Acquisition of Epithelial Stem-Like Cell Properties in Ameloblastoma Epithelial Cells.
Stem cells (Dayton, Ohio)
Jiang C,Zhang Q,Shanti RM,Shi S,Chang TH,Carrasco L,Alawi F,Le AD
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in Immunofluorescence
Fri Sep 01 00:00:00 EDT 2017
Decitabine, a DNA-demethylating agent, promotes differentiation via NOTCH1 signaling and alters immune-related pathways in muscle-invasive bladder cancer.
Cell death & disease
Ramakrishnan S,Hu Q,Krishnan N,Wang D,Smit E,Granger V,Rak M,Attwood K,Johnson C,Morrison C,Pili R,Chatta G,Guru K,Gueron G,McNally L,Wang J,Woloszynska-Read A
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in T-Cell Activation
Thu Dec 14 00:00:00 EST 2017
Biallelic hypomorphic mutations in a linear deubiquitinase define otulipenia, an early-onset autoinflammatory disease.
Proceedings of the National Academy of Sciences of the United States of America
Zhou Q,Yu X,Demirkaya E,Deuitch N,Stone D,Tsai WL,Kuehn HS,Wang H,Yang D,Park YH,Ombrello AK,Blake M,Romeo T,Remmers EF,Chae JJ,Mullikin JC,Güzel F,Milner JD,Boehm M,Rosenzweig SD,Gadina M,Welch SB,Özen S,Topaloglu R,Abinun M,Kastner DL,Aksentijevich I
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in ELISA
Tue Sep 06 00:00:00 EDT 2016
Biallelic hypomorphic mutations in a linear deubiquitinase define otulipenia, an early-onset autoinflammatory disease.
Proceedings of the National Academy of Sciences of the United States of America
Zhou Q,Yu X,Demirkaya E,Deuitch N,Stone D,Tsai WL,Kuehn HS,Wang H,Yang D,Park YH,Ombrello AK,Blake M,Romeo T,Remmers EF,Chae JJ,Mullikin JC,Güzel F,Milner JD,Boehm M,Rosenzweig SD,Gadina M,Welch SB,Özen S,Topaloglu R,Abinun M,Kastner DL,Aksentijevich I
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in ELISA
Tue Sep 06 00:00:00 EDT 2016
Plasmonically amplified bioassay - Total internal reflection fluorescence vs. epifluorescence geometry.
Talanta
Hageneder S,Bauch M,Dostalek J
14706981 was used to assess IL-6 levels by plasmonic amplification using epifluorescence and total internal reflection fluorescence
Mon Aug 15 00:00:00 EDT 2016
Toll-Like Receptor Activation by Generalized Modules for Membrane Antigens from Lipid A Mutants of Salmonella enterica Serovars Typhimurium and Enteritidis.
Clinical and vaccine immunology : CVI
Rossi O,Caboni M,Negrea A,Necchi F,Alfini R,Micoli F,Saul A,MacLennan CA,Rondini S,Gerke C
14-7069 was used in an ELISA assay to investigate the differences between Salmonella enterica strains in terms of TLR activation by generalised modules for membrane antigens from lipid A mutants.
Fri Apr 01 00:00:00 EDT 2016
Activated human valvular interstitial cells sustain interleukin-17 production to recruit neutrophils in infective endocarditis.
Infection and immunity
Yeh CY,Shun CT,Kuo YM,Jung CJ,Hsieh SC,Chiu YL,Chen JW,Hsu RB,Yang CJ,Chia JS
14-7069 was used in Inhibition experiments to assess the role of activated valvular interstitial cells in valvular damage.
Mon Jun 01 00:00:00 EDT 2015
TL1A induces TCR independent IL-6 and TNF-α production and growth of PLZF⁺ leukocytes.
PloS one
Reichwald K,Jørgensen TZ,Tougaard P,Skov S
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in Flow Cytometry
Mon Sep 08 00:00:00 EDT 2014
Modulation of endotoxicity of Shigella generalized modules for membrane antigens (GMMA) by genetic lipid A modifications: relative activation of TLR4 and TLR2 pathways in different mutants.
The Journal of biological chemistry
Rossi O,Pesce I,Giannelli C,Aprea S,Caboni M,Citiulo F,Valentini S,Ferlenghi I,MacLennan CA,D'Oro U,Saul A,Gerke C
Published figure using IL-6 monoclonal antibody (Product # 14-7069-81) in ELISA
Fri Sep 05 00:00:00 EDT 2014
Monocytes from chronic HBV patients react in vitro to HBsAg and TLR by producing cytokines irrespective of stage of disease.
PloS one
Boltjes A,Groothuismink ZM,van Oord GW,Janssen HL,Woltman AM,Boonstra A
14-7069 was used in Flow cytometry/Cell sorting to investigate the in vitro and in vivo effects of hepatitis B virus surface antigen on monocytes from chronic HBV patients.
Tue Jan 06 00:00:00 EST 2015
Induction of cytokines in a macrophage cell line by proteins of Clostridium difficile.
FEMS immunology and medical microbiology
Vohra P,Poxton IR
14-7069 was used in an ELISA assay to assess the induction of cytokines in a macrophage cell line in response to different antigens prepared from five Clostridium difficile strains.
Fri Jun 01 00:00:00 EDT 2012
De novo generation and enhanced suppression of human CD4+CD25+ regulatory T cells by retinoic acid.
Journal of immunology (Baltimore, Md. : 1950)
Wang J,Huizinga TW,Toes RE
14-7069 was used in Neutralization experiments to investigate methods to convert T cells into Treg cells, showing that retinoic acid can be used for de novo generation of Tregs.
Tue Sep 15 00:00:00 EDT 2009