Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALSGSTPA PCWEEDECLD YYGMLSLHRM FEVVGGQLTE CELELLAFLL DEAPGAAGGL ARARSGLELL LELERRGQCD ESNLRLLGQL LRVLARHDLL PHLARKRRRP VSPERYSYGT SSSSKRTEGS CRRRRQSSSS ANSQQGQWET GSPPTKRQRR SRGRPSGGAR RRRRGAPAAP QQQSEPARPS SEGKVTCDIR LRVRAEYCEH GPALEQGVAS RRPQALARQL DVFGQATAVL RSRDLGSVVC DIKFSELSYL DAFWGDYLSG ALLQALRGVF LTEALREAVG REAVRLLVSV DEADYEAGRR RLLLMEEEGG RRPTEAS (1-326 aa encoded by BC027930)
Apoptotic signals are often triggered by cell surface death receptors through protein-protein interactions mediated by conserved domains such as the death effector domain. A novel death effector domain (DED)-containing protein, DEDD2, has been recently identified. Over-expression of DEDD2 in transfected cells induces moderate apoptosis and results in substantial sensitization to apoptosis induced by Fas, TRAIL, and FADD. More recently, it has been shown that DEDD2 can bind caspase-8 and -10 in addition to FLIP but not FADD. Like the related protein DEDD, DEDD2 translocates from the cytosol to the nucleus upon induction of apoptosis, and it has been suggested that DEDD2 may target caspase-8 to the nucleus and that DEDD2 thus plays a critical role in death receptor-induced apoptosis. At least two alternatively spliced transcript variants encoding distinct isoforms have been found for DEDD2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: death effector domain containing 2; death effector domain-containing DNA-binding protein 2; DED-containing protein FLAME-3; DNA-binding death effector domain-containing protein 2; FADD-like anti-apoptotic molecule 3
Gene Aliases: 2410050E11Rik; DEDD2; FLAME-3; FLAME3; PSEC0004
UniProt ID: (Human) Q8WXF8, (Mouse) Q8QZV0
Entrez Gene ID: (Human) 162989, (Mouse) 67379, (Rat) 687118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support