Product References
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
Diversity in medullary thymic epithelial cells controls the activity and availability of iNKT cells.
Nature communications
Lucas B,White AJ,Cosway EJ,Parnell SM,James KD,Jones ND,Ohigashi I,Takahama Y,Jenkinson WE,Anderson G
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Mon May 04 00:00:00 EDT 2020
Requirements for the differentiation of innate T-bethigh memory-phenotype CD4+ T lymphocytes under steady state.
Nature communications
Kawabe T,Yi J,Kawajiri A,Hilligan K,Fang D,Ishii N,Yamane H,Zhu J,Jankovic D,Kim KS,Trinchieri G,Sher A
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2020
B7-CD28 co-stimulation modulates central tolerance via thymic clonal deletion and Treg generation through distinct mechanisms.
Nature communications
Watanabe M,Lu Y,Breen M,Hodes RJ
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
CD160 serves as a negative regulator of NKT cells in acute hepatic injury.
Nature communications
Kim TJ,Park G,Kim J,Lim SA,Kim J,Im K,Shin MH,Fu YX,Del Rio ML,Rodriguez-Barbosa JI,Yee C,Suh KS,Kim SJ,Ha SJ,Lee KM
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Mon Jul 22 00:00:00 EDT 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
Complement Activation via a C3a Receptor Pathway Alters CD4+ T Lymphocytes and Mediates Lung Cancer Progression.
Cancer research
Kwak JW,Laskowski J,Li HY,McSharry MV,Sippel TR,Bullock BL,Johnson AM,Poczobutt JM,Neuwelt AJ,Malkoski SP,Weiser-Evans MC,Lambris JD,Clambey ET,Thurman JM,Nemenoff RA
15-5961 was used in Flow cytometry/Cell sorting to study the mechanism relating complement activation to tumour progression in multiple immunocompetent orthotopic models of lung cancer.
Mon Jan 01 00:00:00 EST 2018
STAT1 is a sex-specific tumor suppressor in colitis-associated colorectal cancer.
Molecular oncology
Crnčec I,Modak M,Gordziel C,Svinka J,Scharf I,Moritsch S,Pathria P,Schlederer M,Kenner L,Timelthaler G,Müller M,Strobl B,Casanova E,Bayer E,Mohr T,Stöckl J,Friedrich K,Eferl R
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Dysregulation of Kupffer Cells/Macrophages and Natural Killer T Cells in Steatohepatitis in LXRα Knockout Male Mice.
Endocrinology
Endo-Umeda K,Nakashima H,Umeda N,Seki S,Makishima M
15-5961 was used in Flow cytometry/Cell sorting to study the role fof LXRα in the pathogenesis of steatohepatitis.
Thu Mar 01 00:00:00 EST 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
A Paradoxical Tumor-Suppressor Role for the Rac1 Exchange Factor Vav1 in T Cell Acute Lymphoblastic Leukemia.
Cancer cell
Robles-Valero J,Lorenzo-Martín LF,Menacho-Márquez M,Fernández-Pisonero I,Abad A,Camós M,Toribio ML,Espinosa L,Bigas A,Bustelo XR
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Mon Nov 13 00:00:00 EST 2017
1810011o10 Rik Inhibits the Antitumor Effect of Intratumoral CD8+ T Cells through Suppression of Notch2 Pathway in a Murine Hepatocellular Carcinoma Model.
Frontiers in immunology
Dai K,Huang L,Huang YB,Chen ZB,Yang LH,Jiang YA
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
CD40-mediated signalling influences trafficking, T-cell receptor expression, and T-cell pathogenesis, in the NOD model of type 1 diabetes.
Immunology
Vaitaitis GM,Waid DM,Yussman MG,Wagner DH
15-5961 was used in Flow cytometry/Cell sorting to determine the mechanism by which CD40 results in the pathogenesis of type 1 diabetes.
Sun Oct 01 00:00:00 EDT 2017
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Hypoxia-Inducible Factor-2α Limits Natural Killer T Cell Cytotoxicity in Renal Ischemia/Reperfusion Injury.
Journal of the American Society of Nephrology : JASN
Zhang J,Han C,Dai H,Hou J,Dong Y,Cui X,Xu L,Zhang M,Xia Q
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Environmental and Genetic Activation of Hypothalamic BDNF Modulates T-cell Immunity to Exert an Anticancer Phenotype.
Cancer immunology research
Xiao R,Bergin SM,Huang W,Slater AM,Liu X,Judd RT,Lin ED,Widstrom KJ,Scoville SD,Yu J,Caligiuri MA,Cao L
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Protein kinase D regulates positive selection of CD4+ thymocytes through phosphorylation of SHP-1.
Nature communications
Ishikawa E,Kosako H,Yasuda T,Ohmuraya M,Araki K,Kurosaki T,Saito T,Yamasaki S
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Tue Sep 27 00:00:00 EDT 2016
MHC-mismatched mixed chimerism augments thymic regulatory T-cell production and prevents relapse of EAE in mice.
Proceedings of the National Academy of Sciences of the United States of America
Wu L,Li N,Zhang M,Xue SL,Cassady K,Lin Q,Riggs AD,Zeng D
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Tue Dec 29 00:00:00 EST 2015
Antibiotics in neonatal life increase murine susceptibility to experimental psoriasis.
Nature communications
Zanvit P,Konkel JE,Jiao X,Kasagi S,Zhang D,Wu R,Chia C,Ajami NJ,Smith DP,Petrosino JF,Abbatiello B,Nakatsukasa H,Chen Q,Belkaid Y,Chen ZJ,Chen W
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2015
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
Published figure using TCR beta monoclonal antibody (Product # 15-5961-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Glucocorticoid receptor-dependent immunomodulatory effect of ursodeoxycholic acid on liver lymphocytes in mice.
American journal of physiology. Gastrointestinal and liver physiology
Takigawa T,Miyazaki H,Kinoshita M,Kawarabayashi N,Nishiyama K,Hatsuse K,Ono S,Saitoh D,Seki S,Yamamoto J
15-5961 was used in Flow cytometry/Cell sorting to investigate the immunomodulatory effect of ursodeoxycholic acid on experimental hepatitis, endotoxin shock, and bacterial infection in mice.
Sun Sep 15 00:00:00 EDT 2013
PD-L1 and PD-L2 modulate airway inflammation and iNKT-cell-dependent airway hyperreactivity in opposing directions.
Mucosal immunology
Akbari O,Stock P,Singh AK,Lombardi V,Lee WL,Freeman GJ,Sharpe AH,Umetsu DT,Dekruyff RH
15-5961 was used in Flow cytometry/Cell sorting to investigate the roles of PD-L1 and PD-L2 in regulating invariant natural killer T cell mediated airway hyperreactivity in a mouse model of asthma.
Fri Jan 01 00:00:00 EST 2010
A CD1d-dependent antagonist inhibits the activation of invariant NKT cells and prevents development of allergen-induced airway hyperreactivity.
Journal of immunology (Baltimore, Md. : 1950)
Lombardi V,Stock P,Singh AK,Kerzerho J,Yang W,Sullivan BA,Li X,Shiratsuchi T,Hnatiuk NE,Howell AR,Yu KO,Porcelli SA,Tsuji M,Kronenberg M,Wilson SB,Akbari O
15-5961 was used in Flow cytometry/Cell sorting to test whether iNKT cell-targeted therapy could be used to treat allergen-induced airway disease.
Mon Feb 15 00:00:00 EST 2010
Induction of airway hyperreactivity by IL-25 is dependent on a subset of invariant NKT cells expressing IL-17RB.
Journal of immunology (Baltimore, Md. : 1950)
Stock P,Lombardi V,Kohlrautz V,Akbari O
15-5961 was used in Flow cytometry/Cell sorting to investigate the invariant NKT cell dependent pathway by which IL-25 induces Th2 responses and airway hyperreactivity in mice.
Wed Apr 15 00:00:00 EDT 2009
DNAM-1 promotes activation of cytotoxic lymphocytes by nonprofessional antigen-presenting cells and tumors.
The Journal of experimental medicine
Gilfillan S,Chan CJ,Cella M,Haynes NM,Rapaport AS,Boles KS,Andrews DM,Smyth MJ,Colonna M
15-5961 was used in Flow cytometry/Cell sorting to study the promoted activation of cytotoxic lymphocytes by nonprofessional antigen-presenting cells and tumors by dnam-1.
Mon Dec 22 00:00:00 EST 2008
CXCL9, but not CXCL10, promotes CXCR3-dependent immune-mediated kidney disease.
Journal of the American Society of Nephrology : JASN
Menke J,Zeller GC,Kikawada E,Means TK,Huang XR,Lan HY,Lu B,Farber J,Luster AD,Kelley VR
15-5961 was used in Flow cytometry/Cell sorting to identify additional therapeutic chemokine targets for immune-mediated nephritis.
Sun Jun 01 00:00:00 EDT 2008
ICOS/ICOSL interaction is required for CD4+ invariant NKT cell function and homeostatic survival.
Journal of immunology (Baltimore, Md. : 1950)
Akbari O,Stock P,Meyer EH,Freeman GJ,Sharpe AH,Umetsu DT,DeKruyff RH
15-5961 was used in Flow cytometry/Cell sorting to investigate the role of ICOS and iNKT cells in the development of airway hyperreactivity.
Tue Apr 15 00:00:00 EDT 2008