Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize Rac1, Rac2, and Rac3.
Immunogen sequence: MQAIKCVVV GDGAVGKTCL LISYTTNAFP GEYIPTVFDN YSANVMVDGK PVNLGLWDTA GQEDYDRLRP LSYPQTDVFL ICFSLVSPAS FENVRAKWYP EVRHHCPHTP ILLVGTKLDL RDDKDTIERL RDKKLAPITY PQGLAMAREI GSVKYLECSA LTQRGLKTVF DEAIRAVLCP PPVKKPGKKC TVF (1-192 aa encoded by BC015197)
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell migration-inducing gene 5 protein; EN-7 protein; GX; p21-Rac1; p21-Rac2; p21-Rac3; Protein EN-7; Ras-like protein TC25; RAS-related C3 botulinum substrate 2; Ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); Ras-related C3 botulinum toxin substrate 2; Ras-related C3 botulinum toxin substrate 3; ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3); Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); rho family, small GTP binding protein Rac1; rho family, small GTP binding protein Rac3; Small G protein
Gene Aliases: AI323801; AI452260; AL023026; D5Ertd559e; EN-7; Gx; HSPC022; MIG5; p21-Rac1; p21-Rac2; Rac; Rac-1; RAC1; Rac1B; RAC2; RAC3; TC-25; TC25
UniProt ID: (Human) P63000, (Human) P15153, (Human) P60763, (Mouse) P63001, (Rat) Q6RUV5, (Mouse) Q9D8X9, (Mouse) P60764
Entrez Gene ID: (Human) 5879, (Human) 5880, (Human) 5881, (Mouse) 19353, (Rat) 363875, (Rat) 366957, (Mouse) 19354, (Mouse) 170758, (Rat) 688319
Molecular Function: small GTPase immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support