Product References
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
ROS-responsive polymer nanoparticles with enhanced loading of dexamethasone effectively modulate the lung injury microenvironment.
Acta biomaterialia
Muhammad W,Zhu J,Zhai Z,Xie J,Zhou J,Feng X,Feng B,Pan Q,Li S,Venkatesan R,Li P,Cao H,Gao C
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
Neuropeptide Y attenuates cardiac remodeling and deterioration of function following myocardial infarction.
Molecular therapy : the journal of the American Society of Gene Therapy
Qin YY,Huang XR,Zhang J,Wu W,Chen J,Wan S,Yu XY,Lan HY
17-5920-82 was used in Flow cytometry/Cell sorting to find that AMI mice lacking NPY developed more severe AMI than WT mice with worse cardiac dysfunction, progressive cardiac inflammation and fibrosis, and excessive apoptosis but impairing angiogenesis.
Wed Feb 02 00:00:00 EST 2022
Nitric oxide controls proliferation of Leishmania major by inhibiting the recruitment of permissive host cells.
Immunity
Formaglio P,Alabdullah M,Siokis A,Handschuh J,Sauerland I,Fu Y,Krone A,Gintschel P,Stettin J,Heyde S,Mohr J,Philipsen L,Schröder A,Robert PA,Zhao G,Khailaie S,Dudeck A,Bertrand J,Späth GF,Kahlfuß S,Bousso P,Schraven B,Huehn J,Binder S,Meyer-Hermann M,Müller AJ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Dec 14 00:00:00 EST 2021
Nitric oxide controls proliferation of Leishmania major by inhibiting the recruitment of permissive host cells.
Immunity
Formaglio P,Alabdullah M,Siokis A,Handschuh J,Sauerland I,Fu Y,Krone A,Gintschel P,Stettin J,Heyde S,Mohr J,Philipsen L,Schröder A,Robert PA,Zhao G,Khailaie S,Dudeck A,Bertrand J,Späth GF,Kahlfuß S,Bousso P,Schraven B,Huehn J,Binder S,Meyer-Hermann M,Müller AJ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Dec 14 00:00:00 EST 2021
Immunogenic Chemotherapy Enhances Recruitment of CAR-T Cells to Lung Tumors and Improves Antitumor Efficacy when Combined with Checkpoint Blockade.
Cancer cell
Srivastava S,Furlan SN,Jaeger-Ruckstuhl CA,Sarvothama M,Berger C,Smythe KS,Garrison SM,Specht JM,Lee SM,Amezquita RA,Voillet V,Muhunthan V,Yechan-Gunja S,Pillai SPS,Rader C,Houghton AM,Pierce RH,Gottardo R,Maloney DG,Riddell SR
17-5920-82 was used in Flow Cytometry to test strategies for enhancing efficacy of CAR-T cells targeting the tumor-associated antigen receptor tyrosine kinase-like orphan receptor 1 (ROR1) infiltrating tumors.
Mon Feb 08 00:00:00 EST 2021
Neutrophil-Macrophage Imbalance Drives the Development of Renal Scarring during Experimental Pyelonephritis.
Journal of the American Society of Nephrology : JASN
Ruiz-Rosado JD,Robledo-Avila F,Cortado H,Rangel-Moreno J,Justice SS,Yang C,Spencer JD,Becknell B,Partida-Sanchez S
17-5920-82 was used in Flow Cytometry to reveal a previously unappreciated outcome for neutrophil-macrophage imbalance in promoting host susceptibility to acute pyelonephritis and the development of permanent renal damage.
Fri Jan 01 00:00:00 EST 2021
AXL Is a Key Factor for Cell Plasticity and Promotes Metastasis in Pancreatic Cancer.
Molecular cancer research : MCR
Du W,Phinney NZ,Huang H,Wang Z,Westcott J,Toombs JE,Zhang Y,Beg MS,Wilkie TM,Lorens JB,Brekken RA
17-5920-82 was used in Western Blot to implicate AXL as a marker of undifferentiated PDA cells and a target for therapy.
Sun Aug 01 00:00:00 EDT 2021
Prostaglandin E3 attenuates macrophage-associated inflammation and prostate tumour growth by modulating polarization.
Journal of cellular and molecular medicine
Cui J,Shan K,Yang Q,Qi Y,Qu H,Li J,Wang R,Jia L,Chen W,Feng N,Chen YQ
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Regulation of the long noncoding RNA XIST on the inflammatory polarization of microglia in cerebral infarction.
Experimental and therapeutic medicine
Zhang M,Yang JK,Ma J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 2021
Computational modeling reveals a key role for polarized myeloid cells in controlling osteoclast activity during bone injury repair.
Scientific reports
Lo CH,Baratchart E,Basanta D,Lynch CC
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Mar 15 00:00:00 EDT 2021
Computational modeling reveals a key role for polarized myeloid cells in controlling osteoclast activity during bone injury repair.
Scientific reports
Lo CH,Baratchart E,Basanta D,Lynch CC
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Mar 15 00:00:00 EDT 2021
Exosomes from human umbilical cord mesenchymal stem cells attenuate the inflammation of severe steroid-resistant asthma by reshaping macrophage polarization.
Stem cell research & therapy
Dong B,Wang C,Zhang J,Zhang J,Gu Y,Guo X,Zuo X,Pan H,Hsu AC,Wang G,Wang F
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Mar 24 00:00:00 EDT 2021
Alphavirus-Driven Interferon Gamma (IFNg) Expression Inhibits Tumor Growth in Orthotopic 4T1 Breast Cancer Model.
Vaccines
Trofimova O,Korotkaja K,Skrastina D,Jansons J,Spunde K,Isaguliants M,Zajakina A
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Oct 27 00:00:00 EDT 2021
PD-L1 Improves Motor Function and Alleviates Neuropathic Pain in Male Mice After Spinal Cord Injury by Inhibiting MAPK Pathway.
Frontiers in immunology
Kong F,Sun K,Zhu J,Li F,Lin F,Sun X,Luo X,Ren C,Lu L,Zhao S,Sun J,Wang Y,Shi J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
TREM2 is a receptor for non-glycosylated mycolic acids of mycobacteria that limits anti-mycobacterial macrophage activation.
Nature communications
Iizasa E,Chuma Y,Uematsu T,Kubota M,Kawaguchi H,Umemura M,Toyonaga K,Kiyohara H,Yano I,Colonna M,Sugita M,Matsuzaki G,Yamasaki S,Yoshida H,Hara H
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Macrophages Expressing GALC Improve Peripheral Krabbe Disease by a Mechanism Independent of Cross-Correction.
Neuron
Weinstock NI,Shin D,Dhimal N,Hong X,Irons EE,Silvestri NJ,Reed CB,Nguyen D,Sampson O,Cheng YC,Lau JTY,Bongarzone ER,Kofler J,Escolar ML,Gelb MH,Wrabetz L,Feltri ML
17-5920 was used in Flow cytometry/Cell sorting to show that cross-correction does not occur efficiently in vivo and that Galc-deficient Schwann cells autonomously produce psychosine.
Wed Jul 08 00:00:00 EDT 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
Astrocytes promote a protective immune response to brain Toxoplasma gondii infection via IL-33-ST2 signaling.
PLoS pathogens
Still KM,Batista SJ,O'Brien CA,Oyesola OO,Früh SP,Webb LM,Smirnov I,Kovacs MA,Cowan MN,Hayes NW,Thompson JA,Tait Wojno ED,Harris TH
17-5920 was used in Flow cytometry/Cell sorting to show that IL-33 is expressed by oligodendrocytes and astrocytes during T. gondii infection, is released locally into the cerebrospinal fluid of T. gondii-infected animals, and is required for control of infection.
Thu Oct 01 00:00:00 EDT 2020
Anticancer Activity of Liquid Treated with Microwave Plasma-Generated Gas through Macrophage Activation.
Oxidative medicine and cellular longevity
Lee CB,Seo IH,Chae MW,Park JW,Choi EH,Uhm HS,Baik KY
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Mycobacterium tuberculosis-infected alveolar epithelial cells modulate dendritic cell function through the HIF-1α-NOS2 axis.
Journal of leukocyte biology
Rodrigues TS,Alvarez ARP,Gembre AF,Forni MFPAD,de Melo BMS,Alves Filho JCF,Câmara NOS,Bonato VLD
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Enhanced alleviation of aGVHD by TGF-β1-modified mesenchymal stem cells in mice through shifting MΦ into M2 phenotype and promoting the differentiation of Treg cells.
Journal of cellular and molecular medicine
Wu R,Liu C,Deng X,Chen L,Hao S,Ma L
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Proimmunogenic impact of MEK inhibition synergizes with agonist anti-CD40 immunostimulatory antibodies in tumor therapy.
Nature communications
Baumann D,Hägele T,Mochayedi J,Drebant J,Vent C,Blobner S,Noll JH,Nickel I,Schumacher C,Boos SL,Daniel AS,Wendler S,Volkmar M,Strobel O,Offringa R
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Proimmunogenic impact of MEK inhibition synergizes with agonist anti-CD40 immunostimulatory antibodies in tumor therapy.
Nature communications
Baumann D,Hägele T,Mochayedi J,Drebant J,Vent C,Blobner S,Noll JH,Nickel I,Schumacher C,Boos SL,Daniel AS,Wendler S,Volkmar M,Strobel O,Offringa R
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
CCL2 and CXCL12 Derived from Mesenchymal Stromal Cells Cooperatively Polarize IL-10+ Tissue Macrophages to Mitigate Gut Injury.
Cell reports
Giri J,Das R,Nylen E,Chinnadurai R,Galipeau J
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue Feb 11 00:00:00 EST 2020
Select autophagy genes maintain quiescence of tissue-resident macrophages and increase susceptibility to Listeria monocytogenes.
Nature microbiology
Wang YT,Zaitsev K,Lu Q,Li S,Schaiff WT,Kim KW,Droit L,Wilen CB,Desai C,Balce DR,Orchard RC,Orvedahl A,Park S,Kreamalmeyer D,Handley SA,Pfeifer JD,Baldridge MT,Artyomov MN,Stallings CL,Virgin HW
17-5920 was used in Flow cytometry/Cell sorting to delineate a function of the autophagy proteins Beclin 1 and FIP200-but not of other essential autophagy components ATG5, ATG16L1 or ATG7-in mediating quiescence of tissue-resident macrophages by limiting the effects of systemic interferon-γ.
Sat Feb 01 00:00:00 EST 2020
Inducing cancer indolence by targeting mitochondrial Complex I is potentiated by blocking macrophage-mediated adaptive responses.
Nature communications
Kurelac I,Iommarini L,Vatrinet R,Amato LB,De Luise M,Leone G,Girolimetti G,Umesh Ganesh N,Bridgeman VL,Ombrato L,Columbaro M,Ragazzi M,Gibellini L,Sollazzo M,Feichtinger RG,Vidali S,Baldassarre M,Foriel S,Vidone M,Cossarizza A,Grifoni D,Kofler B,Malanchi I,Porcelli AM,Gasparre G
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Feb 22 00:00:00 EST 2019
Irg1 expression in myeloid cells prevents immunopathology during M. tuberculosis infection.
The Journal of experimental medicine
Nair S,Huynh JP,Lampropoulou V,Loginicheva E,Esaulova E,Gounder AP,Boon ACM,Schwarzkopf EA,Bradstreet TR,Edelson BT,Artyomov MN,Stallings CL,Diamond MS
17-5920 was used in Flow cytometry/Cell sorting to study the role of Immune responsive gene 1 in the modulation of inflammation during Mycobacterium tuberculosis infection.
Mon Apr 02 00:00:00 EDT 2018
Chloroquine modulates antitumor immune response by resetting tumor-associated macrophages toward M1 phenotype.
Nature communications
Chen D,Xie J,Fiskesund R,Dong W,Liang X,Lv J,Jin X,Liu J,Mo S,Zhang T,Cheng F,Zhou Y,Zhang H,Tang K,Ma J,Liu Y,Huang B
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Wed Feb 28 00:00:00 EST 2018
Oncostatin M is overexpressed in skin squamous-cell carcinoma and promotes tumor progression.
Oncotarget
Simonneau M,Frouin E,Huguier V,Jermidi C,Jégou JF,Godet J,Barra A,Paris I,Levillain P,Cordier-Dirikoc S,Pedretti N,Bernard FX,Lecron JC,Morel F,Favot L
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Nov 23 00:00:00 EST 2018
Oncostatin M is overexpressed in skin squamous-cell carcinoma and promotes tumor progression.
Oncotarget
Simonneau M,Frouin E,Huguier V,Jermidi C,Jégou JF,Godet J,Barra A,Paris I,Levillain P,Cordier-Dirikoc S,Pedretti N,Bernard FX,Lecron JC,Morel F,Favot L
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Nov 23 00:00:00 EST 2018
Mitochondrial permeabilization engages NF-κB-dependent anti-tumour activity under caspase deficiency.
Nature cell biology
Giampazolias E,Zunino B,Dhayade S,Bock F,Cloix C,Cao K,Roca A,Lopez J,Ichim G,Proïcs E,Rubio-Patiño C,Fort L,Yatim N,Woodham E,Orozco S,Taraborrelli L,Peltzer N,Lecis D,Machesky L,Walczak H,Albert ML,Milling S,Oberst A,Ricci JE,Ryan KM,Blyth K,Tait SWG
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2017
The pattern recognition receptor, Mincle, is essential for maintaining the M1 macrophage phenotype in acute renal inflammation.
Kidney international
Lv LL,Tang PM,Li CJ,You YK,Li J,Huang XR,Ni J,Feng M,Liu BC,Lan HY
17-5920-82 was used in Flow cytometry/Cell sorting to investigate the role of Mincle (macrophage-inducible C-type lectin, Clec4e) in kidney disease.
Wed Mar 01 00:00:00 EST 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
TGF-β receptor 1 inhibition prevents stenosis of tissue-engineered vascular grafts by reducing host mononuclear phagocyte activation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Lee YU,de Dios Ruiz-Rosado J,Mahler N,Best CA,Tara S,Yi T,Shoji T,Sugiura T,Lee AY,Robledo-Avila F,Hibino N,Pober JS,Shinoka T,Partida-Sanchez S,Breuer CK
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
TNF-Mediated Restriction of Arginase 1 Expression in Myeloid Cells Triggers Type 2 NO Synthase Activity at the Site of Infection.
Cell reports
Schleicher U,Paduch K,Debus A,Obermeyer S,König T,Kling JC,Ribechini E,Dudziak D,Mougiakakos D,Murray PJ,Ostuni R,Körner H,Bogdan C
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Tue May 03 00:00:00 EDT 2016
Dendritic cell-derived nitric oxide inhibits the differentiation of effector dendritic cells.
Oncotarget
Si C,Zhang R,Wu T,Lu G,Hu Y,Zhang H,Xu F,Wei P,Chen K,Tang H,Yeretssian G,Xiong H
17-5920 was used in Flow cytometry/Cell sorting to demonstrate that DC-derived nitric oxide restrains effector DC development to treat autoimmune and inflammatory diseases.
Tue Nov 15 00:00:00 EST 2016
Commensal Gram-positive bacteria initiates colitis by inducing monocyte/macrophage mobilization.
Mucosal immunology
Nakanishi Y,Sato T,Ohteki T
Thu Jan 01 00:00:00 EST 2015
Expression of Cationic Amino Acid Transporter 2 Is Required for Myeloid-Derived Suppressor Cell-Mediated Control of T Cell Immunity.
Journal of immunology (Baltimore, Md. : 1950)
Cimen Bozkus C,Elzey BD,Crist SA,Ellies LG,Ratliff TL
17-5920 was used in Flow cytometry/Cell sorting to investigate whether restricting myeloid-derived suppressor cell uptake of l-Arginine is a critical control point to modulate their suppressor activity.
Tue Dec 01 00:00:00 EST 2015
Maternal immune activation evoked by polyinosinic:polycytidylic acid does not evoke microglial cell activation in the embryo.
Frontiers in cellular neuroscience
Smolders S,Smolders SM,Swinnen N,Gärtner A,Rigo JM,Legendre P,Brône B
Published figure using iNOS monoclonal antibody (Product # 17-5920-82) in Flow Cytometry
Mon Aug 24 00:00:00 EDT 2015
Myeloid cell-derived inducible nitric oxide synthase suppresses M1 macrophage polarization.
Nature communications
Lu G,Zhang R,Geng S,Peng L,Jayaraman P,Chen C,Xu F,Yang J,Li Q,Zheng H,Shen K,Wang J,Liu X,Wang W,Zheng Z,Qi CF,Si C,He JC,Liu K,Lira SA,Sikora AG,Li L,Xiong H
17-5920 was used in Flow cytometry/Cell sorting to show that iNOS-deficient mice display enhanced classically activated M1 macrophage polarization without major effects on alternatively activated M2 macrophages.
Fri Mar 27 00:00:00 EDT 2015