Product References
Trehalose induces autophagy via lysosomal-mediated TFEB activation in models of motoneuron degeneration.
Autophagy
Rusmini P,Cortese K,Crippa V,Cristofani R,Cicardi ME,Ferrari V,Vezzoli G,Tedesco B,Meroni M,Messi E,Piccolella M,Galbiati M,Garrè M,Morelli E,Vaccari T,Poletti A
Mon Apr 01 00:00:00 EDT 2019
TGFβi is involved in the chondrogenic differentiation of mesenchymal stem cells and is dysregulated in osteoarthritis.
Osteoarthritis and cartilage
Ruiz M,Maumus M,Fonteneau G,Pers YM,Ferreira R,Dagneaux L,Delfour C,Houard X,Berenbaum F,Rannou F,Jorgensen C,Noël D
Fri Mar 01 00:00:00 EST 2019
Biomimetic electrical stimulation induces rat bone marrow mesenchymal stem cells to differentiate into cardiomyocyte-like cells via TGF-beta 1 in vitro.
Progress in biophysics and molecular biology
He X,Li L,Tang M,Zeng Y,Li H,Yu X
Fri Nov 01 00:00:00 EDT 2019
Inhibition of tumor-promoting stroma to enforce subsequently targeting AT1R on tumor cells by pathological inspired micelles.
Biomaterials
Zhu Y,Wen L,Shao S,Tan Y,Meng T,Yang X,Liu Y,Liu X,Yuan H,Hu F
Sun Apr 01 00:00:00 EDT 2018
Combination antitumor immunotherapy with VEGF and PIGF siRNA via systemic delivery of multi-functionalized nanoparticles to tumor-associated macrophages and breast cancer cells.
Biomaterials
Song Y,Tang C,Yin C
Sat Dec 01 00:00:00 EST 2018
Total glucosides of paeony improves the immunomodulatory capacity of MSCs partially via the miR-124/STAT3 pathway in oral lichen planus.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhao Z,Han Y,Zhang Z,Li W,Ji X,Liu X,Jin J,Xu S,Cui H,Cheng Z,Wang Q,Wang X,Guo X,Wang Y,Liu H
Sat Sep 01 00:00:00 EDT 2018
HOXC8 promotes proliferation and migration through transcriptional up-regulation of TGFβ1 in non-small cell lung cancer.
Oncogenesis
Liu H,Zhang M,Xu S,Zhang J,Zou J,Yang C,Zhang Y,Gong C,Kai Y,Li Y
Wed Jan 17 00:00:00 EST 2018
Hepatoprotection of auraptene from peels of citrus fruits against thioacetamide-induced hepatic fibrosis in mice by activating farnesoid X receptor.
Food & function
Gao X,Wang C,Ning C,Liu K,Wang X,Liu Z,Sun H,Ma X,Sun P,Meng Q
Wed May 23 00:00:00 EDT 2018
Quercetin ameliorates kidney injury and fibrosis by modulating M1/M2 macrophage polarization.
Biochemical pharmacology
Lu H,Wu L,Liu L,Ruan Q,Zhang X,Hong W,Wu S,Jin G,Bai Y
Wed Aug 01 00:00:00 EDT 2018
Photobiomodulation therapy promotes neurogenesis by improving post-stroke local microenvironment and stimulating neuroprogenitor cells.
Experimental neurology
Yang L,Tucker D,Dong Y,Wu C,Lu Y,Li Y,Zhang J,Liu TC,Zhang Q
Mon Jan 01 00:00:00 EST 2018
Regulation of TLR4 expression mediates the attenuating effect of erythropoietin on inflammation and myocardial fibrosis in rat heart.
International journal of molecular medicine
Liu F,Wen Y,Kang J,Wei C,Wang M,Zheng Z,Peng J
Sat Sep 01 00:00:00 EDT 2018
Knockdown of NLRP3 alleviates high glucose or TGFB1-induced EMT in human renal tubular cells.
Journal of molecular endocrinology
Song S,Qiu D,Luo F,Wei J,Wu M,Wu H,Du C,Du Y,Ren Y,Chen N,Duan H,Shi Y
Mon Oct 01 00:00:00 EDT 2018
NLRP3 deficiency ameliorates renal inflammation and fibrosis in diabetic mice.
Molecular and cellular endocrinology
Wu M,Han W,Song S,Du Y,Liu C,Chen N,Wu H,Shi Y,Duan H
Sat Dec 15 00:00:00 EST 2018
NLRP3 deficiency ameliorates renal inflammation and fibrosis in diabetic mice.
Molecular and cellular endocrinology
Wu M,Han W,Song S,Du Y,Liu C,Chen N,Wu H,Shi Y,Duan H
Sat Dec 15 00:00:00 EST 2018
Protective effects of dioscin against fructose-induced renal damage via adjusting Sirt3-mediated oxidative stress, fibrosis, lipid metabolism and inflammation.
Toxicology letters
Qiao Y,Xu L,Tao X,Yin L,Qi Y,Xu Y,Han X,Tang Z,Ma X,Liu K,Peng J
Thu Mar 01 00:00:00 EST 2018
Baicalin inhibits pressure overload-induced cardiac fibrosis through regulating AMPK/TGF-β/Smads signaling pathway.
Archives of biochemistry and biophysics
Xiao Y,Ye J,Zhou Y,Huang J,Liu X,Huang B,Zhu L,Wu B,Zhang G,Cai Y
Thu Feb 15 00:00:00 EST 2018
Effects of garlic polysaccharide on alcoholic liver fibrosis and intestinal microflora in mice.
Pharmaceutical biology
Wang Y,Guan M,Zhao X,Li X
Sat Dec 01 00:00:00 EST 2018
Intrinsic renal cells induce lymphocytosis of Th22 cells from IgA nephropathy patients through B7-CTLA-4 and CCL-CCR pathways.
Molecular and cellular biochemistry
Gan L,Zhou Q,Li X,Chen C,Meng T,Pu J,Zhu M,Xiao C
Sun Apr 01 00:00:00 EDT 2018
Role of TGF-β1/Smad3-mediated fibrosis in drug resistance mechanism of prolactinoma.
Brain research
Hu B,Mao Z,Jiang X,He D,Wang Z,Wang X,Zhu Y,Wang H
Thu Nov 01 00:00:00 EDT 2018
Role of TGF-β1/Smad3-mediated fibrosis in drug resistance mechanism of prolactinoma.
Brain research
Hu B,Mao Z,Jiang X,He D,Wang Z,Wang X,Zhu Y,Wang H
Thu Nov 01 00:00:00 EDT 2018
Angiotensin II upregulates fibroblast-myofibroblast transition through Cx43-dependent CaMKII and TGF-β1 signaling in neonatal rat cardiac fibroblasts.
Acta biochimica et biophysica Sinica
Cao L,Chen Y,Lu L,Liu Y,Wang Y,Fan J,Yin Y
Sat Sep 01 00:00:00 EDT 2018
Downregulation of Glt25d1 aggravates carbon tetrachloride‑induced acute hepatic injury through activation of the TGF‑β1/Smad2 signaling pathway.
Molecular medicine reports
Ye X,He L,Ma J,Li Y,Zhang M,Yang J,Zhang J,Xiao F,Wei H
Mon Oct 01 00:00:00 EDT 2018
Antifibrotic Agent Pirfenidone Protects against Development of Radiation-Induced Pulmonary Fibrosis in a Murine Model.
Radiation research
Qin W,Liu B,Yi M,Li L,Tang Y,Wu B,Yuan X
Mon Oct 01 00:00:00 EDT 2018
Glucagon-Like Peptide-1 Mediates the Protective Effect of the Dipeptidyl Peptidase IV Inhibitor on Renal Fibrosis via Reducing the Phenotypic Conversion of Renal Microvascular Cells in Monocrotaline-Treated Rats.
BioMed research international
Xu J,Wang J,Cheng Y,Li X,He M,Zhu J,Han H,Wei G,Kong H,Xie W,Wang H,Zuo X
Wed Sep 05 00:00:00 EDT 2018
Gossypol Promotes Bone Formation in Ovariectomy-Induced Osteoporosis through Regulating Cell Apoptosis.
BioMed research international
Liang J,Chen C,Liu H,Liu X,Li Z,Hu J,Zhao H
Wed Apr 24 00:00:00 EDT 2019
Acteoside relieves mesangial cell injury by regulating Th22 cell chemotaxis and proliferation in IgA nephropathy.
Renal failure
Gan L,Li X,Zhu M,Chen C,Luo H,Zhou Q
Thu Nov 01 00:00:00 EDT 2018
Overexpression of YOD1 Promotes the Migration of Human Oral Keratinocytes by Enhancing TGF-β3 Signaling.
Biomedical and environmental sciences : BES
Ju Q,Li MX,Chen G,Wang HX,Shi QM,Ge X,Ding Z,Wang Q,Xu LC
Sun Jul 01 00:00:00 EDT 2018
EDIL3 depletion suppress epithelial-mesenchymal transition of lens epithelial cells via transforming growth factor β pathway.
International journal of ophthalmology
Zhang R,Wei YH,Zhao CY,Song HY,Shen N,Cui X,Gao X,Qi ZT,Zhong M,Shen W
Wed Sep 30 00:00:00 EDT 2020
Overexpression of zinc-α2-glycoprotein suppressed seizures and seizure-related neuroflammation in pentylenetetrazol-kindled rats.
Journal of neuroinflammation
Liu Y,Wang T,Liu X,Wen Y,Xu T,Yu X,Wei X,Ding X,Mo L,Yin M,Tan X,Chen L
Thu Mar 22 00:00:00 EDT 2018
LincRNA 1700020I14Rik alleviates cell proliferation and fibrosis in diabetic nephropathy via miR-34a-5p/Sirt1/HIF-1α signaling.
Cell death & disease
Li A,Peng R,Sun Y,Liu H,Peng H,Zhang Z
Tue May 01 00:00:00 EDT 2018
LincRNA 1700020I14Rik alleviates cell proliferation and fibrosis in diabetic nephropathy via miR-34a-5p/Sirt1/HIF-1α signaling.
Cell death & disease
Li A,Peng R,Sun Y,Liu H,Peng H,Zhang Z
Tue May 01 00:00:00 EDT 2018
The Role of Alpha-Lipoic Acid in the Pathomechanism of Acute Ischemic Stroke.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Q,Lv C,Sun Y,Han X,Wang S,Mao Z,Xin Y,Zhang B
Tue Sep 04 00:00:00 EDT 2018
microRNA-21 regulates astrocytic reaction post-acute phase of spinal cord injury through modulating TGF-β signaling.
Aging
Liu R,Wang W,Wang S,Xie W,Li H,Ning B
Sat Jun 23 00:00:00 EDT 2018
Ursolic Acid Attenuates High Glucose-Mediated Mesangial Cell Injury by Inhibiting the Phosphatidylinositol 3-Kinase/Akt/Mammalian Target of Rapamycin (PI3K/Akt/mTOR) Signaling Pathway.
Medical science monitor : international medical journal of experimental and clinical research
Wang EM,Fan QL,Yue Y,Xu L
Sun Feb 11 00:00:00 EST 2018
Study on Cardiotoxicity and Mechanism of "Fuzi" Extracts Based on Metabonomics.
International journal of molecular sciences
Huang G,Yang L,Zhou W,Tang X,Wang Y,Ma Z,Gao S,Gao Y
Wed Nov 07 00:00:00 EST 2018
Study on Cardiotoxicity and Mechanism of "Fuzi" Extracts Based on Metabonomics.
International journal of molecular sciences
Huang G,Yang L,Zhou W,Tang X,Wang Y,Ma Z,Gao S,Gao Y
Wed Nov 07 00:00:00 EST 2018
Ambient fine particulate matter exposure induces reversible cardiac dysfunction and fibrosis in juvenile and older female mice.
Particle and fibre toxicology
Qin G,Xia J,Zhang Y,Guo L,Chen R,Sang N
Mon Jun 25 00:00:00 EDT 2018
FSTL1 interacts with VIM and promotes colorectal cancer metastasis via activating the focal adhesion signalling pathway.
Cell death & disease
Gu C,Wang X,Long T,Wang X,Zhong Y,Ma Y,Hu Z,Li Z
Tue May 29 00:00:00 EDT 2018
Renal Denervation Effects on Myocardial Fibrosis and Ventricular Arrhythmias in Rats with Ischemic Cardiomyopathy.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zhang B,Li X,Chen C,Jiang W,Lu D,Liu Q,Wang K,Yan Y,Jiang Z,Geng J,Xu H,Shan Q
Fri Aug 17 00:00:00 EDT 2018
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Cardioprotection of CAPE-oNO2 against myocardial ischemia/reperfusion induced ROS generation via regulating the SIRT1/eNOS/NF-κB pathway in vivo and in vitro.
Redox biology
Li D,Wang X,Huang Q,Li S,Zhou Y,Li Z
Tue May 01 00:00:00 EDT 2018
Emodin Alleviates Sodium Taurocholate-Induced Pancreatic Acinar Cell Injury via MicroRNA-30a-5p-Mediated Inhibition of High-Temperature Requirement A/Transforming Growth Factor Beta 1 Inflammatory Signaling.
Frontiers in immunology
Xiang H,Tao X,Xia S,Qu J,Song H,Liu J,Shang D
Wed Nov 20 00:00:00 EST 2019
Anthocyanins Delay Ageing-Related Degenerative Changes in the Liver.
Plant foods for human nutrition (Dordrecht, Netherlands)
Wei J,Zhang G,Zhang X,Xu D,Gao J,Fan J
Fri Dec 01 00:00:00 EST 2017
TGF-β1/Smad3 Signaling Pathway Mediates T-2 Toxin-Induced Decrease of Type II Collagen in Cultured Rat Chondrocytes.
Toxins
Li Y,Zou N,Wang J,Wang KW,Li FY,Chen FX,Sun BY,Sun DJ
Sun Nov 05 00:00:00 EDT 2017
Exosomes secreted by human adipose mesenchymal stem cells promote scarless cutaneous repair by regulating extracellular matrix remodelling.
Scientific reports
Wang L,Hu L,Zhou X,Xiong Z,Zhang C,Shehada HMA,Hu B,Song J,Chen L
Tue Oct 17 00:00:00 EDT 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-κB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
Metformin reverses prostate cancer resistance to enzalutamide by targeting TGF-β1/STAT3 axis-regulated EMT.
Cell death & disease
Liu Q,Tong D,Liu G,Xu J,Do K,Geary K,Zhang D,Zhang J,Zhang Y,Li Y,Bi G,Lan W,Jiang J
Thu Aug 24 00:00:00 EDT 2017
Metformin reverses prostate cancer resistance to enzalutamide by targeting TGF-β1/STAT3 axis-regulated EMT.
Cell death & disease
Liu Q,Tong D,Liu G,Xu J,Do K,Geary K,Zhang D,Zhang J,Zhang Y,Li Y,Bi G,Lan W,Jiang J
Thu Aug 24 00:00:00 EDT 2017
TGFβ1 Promotes Gemcitabine Resistance through Regulating the LncRNA-LET/NF90/miR-145 Signaling Axis in Bladder Cancer.
Theranostics
Zhuang J,Shen L,Yang L,Huang X,Lu Q,Cui Y,Zheng X,Zhao X,Zhang D,Huang R,Guo H,Yan J
Mon Apr 30 00:00:00 EDT 2018
Let‑7a suppresses cell proliferation via the TGF‑β/SMAD signaling pathway in cervical cancer.
Oncology reports
Wu T,Chen X,Peng R,Liu H,Yin P,Peng H,Zhou Y,Sun Y,Wen L,Yi H,Li A,Zhang Z
Thu Dec 01 00:00:00 EST 2016