Product References
Niche-specific MHC II and PD-L1 regulate CD4+CD8αα+ intraepithelial lymphocyte differentiation.
The Journal of experimental medicine
Moon S,Park Y,Hyeon S,Kim YM,Kim JH,Kim H,Park S,Lee KJ,Koo BK,Ha SJ,Lee SW
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Mon Apr 05 00:00:00 EDT 2021
CCL17-expressing dendritic cells in the intestine are preferentially infected by Salmonella but CCL17 plays a redundant role in systemic dissemination.
Immunity, inflammation and disease
Erazo AB,Wang N,Standke L,Semeniuk AD,Fülle L,Cengiz SC,Thiem M,Weighardt H,Strugnell RA,Förster I
25-5961-82 was used in Flow Cytometry to study the role of CCL17 during the early phase of infection, including bacterial spreading from the intestine to the draining lymph nodes.
Wed Sep 01 00:00:00 EDT 2021
Physical restraint mouse models to assess immune responses under stress with or without habituation.
STAR protocols
Ding JX,Rudak PT,Inoue W,Haeryfar SMM
25-5961-82 was used in Flow Cytometry to provide protocols and guidelines in using three mouse restraint models, namely prolonged restraint stress, repeated restraint stress, and chronic variable stress, to examine immunological homeostasis/competence, or lack thereof, under stress with or without habituation.
Fri Dec 17 00:00:00 EST 2021
Dynamic regulation of chromatin organizer SATB1 via TCR-induced alternative promoter switch during T-cell development.
Nucleic acids research
Patta I,Madhok A,Khare S,Gottimukkala KP,Verma A,Giri S,Dandewad V,Seshadri V,Lal G,Misra-Sen J,Galande S
25-5961 was used in Flow cytometry/Cell sorting to study the mechanism regulating expression of SATB1 during T-cell development.
Fri Jun 19 00:00:00 EDT 2020
Diversity in medullary thymic epithelial cells controls the activity and availability of iNKT cells.
Nature communications
Lucas B,White AJ,Cosway EJ,Parnell SM,James KD,Jones ND,Ohigashi I,Takahama Y,Jenkinson WE,Anderson G
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Mon May 04 00:00:00 EDT 2020
Requirements for the differentiation of innate T-bethigh memory-phenotype CD4+ T lymphocytes under steady state.
Nature communications
Kawabe T,Yi J,Kawajiri A,Hilligan K,Fang D,Ishii N,Yamane H,Zhu J,Jankovic D,Kim KS,Trinchieri G,Sher A
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2020
B7-CD28 co-stimulation modulates central tolerance via thymic clonal deletion and Treg generation through distinct mechanisms.
Nature communications
Watanabe M,Lu Y,Breen M,Hodes RJ
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Dietary Glucose Consumption Promotes RALDH Activity in Small Intestinal CD103+CD11b+ Dendritic Cells.
Frontiers in immunology
Ko HJ,Hong SW,Verma R,Jung J,Lee M,Kim N,Kim D,Surh CD,Kim KS,Rudra D,Im SH
25-5961 was used in Flow cytometry/Cell sorting to reveal previously underappreciated role of dietary glucose concentration in establishing regulatory properties in intestinal DCs, thereby extending a potential therapeutic module against intestinal inflammation.
Mon Apr 19 00:00:00 EDT 2021
CD160 serves as a negative regulator of NKT cells in acute hepatic injury.
Nature communications
Kim TJ,Park G,Kim J,Lim SA,Kim J,Im K,Shin MH,Fu YX,Del Rio ML,Rodriguez-Barbosa JI,Yee C,Suh KS,Kim SJ,Ha SJ,Lee KM
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Mon Jul 22 00:00:00 EDT 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
STAT1 is a sex-specific tumor suppressor in colitis-associated colorectal cancer.
Molecular oncology
Crnčec I,Modak M,Gordziel C,Svinka J,Scharf I,Moritsch S,Pathria P,Schlederer M,Kenner L,Timelthaler G,Müller M,Strobl B,Casanova E,Bayer E,Mohr T,Stöckl J,Friedrich K,Eferl R
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
A Paradoxical Tumor-Suppressor Role for the Rac1 Exchange Factor Vav1 in T Cell Acute Lymphoblastic Leukemia.
Cancer cell
Robles-Valero J,Lorenzo-Martín LF,Menacho-Márquez M,Fernández-Pisonero I,Abad A,Camós M,Toribio ML,Espinosa L,Bigas A,Bustelo XR
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Mon Nov 13 00:00:00 EST 2017
1810011o10 Rik Inhibits the Antitumor Effect of Intratumoral CD8+ T Cells through Suppression of Notch2 Pathway in a Murine Hepatocellular Carcinoma Model.
Frontiers in immunology
Dai K,Huang L,Huang YB,Chen ZB,Yang LH,Jiang YA
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Hypoxia-Inducible Factor-2α Limits Natural Killer T Cell Cytotoxicity in Renal Ischemia/Reperfusion Injury.
Journal of the American Society of Nephrology : JASN
Zhang J,Han C,Dai H,Hou J,Dong Y,Cui X,Xu L,Zhang M,Xia Q
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Environmental and Genetic Activation of Hypothalamic BDNF Modulates T-cell Immunity to Exert an Anticancer Phenotype.
Cancer immunology research
Xiao R,Bergin SM,Huang W,Slater AM,Liu X,Judd RT,Lin ED,Widstrom KJ,Scoville SD,Yu J,Caligiuri MA,Cao L
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Protein kinase D regulates positive selection of CD4+ thymocytes through phosphorylation of SHP-1.
Nature communications
Ishikawa E,Kosako H,Yasuda T,Ohmuraya M,Araki K,Kurosaki T,Saito T,Yamasaki S
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Tue Sep 27 00:00:00 EDT 2016
Cell Type Preference of a Novel Human Derived Cell-Permeable Peptide dNP2 and TAT in Murine Splenic Immune Cells.
PloS one
Lim S,Lee JA,Koo JH,Kang TG,Ha SJ,Choi JM
25-5961 was used in Flow cytometry/Cell sorting to demonstrate that uptake of cell-permeable peptides differs between phagocytic and non-phagocytic immune cells.
Tue Jul 11 00:00:00 EDT 2017
Enhanced Immune Response in Immunodeficient Mice Improves Peripheral Nerve Regeneration Following Axotomy.
Frontiers in cellular neuroscience
Bombeiro AL,Santini JC,Thomé R,Ferreira ER,Nunes SL,Moreira BM,Bonet IJ,Sartori CR,Verinaud L,Oliveira AL
25-5961 was used in Flow cytometry/Cell sorting to investigate the contribution of lymphocytes during peripheral nerve regeneration in immunocompetent versus immunodeficient mice.
Wed Jul 06 00:00:00 EDT 2016
MHC-mismatched mixed chimerism augments thymic regulatory T-cell production and prevents relapse of EAE in mice.
Proceedings of the National Academy of Sciences of the United States of America
Wu L,Li N,Zhang M,Xue SL,Cassady K,Lin Q,Riggs AD,Zeng D
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Tue Dec 29 00:00:00 EST 2015
Antibiotics in neonatal life increase murine susceptibility to experimental psoriasis.
Nature communications
Zanvit P,Konkel JE,Jiao X,Kasagi S,Zhang D,Wu R,Chia C,Ajami NJ,Smith DP,Petrosino JF,Abbatiello B,Nakatsukasa H,Chen Q,Belkaid Y,Chen ZJ,Chen W
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2015
Administration of anti-CD20 mAb is highly effective in preventing but ineffective in treating chronic graft-versus-host disease while preserving strong graft-versus-leukemia effects.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Johnston HF,Xu Y,Racine JJ,Cassady K,Ni X,Wu T,Chan A,Forman S,Zeng D
25-5961 was used in Flow cytometry/Cell sorting to indicate that administration of anti-CD20 mAb before signs of cGVHD can prevent induction of autoimmune-like cGVHD while preserving a GVL effect.
Fri Aug 01 00:00:00 EDT 2014
Autophagy is a critical regulator of memory CD8(+) T cell formation.
eLife
Puleston DJ,Zhang H,Powell TJ,Lipina E,Sims S,Panse I,Watson AS,Cerundolo V,Townsend AR,Klenerman P,Simon AK
25-5961 was used in Flow cytometry/Cell sorting to show that mice lacking the autophagy gene Atg7 in T cells failed to establish CD8(+) T cell memory to influenza and MCMV infection.
Tue Nov 11 00:00:00 EST 2014
Mutual expression of the transcription factors Runx3 and ThPOK regulates intestinal CD4⁺ T cell immunity.
Nature immunology
Reis BS,Rogoz A,Costa-Pinto FA,Taniuchi I,Mucida D
Published figure using TCR beta monoclonal antibody (Product # 25-5961-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Blockade of invariant TCR-CD1d interaction specifically inhibits antibody production against blood group A carbohydrates.
Blood
Tazawa H,Irei T,Tanaka Y,Igarashi Y,Tashiro H,Ohdan H
25-5961 was used in Flow cytometry/Cell sorting to elucidate the role of CD1d-restricted natural killer T cells and IL-5 in the production of antibodies against blood group A carbohydrates.
Thu Oct 10 00:00:00 EDT 2013
γδ T cells are essential effectors of type 1 diabetes in the nonobese diabetic mouse model.
Journal of immunology (Baltimore, Md. : 1950)
Markle JG,Mortin-Toth S,Wong AS,Geng L,Hayday A,Danska JS
25-5961 was used in Flow cytometry/Cell sorting to elucidate the role of various subsets of γδ T cells in a murine model for autoimmune disease.
Sat Jun 01 00:00:00 EDT 2013
Additive and global functions of HoxA cluster genes in mesoderm derivatives.
Developmental biology
Di-Poï N,Koch U,Radtke F,Duboule D
25-5961 was used in Flow cytometry/Cell sorting to report that mutant mice lacking the entire HoxA cluster in mesodermal lineages display the expected spectrum of postnatal respiratory, cardiac and urogenital defects, previously reported for single gene mutations.
Sat May 15 00:00:00 EDT 2010