Product References
Alleviative Effect of Lactoferrin Interventions Against the Hepatotoxicity Induced by Titanium Dioxide Nanoparticles.
Biological trace element research
Zhang W,Sun J,Liu F,Li S,Wang X,Su L,Liu G
Thu Feb 01 00:00:00 EST 2024
Luteolin alleviated neutrophilic asthma by inhibiting IL-36γ secretion-mediated MAPK pathways.
Pharmaceutical biology
Qiao XR,Feng T,Zhang D,Zhi LL,Zhang JT,Liu XF,Pan Y,Xu JW,Cui WJ,Dong L
Fri Dec 01 00:00:00 EST 2023
BMP7 Attenuates Neuroinflammation after Spinal Cord Injury by Suppressing the Microglia Activation and Inducing Microglial Polarization Via the STAT3 Pathway.
Neurochemical research
Wei X,Huang C,Chen K,Liu S,Wang M,Yang L,Wang Y
Fri Sep 01 00:00:00 EDT 2023
Suppression of NLRP3/Caspase-1/GSDMD Mediated Corneal Epithelium Pyroptosis Using Melatonin-Loaded Liposomes to Inhibit Benzalkonium Chloride-Induced Dry Eye Disease.
International journal of nanomedicine
Lou Q,Pan L,Xiang S,Li Y,Jin J,Tan J,Huang B,Nan K,Lin S
Thu May 18 00:00:00 EDT 2023
Ang-(1-7)/MasR axis promotes functional recovery after spinal cord injury by regulating microglia/macrophage polarization.
Cell & bioscience
Gu G,Zhu B,Ren J,Song X,Fan B,Ding H,Shang J,Wu H,Li J,Wang H,Li J,Wei Z,Feng S
Sat Feb 04 00:00:00 EST 2023
EEF1A2 accelerates the protein translation of chemokine in rat myocardial cells induced by ischemia-reperfusion.
Heliyon
Zhang ZJ,Sun ZX,Liu HJ
Sat Apr 01 00:00:00 EDT 2023
Crocetin confers neuroprotection and is anti-inflammatory in rats with induced glaucoma.
Molecular biology reports
Li Q,Feng P,Lin S,Xu Z,Zhao J,Chen Z,Luo Z,Tao Y,Chen S,Wang P
Wed Feb 01 00:00:00 EST 2023
Single-cell RNA sequencing deciphers the mechanism of sepsis-induced liver injury and the therapeutic effects of artesunate.
Acta pharmacologica Sinica
He XL,Chen JY,Feng YL,Song P,Wong YK,Xie LL,Wang C,Zhang Q,Bai YM,Gao P,Luo P,Liu Q,Liao FL,Li ZJ,Jiang Y,Wang JG
Fri Sep 01 00:00:00 EDT 2023
Protective effect of HDACIs in improves survival and organ injury after CLP-induced sepsis.
Surgery open science
Niu K,Qu S,Yang L,Zhang H,Yuan J,Fan H,Li X,Tao K
Wed Mar 01 00:00:00 EST 2023
Tunicamycin induces depression-like behaviors in male rats, accompanied by initiated chaperon-mediated autophagy and decreased synaptic protein expression in the hippocampus.
Neuroscience letters
Yuan G,Xiao L,Xie Y,Si L,Xu H,Xu W,Wang G
Tue Feb 28 00:00:00 EST 2023
Salmonella typhimurium exacerbates injuries but resolves fibrosis in liver and spleen during Schistosoma mansoni infection.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi
Lam HYP,Wu WJ,Liang TR,Li HC,Chang KC,Peng SY
Thu Jun 01 00:00:00 EDT 2023
Swimming training combined with fecal microbial transplantation protects motor functions in rats with spinal cord injury by improving the intestinal system.
Neuroscience letters
Ouyang S,Wang X,Chen Y,Deng L,Yang X,Hu S,Wu S
Thu Mar 16 00:00:00 EDT 2023
Apigenin Inhibits the Progression of Osteoarthritis by Mediating Macrophage Polarization.
Molecules (Basel, Switzerland)
Ji X,Du W,Che W,Wang L,Zhao L
Fri Mar 24 00:00:00 EDT 2023
CARMA3 Deficiency Aggravates Angiotensin II-Induced Abdominal Aortic Aneurysm Development Interacting Between Endoplasmic Reticulum and Mitochondria.
The Canadian journal of cardiology
Yao Y,Cao Y,Xu Y,Chen G,Liu Y,Jiang H,Fan R,Qin W,Wang X,Chai H,Chen X,Qiu Z,Chen W
Sun Oct 01 00:00:00 EDT 2023
Hepatoprotective effect of protocatechuic acid against type 2 diabetes-induced liver injury.
Pharmaceutical biology
Xu K,Lu G,Feng Q,Chen S,Wang Y
Fri Dec 01 00:00:00 EST 2023
Interleukin-37 ameliorates atherosclerosis by regulating autophagy-mediated endothelial cell apoptosis and inflammation.
International immunopharmacology
Xiao S,Song X,Zheng M,Cao X,Ai G,Li B,Zhao G,Yuan H
Mon May 01 00:00:00 EDT 2023
Bufalin alleviates acute kidney injury by regulating NLRP3 inflammasome-mediated pyroptosis.
Apoptosis : an international journal on programmed cell death
Ding L,Yin J,Xu X,Xie D,Xiang D,Tong P,Liu S,Yang X
Sat Apr 01 00:00:00 EDT 2023
ROS-responsive magnesium-containing microspheres for antioxidative treatment of intervertebral disc degeneration.
Acta biomaterialia
Zhang T,Wang Y,Li R,Xin J,Zheng Z,Zhang X,Xiao C,Zhang S
Wed Mar 01 00:00:00 EST 2023
Prussian Blue Nanozyme Treatment of Ischemic Brain Injury via Reducing Oxidative Stress Inhibits Inflammation, Suppresses Apoptosis, and Promotes Neurological Recovery.
ACS chemical neuroscience
Liu J,Sun J,Song Y,Wang M,Zhao P,Wang W,Yang M,Wang Y,Song Y,Sun B
Mon Apr 10 00:00:00 EDT 2023
Normothermic ex vivo heart perfusion with NLRP3 inflammasome inhibitor Mcc950 treatment improves cardiac function of circulatory death hearts after transplantation.
Frontiers in cardiovascular medicine
Xu L,Zeng Z,Niu C,Liu D,Lin S,Liu X,Szabó G,Lu J,Zheng S,Zhou P
Tue Apr 04 00:00:00 EDT 2023
Normothermic ex vivo heart perfusion with NLRP3 inflammasome inhibitor Mcc950 treatment improves cardiac function of circulatory death hearts after transplantation.
Frontiers in cardiovascular medicine
Xu L,Zeng Z,Niu C,Liu D,Lin S,Liu X,Szabó G,Lu J,Zheng S,Zhou P
Tue Apr 04 00:00:00 EDT 2023
Analyses of Transcriptomics upon IL-1β-Stimulated Mouse Chondrocytes and the Protective Effect of Catalpol through the NOD2/NF-κB/MAPK Signaling Pathway.
Molecules (Basel, Switzerland)
Pang Y,Zhao L,Ji X,Guo K,Yin X
Tue Feb 07 00:00:00 EST 2023
Microglial voltage-dependent anion channel 1 signaling modulates sleep deprivation-induced transition to chronic postsurgical pain.
Sleep
Wei SN,Zhang H,Lu Y,Yu HJ,Ma T,Wang SN,Yang K,Tian ML,Huang AH,Wang W,Li FS,Li YW
Wed Nov 08 00:00:00 EST 2023
Bu Shen Yi Sui Capsule Promotes Myelin Repair by Modulating the Transformation of A1/A2 Reactive Astrocytes In Vivo and In Vitro.
Oxidative medicine and cellular longevity
Zha Z,Liu YJ,Liu SS,Zhang N,Li JL,Qi F,Jin LY,Xue B,Yang T,Fan YP,Zhao H,Wang L
Tue Sep 13 00:00:00 EDT 2022
Adipose stem cells preincubated with theanine exert liver regeneration through increase of stem cell paracrine VEGF and suppression of ROS, pyroptosis as well as autophagy markers in liver damage induced by N-nitrosodiethylamine.
Life sciences
Chen TS,Lai YA,Lai YJ,Chien CT
Tue Nov 01 00:00:00 EDT 2022
Acetate Improves the Killing of Streptococcus pneumoniae by Alveolar Macrophages via NLRP3 Inflammasome and Glycolysis-HIF-1α Axis.
Frontiers in immunology
Machado MG,Patente TA,Rouillé Y,Heumel S,Melo EM,Deruyter L,Pourcet B,Sencio V,Teixeira MM,Trottein F
Wed Mar 02 00:00:00 EST 2022
Intrahepatic Cholestasis of Pregnancy Increases Inflammatory Susceptibility in Neonatal Offspring by Modulating Gut Microbiota.
Frontiers in immunology
Lin QX,Huang WW,Shen W,Deng XS,Tang ZY,Chen ZH,Zhao W,Fan HY
Fri Jul 01 00:00:00 EDT 2022
Tanshinone IIA Improves Acute Gouty Arthritis in Rats through Regulating Neutrophil Activation and the NLRP3 Inflammasome.
Disease markers
Xu L,Liu X,Zhang Y,Jia T,Li L,Du Y,Chen W,Zhang S
Fri Dec 30 00:00:00 EST 2022
Panax notoginseng saponins alleviates inflammation induced by microglial activation and protects against ischemic brain injury via inhibiting HIF-1α/PKM2/STAT3 signaling.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gao J,Yao M,Zhang W,Yang B,Yuan G,Liu JX,Zhang Y
Tue Nov 01 00:00:00 EDT 2022
Trans-anethole ameliorates LPS-induced inflammation via suppression of TLR4/NF-κB pathway in IEC-6 cells.
International immunopharmacology
Yu C,Wang D,Li Q,Tong Y,Yang Z,Wang T
Fri Jul 01 00:00:00 EDT 2022
Targeting A-kinase anchoring protein 12 phosphorylation in hepatic stellate cells regulates liver injury and fibrosis in mouse models.
eLife
Ramani K,Mavila N,Abeynayake A,Tomasi ML,Wang J,Matsuda M,Seki E
Tue Oct 04 00:00:00 EDT 2022
Continuous release of mefloquine featured in electrospun fiber membranes alleviates epidural fibrosis and aids in sensory neurological function after lumbar laminectomy.
Materials today. Bio
Yue Z,Hu B,Chen Z,Zheng G,Wang Y,Yang C,Cao P,Wu X,Liang L,Zang F,Wang J,Li J,Zhang T,Wu J,Chen H
Thu Dec 15 00:00:00 EST 2022
Chitosan/Sodium Alginate/Velvet Antler Blood Peptides Hydrogel Promotes Diabetic Wound Healing via Regulating Angiogenesis, Inflammatory Response and Skin Flora.
Journal of inflammation research
Hao M,Ding C,Sun S,Peng X,Liu W
Wed Sep 07 00:00:00 EDT 2022
Inhibition of Dectin-1 Alleviates Neuroinflammatory Injury by Attenuating NLRP3 Inflammasome-Mediated Pyroptosis After Intracerebral Hemorrhage in Mice: Preliminary Study Results.
Journal of inflammation research
Ding Z,Zhong Z,Wang J,Zhang R,Shao J,Li Y,Wu G,Tu H,Yuan W,Sun H,Wang Q
Tue Nov 01 00:00:00 EDT 2022
Cold Exposure Induces Intestinal Barrier Damage and Endoplasmic Reticulum Stress in the Colon via the SIRT1/Nrf2 Signaling Pathway.
Frontiers in physiology
Guo J,Hu H,Chen Z,Xu J,Nie J,Lu J,Ma L,Ji H,Yuan J,Xu B
Sat Jul 16 00:00:00 EDT 2022
Activated AXL Protects Against Hepatic Ischemia-reperfusion Injury by Upregulating SOCS-1 Expression.
Transplantation
Wang Z,Liu D,Yan Q,Liu F,Zhan M,Qi S,Fang Q,Yao L,Wang W,Zhang R,Du J,Chen L
Fri Jul 01 00:00:00 EDT 2022
Single-Cell RNA Sequencing Technology Revealed the Pivotal Role of Fibroblast Heterogeneity in Angiotensin II-Induced Abdominal Aortic Aneurysms.
DNA and cell biology
Weng Y,Lou J,Bao Y,Cai C,Zhu K,Du C,Chen X,Tang L
Sun May 01 00:00:00 EDT 2022
Anti-aging effect of phlorizin on D-galactose-induced aging in mice through antioxidant and anti-inflammatory activity, prevention of apoptosis, and regulation of the gut microbiota.
Experimental gerontology
Chen H,Dong L,Chen X,Ding C,Hao M,Peng X,Zhang Y,Zhu H,Liu W
Wed Jun 15 00:00:00 EDT 2022
Cold exposure-induced endoplasmic reticulum stress regulates autophagy through the SIRT2/FoxO1 signaling pathway.
Journal of cellular physiology
Guo J,Nie J,Chen Z,Wang X,Hu H,Xu J,Lu J,Ma L,Ji H,Yuan J,Xu B
Sat Oct 01 00:00:00 EDT 2022
Assessing the anti-inflammatory effects of quercetin using network pharmacology and in vitro experiments.
Experimental and therapeutic medicine
Zhang J,Li H,Wang W,Li H
Fri Apr 01 00:00:00 EDT 2022
ETC-1002 Attenuates Porphyromonas gingivalis Lipopolysaccharide-Induced Inflammation in RAW264.7 Cells via the AMPK/NF-κB Pathway and Exerts Ameliorative Effects in Experimental Periodontitis in Mice.
Disease markers
Li H,Zhang P,Lin H,Gao H,Yin J
Thu Apr 07 00:00:00 EDT 2022
Treadmill exercise alleviates neuronal damage by suppressing NLRP3 inflammasome and microglial activation in the MPTP mouse model of Parkinson's disease.
Brain research bulletin
Wang W,Lv Z,Gao J,Liu M,Wang Y,Tang C,Xiang J
Wed Sep 01 00:00:00 EDT 2021
The epigenetic regulator BRD4 is involved in cadmium-triggered inflammatory response in rat kidney.
Ecotoxicology and environmental safety
Gong Z,Liu G,Liu W,Zou H,Song R,Zhao H,Yuan Y,Gu J,Bian J,Zhu J,Liu Z
Fri Aug 13 00:00:00 EDT 2021
Inhibitor of apoptosis-stimulating p53 protein protects against inflammatory bowel disease in mice models by inhibiting the nuclear factor kappa B signaling.
Clinical and experimental immunology
Qian K,Yuan L,Wang S,Kuang Y,Jin Q,Long D,Jiang Y,Zhao H,Liu K,Yao H
Sun Aug 01 00:00:00 EDT 2021
Ligand-based substituent-anchoring design of selective receptor-interacting protein kinase 1 necroptosis inhibitors for ulcerative colitis therapy.
Acta pharmaceutica Sinica. B
Zhu J,Xin M,Xu C,He Y,Zhang W,Wang Z,Zhuang C
Fri Oct 01 00:00:00 EDT 2021
Btk knockout attenuates the liver inflammation in STZ-induced diabetic mice by suppressing NLRP3 inflammasome activation.
Biochemical and biophysical research communications
Li Y,Zhao J,Wu Y,Xia L
Fri Apr 16 00:00:00 EDT 2021
Arbutin Ameliorates Murine Colitis by Inhibiting JAK2 Signaling Pathway.
Frontiers in pharmacology
Wang L,Feng Y,Wang J,Luo T,Wang X,Wu M,Wang R,Chen D,Li J,Wang J
Sat Oct 02 00:00:00 EDT 2021
Rufinamide (RUF) suppresses inflammation and maintains the integrity of the blood-brain barrier during kainic acid-induced brain damage.
Open life sciences
Yu H,He B,Han X,Yan T
Tue Sep 14 00:00:00 EDT 2021
A "dual-guide" bioinspired drug delivery strategy of a macrophage-based carrier against postoperative triple-negative breast cancer recurrence.
Journal of controlled release : official journal of the Controlled Release Society
Qiu Y,Ren K,Zhao W,Yu Q,Guo R,He J,Mei L,Liu Y,Tang J,Xu S,Li J,Wei J,Li M,Zhang Z,He Q
Sun Jan 10 00:00:00 EST 2021
Luteolin Alleviates Methamphetamine-Induced Hepatotoxicity by Suppressing the p53 Pathway-Mediated Apoptosis, Autophagy, and Inflammation in Rats.
Frontiers in pharmacology
Zhang KK,Wang H,Qu D,Chen LJ,Wang LB,Li JH,Liu JL,Xu LL,Yoshida JS,Xu JT,Xie XL,Li DR
Wed Mar 10 00:00:00 EST 2021
Deficiency of CARMA3 attenuates the development of bleomycin induced pulmonary fibrosis.
Biochemical and biophysical research communications
Wu Y,Qi Y,Qiu Z,Chen W
Fri Dec 03 00:00:00 EST 2021
Deficiency of CARMA3 attenuates the development of bleomycin induced pulmonary fibrosis.
Biochemical and biophysical research communications
Wu Y,Qi Y,Qiu Z,Chen W
Fri Dec 03 00:00:00 EST 2021
Resveratrol Alleviates Levodopa-Induced Dyskinesia in Rats.
Frontiers in immunology
Zheng CQ,Fan HX,Li XX,Li JJ,Sheng S,Zhang F
Mon Nov 01 00:00:00 EDT 2021
Sodium Sulfite-Induced Mast Cell Pyroptosis and Degranulation.
Journal of agricultural and food chemistry
Liu M,Lu J,Chen Y,Shi X,Li Y,Yang S,Yu J,Guan S
Wed Jul 14 00:00:00 EDT 2021
Ageing affects the proliferation and mineralization of rat dental pulp stem cells under inflammatory conditions.
International endodontic journal
Ning T,Shao J,Zhang X,Luo X,Huang X,Wu H,Xu S,Wu B,Ma D
Wed Jan 01 00:00:00 EST 2020
Neuroprotective effect of liquiritin against neuropathic pain induced by chronic constriction injury of the sciatic nerve in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang MT,Wang B,Jia YN,Liu N,Ma PS,Gong SS,Niu Y,Sun T,Li YX,Yu JQ
Wed Nov 01 00:00:00 EDT 2017
Palmitic acid aggravates inflammation of pancreatic acinar cells by enhancing unfolded protein response induced CCAAT-enhancer-binding protein β-CCAAT-enhancer-binding protein α activation.
The international journal of biochemistry & cell biology
Wu J,Hu G,Lu Y,Zheng J,Chen J,Wang X,Zeng Y
Sat Oct 01 00:00:00 EDT 2016