Product References
Disruption of desmosome function leads to increased centrosome clustering in 14-3-3γ-knockout cells with supernumerary centrosomes.
FEBS letters
Tilwani S,Gandhi K,Narayan S,Ainavarapu SRK,Dalal SN
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Nov 01 00:00:00 EDT 2021
Disruption of desmosome function leads to increased centrosome clustering in 14-3-3γ-knockout cells with supernumerary centrosomes.
FEBS letters
Tilwani S,Gandhi K,Narayan S,Ainavarapu SRK,Dalal SN
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Nov 01 00:00:00 EDT 2021
Leaky Gut as a Potential Culprit for the Paradoxical Dysglycemic Response to Gastric Bypass-Associated Ileal Microbiota.
Metabolites
Hankir MK,Seyfried F,Schellinger IN,Schlegel N,Arora T
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
Leaky Gut as a Potential Culprit for the Paradoxical Dysglycemic Response to Gastric Bypass-Associated Ileal Microbiota.
Metabolites
Hankir MK,Seyfried F,Schellinger IN,Schlegel N,Arora T
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
Leaky Gut as a Potential Culprit for the Paradoxical Dysglycemic Response to Gastric Bypass-Associated Ileal Microbiota.
Metabolites
Hankir MK,Seyfried F,Schellinger IN,Schlegel N,Arora T
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
Leaky Gut as a Potential Culprit for the Paradoxical Dysglycemic Response to Gastric Bypass-Associated Ileal Microbiota.
Metabolites
Hankir MK,Seyfried F,Schellinger IN,Schlegel N,Arora T
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
The cell-cell junctions of mammalian testes: II. The lamellar smooth muscle monolayer cells of the peritubular wall are laterally connected by vertical adherens junctions-a novel architectonic cell-cell junction system.
Cell and tissue research
Domke LM,Franke WW
32-6100 was used in Immunocytochemistry-immunoflourescence to show the LSMCs of the peritubular wall monolayers are SMCs sensu stricto which are laterally connected by a novel architectonic system of arrays of vertical AJs located in overlapping cell protrusions.
Fri Feb 01 00:00:00 EST 2019
A non-canonical role for desmoglein-2 in endothelial cells: implications for neoangiogenesis.
Angiogenesis
Ebert LM,Tan LY,Johan MZ,Min KK,Cockshell MP,Parham KA,Betterman KL,Szeto P,Boyle S,Silva L,Peng A,Zhang Y,Ruszkiewicz A,Zannettino AC,Gronthos S,Koblar S,Harvey NL,Lopez AF,Shackleton M,Bonder CS
326100 was used in flow cytometry and immunohistochemistry - paraffin section to study the function of desmoglein-2 and generate knockout mice
Sat Oct 01 00:00:00 EDT 2016
CUB domain-containing protein 1 and the epidermal growth factor receptor cooperate to induce cell detachment.
Breast cancer research : BCR
Law ME,Ferreira RB,Davis BJ,Higgins PJ,Kim JS,Castellano RK,Chen S,Luesch H,Law BK
326100 was used in western blot to study how CUB domain-containing protein1 contributes to metastasis and drug resistance
Fri Aug 05 00:00:00 EDT 2016
Desmoglein 2 promotes vasculogenic mimicry in melanoma and is associated with poor clinical outcome.
Oncotarget
Tan LY,Mintoff C,Johan MZ,Ebert BW,Fedele C,Zhang YF,Szeto P,Sheppard KE,McArthur GA,Foster-Smith E,Ruszkiewicz A,Brown MP,Bonder CS,Shackleton M,Ebert LM
32-6100 was used in Flow cytometry/Cell sorting to study the mechanisms that regulate vasculogenic mimicry.
Tue Jul 19 00:00:00 EDT 2016
Desmoglein 2 depletion leads to increased migration and upregulation of the chemoattractant secretoneurin in melanoma cells.
PloS one
Peitsch WK,Doerflinger Y,Fischer-Colbrie R,Huck V,Bauer AT,Utikal J,Goerdt S,Schneider SW
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunohistochemistry
Tue Oct 21 00:00:00 EDT 2014
Desmoglein 2 depletion leads to increased migration and upregulation of the chemoattractant secretoneurin in melanoma cells.
PloS one
Peitsch WK,Doerflinger Y,Fischer-Colbrie R,Huck V,Bauer AT,Utikal J,Goerdt S,Schneider SW
Published figure using Desmoglein 2 monoclonal antibody (Product # 32-6100) in Immunohistochemistry
Tue Oct 21 00:00:00 EDT 2014
Gastro-oesophageal reflux disease is associated with up-regulation of desmosomal components in oesophageal mucosa.
Histopathology
Wex T,Mönkemüller K,Stahr A,Kuester D,Fry LC,Völkel S,Kandulski A,Roessner A,Malfertheiner P
32-6100 was used in immunohistochemistry to examine the role of desmosomal proteins in gastro-oesophageal reflux disease.
Wed Feb 01 00:00:00 EST 2012
Assessment of desmosomal components (desmoglein 1-3, plakoglobin) in cardia mucosa in relation to gastroesophageal reflux disease and Helicobacter pylori infection.
Human pathology
Wex T,Kuester D,Mönkemüller K,Stahr A,Fry LC,Kandulski A,Kropf S,Roessner A,Malfertheiner P
32-6100 was used in immunohistochemistry to examine the contribution of the desmosomal compartment to gastroesophageal reflux disease.
Mon Oct 01 00:00:00 EDT 2012
Differential downregulation of e-cadherin and desmoglein by epidermal growth factor.
Dermatology research and practice
Chavez MG,Buhr CA,Petrie WK,Wandinger-Ness A,Kusewitt DF,Hudson LG
32-6100 was used in western blot to investigate the effect of EGF on the expression of E-cadherin and desmoglein and its mechanism.
Thu Aug 23 00:00:00 EDT 2012
Mispolarization of desmosomal proteins and altered intercellular adhesion in autosomal dominant polycystic kidney disease.
American journal of physiology. Renal physiology
Silberberg M,Charron AJ,Bacallao R,Wandinger-Ness A
Wed Jun 01 00:00:00 EDT 2005