Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Specificity: Neutralizes mouse interferon beta.
Activity: 1.55x10^6 NU/mL. Interferon was titrated with the use of the cytopathic effect inhibition assay using L929 cells with EMCV, where 2.5 unit/mL of interferon was necessary to produce an endpoint of 50%. One neutralization unit is defined as the amount of serum required to neutralize one unit of mouse IFN beta to an endpoint of 50%.
For retention of full activity, do not store above -70ºC.
Packaged by and distributed for PBL Assay Science.
Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN b; IFN β; IFN-beta; IFNbeta; IFNβ; Interferon b; Interferon beta; RP11-113D19.1
Gene Aliases: Ifb; IFN-beta; IFNB; Ifnb1
UniProt ID: (Mouse) P01575
Entrez Gene ID: (Mouse) 15977
Molecular Function: interferon superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support