Product References
KIFC1 Regulates the Trajectory of Neuronal Migration.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Muralidharan H,Guha S,Madugula K,Patil A,Bennison SA,Sun X,Toyo-Oka K,Baas PW
33-2100 was used in Immunocytochemistry-immunofluorescence to find that crosslinking of microtubules into a nonsliding mode by KIFC1 is necessary for dynein-driven forces to achieve sufficient traction to thrust the soma forward.
Wed Mar 16 00:00:00 EDT 2022
SATB2-LEMD2 interaction links nuclear shape plasticity to regulation of cognition-related genes.
The EMBO journal
Feurle P,Abentung A,Cera I,Wahl N,Ablinger C,Bucher M,Stefan E,Sprenger S,Teis D,Fischer A,Laighneach A,Whitton L,Morris DW,Apostolova G,Dechant G
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunocytochemistry
Mon Feb 01 00:00:00 EST 2021
SATB2-LEMD2 interaction links nuclear shape plasticity to regulation of cognition-related genes.
The EMBO journal
Feurle P,Abentung A,Cera I,Wahl N,Ablinger C,Bucher M,Stefan E,Sprenger S,Teis D,Fischer A,Laighneach A,Whitton L,Morris DW,Apostolova G,Dechant G
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunocytochemistry
Mon Feb 01 00:00:00 EST 2021
A lamin A/C variant causing striated muscle disease provides insights into filament organization.
Journal of cell science
Kronenberg-Tenga R,Tatli M,Eibauer M,Wu W,Shin JY,Bonne G,Worman HJ,Medalia O
33-2100 was used in Western Blot to investigate nuclear lamina molecular changes occurring in cardiomyopathy and muscular dystrophy using a mouse model.
Mon Mar 22 00:00:00 EDT 2021
A small-molecule ICMT inhibitor delays senescence of Hutchinson-Gilford progeria syndrome cells.
eLife
Chen X,Yao H,Kashif M,Revêchon G,Eriksson M,Hu J,Wang T,Liu Y,Tüksammel E,Strömblad S,Ahearn IM,Philips MR,Wiel C,Ibrahim MX,Bergo MO
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 02 00:00:00 EST 2021
A small-molecule ICMT inhibitor delays senescence of Hutchinson-Gilford progeria syndrome cells.
eLife
Chen X,Yao H,Kashif M,Revêchon G,Eriksson M,Hu J,Wang T,Liu Y,Tüksammel E,Strömblad S,Ahearn IM,Philips MR,Wiel C,Ibrahim MX,Bergo MO
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 02 00:00:00 EST 2021
Chromatin compartment dynamics in a haploinsufficient model of cardiac laminopathy.
The Journal of cell biology
Bertero A,Fields PA,Smith AST,Leonard A,Beussman K,Sniadecki NJ,Kim DH,Tse HF,Pabon L,Shendure J,Noble WS,Murry CE
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Mon Sep 02 00:00:00 EDT 2019
Differential expression of nuclear lamin subtypes in the neural cells of the adult rat cerebral cortex.
IBRO reports
Takamori Y,Hirahara Y,Wakabayashi T,Mori T,Koike T,Kataoka Y,Tamura Y,Kurebayashi S,Kurokawa K,Yamada H
33-2100 was used in Immunohistochemistry to investigate the composition of lamin subtypes in neurons, astrocytes, oligodendrocyte-lineage cells, and microglia in the adult rat cerebral cortex.
Sat Dec 01 00:00:00 EST 2018
Detection of DNA Double Strand Breaks by γH2AX Does Not Result in 53bp1 Recruitment in Mouse Retinal Tissues.
Frontiers in neuroscience
Müller B,Ellinwood NM,Lorenz B,Stieger K
33-2100 was used in Immunohistochemistry-immunofluorescence to investigate the recruitment of the double stranded break repair complex of γH2AX and 53bp1 in both developing and mature mouse neuroretinas.
Fri Sep 24 00:00:00 EDT 2021
Lamin B1 is required for mature neuron-specific gene expression during olfactory sensory neuron differentiation.
Nature communications
Gigante CM,Dibattista M,Dong FN,Zheng X,Yue S,Young SG,Reisert J,Zheng Y,Zhao H
33-2100 was used in Immunohistochemistry-immunofluorescence to identify lamin B1 as a key regulator of gene expression required for the formation of functional olfactory sensory neurons.
Thu Apr 20 00:00:00 EDT 2017
Lamin B1 is required for mature neuron-specific gene expression during olfactory sensory neuron differentiation.
Nature communications
Gigante CM,Dibattista M,Dong FN,Zheng X,Yue S,Young SG,Reisert J,Zheng Y,Zhao H
33-2100 was used in Immunohistochemistry-immunofluorescence to identify lamin B1 as a key regulator of gene expression required for the formation of functional olfactory sensory neurons.
Thu Apr 20 00:00:00 EDT 2017
MET receptor variant R970C favors calpain-dependent generation of a fragment promoting epithelial cell scattering.
Oncotarget
Montagne R,Baranzelli A,Muharram G,Catherine L,Lesaffre M,Vinchent A,Kherrouche Z,Werkmeister E,Cortot AB,Tulasne D
33-2100 was used in Western Blotting to examine the influence of mutations in the MET juxtamembrane domain on its proteolytic cleavages.
Tue Feb 14 00:00:00 EST 2017
Intracellular mGluR5 plays a critical role in neuropathic pain.
Nature communications
Vincent K,Cornea VM,Jong YI,Laferrière A,Kumar N,Mickeviciute A,Fung JST,Bandegi P,Ribeiro-da-Silva A,O'Malley KL,Coderre TJ
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Wed Feb 03 00:00:00 EST 2016
Intracellular mGluR5 plays a critical role in neuropathic pain.
Nature communications
Vincent K,Cornea VM,Jong YI,Laferrière A,Kumar N,Mickeviciute A,Fung JST,Bandegi P,Ribeiro-da-Silva A,O'Malley KL,Coderre TJ
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Wed Feb 03 00:00:00 EST 2016
High-resolution Antibody Array Analysis of Childhood Acute Leukemia Cells.
Molecular & cellular proteomics : MCP
Kanderova V,Kuzilkova D,Stuchly J,Vaskova M,Brdicka T,Fiser K,Hrusak O,Lund-Johansen F,Kalina T
332100 was used to use size exclusion chromatography-microsphere-based affinity proteomics to study clinical samples obtained from pediatric acute leukemia patients
Fri Apr 01 00:00:00 EDT 2016
An absence of nuclear lamins in keratinocytes leads to ichthyosis, defective epidermal barrier function, and intrusion of nuclear membranes and endoplasmic reticulum into the nuclear chromatin.
Molecular and cellular biology
Jung HJ,Tatar A,Tu Y,Nobumori C,Yang SH,Goulbourne CN,Herrmann H,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Mon Dec 01 00:00:00 EST 2014
An absence of nuclear lamins in keratinocytes leads to ichthyosis, defective epidermal barrier function, and intrusion of nuclear membranes and endoplasmic reticulum into the nuclear chromatin.
Molecular and cellular biology
Jung HJ,Tatar A,Tu Y,Nobumori C,Yang SH,Goulbourne CN,Herrmann H,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Mon Dec 01 00:00:00 EST 2014
Reciprocal knock-in mice to investigate the functional redundancy of lamin B1 and lamin B2.
Molecular biology of the cell
Lee JM,Tu Y,Tatar A,Wu D,Nobumori C,Jung HJ,Yoshinaga Y,Coffinier C,de Jong PJ,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Thu May 01 00:00:00 EDT 2014
Reciprocal knock-in mice to investigate the functional redundancy of lamin B1 and lamin B2.
Molecular biology of the cell
Lee JM,Tu Y,Tatar A,Wu D,Nobumori C,Jung HJ,Yoshinaga Y,Coffinier C,de Jong PJ,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Thu May 01 00:00:00 EDT 2014
Farnesylation of lamin B1 is important for retention of nuclear chromatin during neuronal migration.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Nobumori C,Goulbourne CN,Tu Y,Lee JM,Tatar A,Wu D,Yoshinaga Y,de Jong PJ,Coffinier C,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Tue May 21 00:00:00 EDT 2013
Farnesylation of lamin B1 is important for retention of nuclear chromatin during neuronal migration.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Nobumori C,Goulbourne CN,Tu Y,Lee JM,Tatar A,Wu D,Yoshinaga Y,de Jong PJ,Coffinier C,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Tue May 21 00:00:00 EDT 2013
Farnesylation of lamin B1 is important for retention of nuclear chromatin during neuronal migration.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Nobumori C,Goulbourne CN,Tu Y,Lee JM,Tatar A,Wu D,Yoshinaga Y,de Jong PJ,Coffinier C,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Tue May 21 00:00:00 EDT 2013
Farnesylation of lamin B1 is important for retention of nuclear chromatin during neuronal migration.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Nobumori C,Goulbourne CN,Tu Y,Lee JM,Tatar A,Wu D,Yoshinaga Y,de Jong PJ,Coffinier C,Fong LG,Young SG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Immunofluorescence
Tue May 21 00:00:00 EDT 2013
Regulation of prelamin A but not lamin C by miR-9, a brain-specific microRNA.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Coffinier C,Choe Y,Beigneux AP,Davies BS,Yang SH,Barnes RH,Hong J,Sun T,Pleasure SJ,Young SG,Fong LG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 14 00:00:00 EST 2012
Regulation of prelamin A but not lamin C by miR-9, a brain-specific microRNA.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Coffinier C,Choe Y,Beigneux AP,Davies BS,Yang SH,Barnes RH,Hong J,Sun T,Pleasure SJ,Young SG,Fong LG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 14 00:00:00 EST 2012
Regulation of prelamin A but not lamin C by miR-9, a brain-specific microRNA.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Coffinier C,Choe Y,Beigneux AP,Davies BS,Yang SH,Barnes RH,Hong J,Sun T,Pleasure SJ,Young SG,Fong LG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 14 00:00:00 EST 2012
Regulation of prelamin A but not lamin C by miR-9, a brain-specific microRNA.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Coffinier C,Choe Y,Beigneux AP,Davies BS,Yang SH,Barnes RH,Hong J,Sun T,Pleasure SJ,Young SG,Fong LG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 14 00:00:00 EST 2012
Regulation of prelamin A but not lamin C by miR-9, a brain-specific microRNA.
Proceedings of the National Academy of Sciences of the United States of America
Jung HJ,Coffinier C,Choe Y,Beigneux AP,Davies BS,Yang SH,Barnes RH,Hong J,Sun T,Pleasure SJ,Young SG,Fong LG
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Tue Feb 14 00:00:00 EST 2012
Nuclear lamins are differentially expressed in retinal neurons of the adult rat retina.
Histochemistry and cell biology
Wakabayashi T,Mori T,Hirahara Y,Koike T,Kubota Y,Takamori Y,Yamada H
33-2100 was used in immunohistochemistry and western blot to study the expression patterns of lamin subtypes in the adult rat retina.
Sat Oct 01 00:00:00 EDT 2011
A {gamma}-secretase-independent mechanism of signal transduction by the amyloid precursor protein.
The Journal of biological chemistry
Hass MR,Yankner BA
Published figure using Lamin B2 monoclonal antibody (Product # 33-2100) in Western Blot
Fri Nov 04 00:00:00 EST 2005
Functional metabotropic glutamate receptors on nuclei from brain and primary cultured striatal neurons. Role of transporters in delivering ligand.
The Journal of biological chemistry
Jong YJ,Kumar V,Kingston AE,Romano C,O'Malley KL
Fri Aug 26 00:00:00 EDT 2005
Activation of metabotropic glutamate receptor mGlu5 on nuclear membranes mediates intranuclear Ca2+ changes in heterologous cell types and neurons.
The Journal of biological chemistry
O'Malley KL,Jong YJ,Gonchar Y,Burkhalter A,Romano C
Fri Jul 25 00:00:00 EDT 2003
Binding of neuronal nitric-oxide synthase (nNOS) to carboxyl-terminal-binding protein (CtBP) changes the localization of CtBP from the nucleus to the cytosol: a novel function for targeting by the PDZ domain of nNOS.
The Journal of biological chemistry
Riefler GM,Firestein BL
Fri Dec 21 00:00:00 EST 2001