Product References
Effect of immune modulation on the skeletal muscle mitochondrial exercise response: An exploratory study in mice with cancer.
PloS one
Buss LA,Hock B,Merry TL,Ang AD,Robinson BA,Currie MJ,Dachs GU
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Western Blot
Wed Nov 24 00:00:00 EST 2021
Impact of Exercise and Detraining during Childhood on Brown Adipose Tissue Whitening in Obesity.
Metabolites
Takaishi K,Oshima T,Eto H,Nishihira M,Nguyen ST,Ochi R,Fujita N,Urakawa S
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Western Blot
Fri Oct 01 00:00:00 EDT 2021
The Effects of Potassium Cyanide on the Functional Recovery of Isolated Rat Hearts after Ischemia and Reperfusion: The Role of Oxidative Stress.
Oxidative medicine and cellular longevity
Petkovic AM,Jakovljevic VL,Bradic JV,Jeremic JN,Jeremic NS,Nikolic Turnic TR,Jovicic NU,Rosic VZ,Srejovic IM,Zivkovic VI
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Immunohistochemistry
Wed Dec 19 00:00:00 EST 2018
Tumor protein Tctp regulates axon development in the embryonic visual system.
Development (Cambridge, England)
Roque CG,Wong HH,Lin JQ,Holt CE
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Western Blot
Fri Apr 01 00:00:00 EDT 2016
Morphine stimulates nitric oxide release in human mitochondria.
Journal of bioenergetics and biomembranes
Stefano GB,Mantione KJ,Capellan L,Casares FM,Challenger S,Ramin R,Samuel JM,Snyder C,Kream RM
33-8500 was used in western blot to discuss morphinergic/NO-coupled regulation.
Thu Oct 01 00:00:00 EDT 2015
The BRCA1 tumor suppressor binds to inositol 1,4,5-trisphosphate receptors to stimulate apoptotic calcium release.
The Journal of biological chemistry
Hedgepeth SC,Garcia MI,Wagner LE,Rodriguez AM,Chintapalli SV,Snyder RR,Hankins GD,Henderson BR,Brodie KM,Yule DI,van Rossum DB,Boehning D
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Western Blot
Fri Mar 13 00:00:00 EDT 2015
The BRCA1 tumor suppressor binds to inositol 1,4,5-trisphosphate receptors to stimulate apoptotic calcium release.
The Journal of biological chemistry
Hedgepeth SC,Garcia MI,Wagner LE,Rodriguez AM,Chintapalli SV,Snyder RR,Hankins GD,Henderson BR,Brodie KM,Yule DI,van Rossum DB,Boehning D
Published figure using Cytochrome C monoclonal antibody (Product # 33-8500) in Western Blot
Fri Mar 13 00:00:00 EDT 2015
Targeting the GD3 acetylation pathway selectively induces apoptosis in glioblastoma.
Neuro-oncology
Birks SM,Danquah JO,King L,Vlasak R,Gorecki DC,Pilkington GJ
Thu Sep 01 00:00:00 EDT 2011
Protective effect of sulforaphane against cisplatin-induced mitochondrial alterations and impairment in the activity of NAD(P)H: quinone oxidoreductase 1 and γ glutamyl cysteine ligase: studies in mitochondria isolated from rat kidney and in LLC-PK1 cells.
Toxicology letters
Guerrero-Beltrán CE,Calderón-Oliver M,Martínez-Abundis E,Tapia E,Zarco-Márquez G,Zazueta C,Pedraza-Chaverri J
33-8500 was used in western blot to test if sulforaphane alters cisplatin-induced mitochondrial alterations and impairs cytoprotective enzyme activity
Wed Nov 10 00:00:00 EST 2010
Lindane and cell death: at the crossroads between apoptosis, necrosis and autophagy.
Toxicology
Zucchini-Pascal N,de Sousa G,Rahmani R
33-8500 was used in western blot to examine the effects of lindane on autophagic, apoptotic, and necrotic cell death in rat hepatocytes
Wed Feb 04 00:00:00 EST 2009
Mitochondrial disruption in Drosophila apoptosis.
Developmental cell
Abdelwahid E,Yokokura T,Krieser RJ,Balasundaram S,Fowle WH,White K
33-8500 was used in immunohistochemistry and western blot to study how Reaper, Hid, and Drp1 regulate mitochondrial disruption in fly apoptosis
Tue May 01 00:00:00 EDT 2007
Mitochondrial impairment induced by poly(ADP-ribose) polymerase-1 activation in cortical neurons after oxygen and glucose deprivation.
Journal of neurochemistry
Tanaka S,Takehashi M,Iida S,Kitajima T,Kamanaka Y,Stedeford T,Banasik M,Ueda K
33-8500 was used in western blot to report that PARP-1 contributes to mitochondrial impairment that ultimately leads to neuronal apoptosis after cerebral ischemia.
Sat Oct 01 00:00:00 EDT 2005
Cadmium-induced apoptosis through the mitochondrial pathway in rainbow trout hepatocytes: involvement of oxidative stress.
Aquatic toxicology (Amsterdam, Netherlands)
Risso-de Faverney C,Orsini N,de Sousa G,Rahmani R
Wed Aug 25 00:00:00 EDT 2004
Sphingosine-phosphate lyase enhances stress-induced ceramide generation and apoptosis.
The Journal of biological chemistry
Reiss U,Oskouian B,Zhou J,Gupta V,Sooriyakumaran P,Kelly S,Wang E,Merrill AH,Saba JD
Fri Jan 09 00:00:00 EST 2004
Mitochondrial and nucleocytoplasmic targeting of O-linked GlcNAc transferase.
Journal of cell science
Love DC,Kochan J,Cathey RL,Shin SH,Hanover JA
Sat Feb 15 00:00:00 EST 2003
Mycobacterium tuberculosis virulence correlates with mitochondrial cytochrome c release in infected macrophages.
Scandinavian journal of immunology
Abarca-Rojano E,Rosas-Medina P,Zamudio-Cortéz P,Mondragón-Flores R,Sánchez-García FJ
33-8500 was used in western blot to examine mitochondrial function in the course of intracellular infections.
Wed Oct 01 00:00:00 EDT 2003
Afferent regulation of cytochrome-c and active caspase-9 in the avian cochlear nucleus.
Neuroscience
Wilkinson BL,Elam JS,Fadool DA,Hyson RL
33-8500 was used in Western Blotting to examine the extrusion of cytochrome-c from the mitochondria into the cytosol and the subsequent activation of proteolytic caspase cascades in the nucleus magnocellularis following deafferentation.
Mon Nov 10 00:00:00 EST 2003
Afferent regulation of cytochrome-c and active caspase-9 in the avian cochlear nucleus.
Neuroscience
Wilkinson BL,Elam JS,Fadool DA,Hyson RL
33-8500 was used in Western Blotting to examine the extrusion of cytochrome-c from the mitochondria into the cytosol and the subsequent activation of proteolytic caspase cascades in the nucleus magnocellularis following deafferentation.
Mon Nov 10 00:00:00 EST 2003
Microcin E492, a channel-forming bacteriocin from Klebsiella pneumoniae, induces apoptosis in some human cell lines.
Proceedings of the National Academy of Sciences of the United States of America
Hetz C,Bono MR,Barros LF,Lagos R
33-8500 was used in Western Blotting to describe a bacteriocin, microcin E492, that has the capacity to induce apoptosis in human cell lines.
Tue Mar 05 00:00:00 EST 2002
Microcin E492, a channel-forming bacteriocin from Klebsiella pneumoniae, induces apoptosis in some human cell lines.
Proceedings of the National Academy of Sciences of the United States of America
Hetz C,Bono MR,Barros LF,Lagos R
33-8500 was used in Western Blotting to describe a bacteriocin, microcin E492, that has the capacity to induce apoptosis in human cell lines.
Tue Mar 05 00:00:00 EST 2002
The human dynamin-related protein OPA1 is anchored to the mitochondrial inner membrane facing the inter-membrane space.
FEBS letters
Olichon A,Emorine LJ,Descoins E,Pelloquin L,Brichese L,Gas N,Guillou E,Delettre C,Valette A,Hamel CP,Ducommun B,Lenaers G,Belenguer P
33-8500 was used in western blot to report that OPA1 resides in the mitochondria.
Wed Jul 17 00:00:00 EDT 2002
Galectin-3 translocates to the perinuclear membranes and inhibits cytochrome c release from the mitochondria. A role for synexin in galectin-3 translocation.
The Journal of biological chemistry
Yu F,Finley RL,Raz A,Kim HR
Fri May 03 00:00:00 EDT 2002
Galectin-3 translocates to the perinuclear membranes and inhibits cytochrome c release from the mitochondria. A role for synexin in galectin-3 translocation.
The Journal of biological chemistry
Yu F,Finley RL,Raz A,Kim HR
Fri May 03 00:00:00 EDT 2002
Cycloheximide-induced T-cell death is mediated by a Fas-associated death domain-dependent mechanism.
The Journal of biological chemistry
Tang D,Lahti JM,Grenet J,Kidd VJ
33-8500 was used in western blot to elucidate the mechanisms that regulate cycloheximide-induced apoptosis
Fri Mar 12 00:00:00 EST 1999