Product References
Mast Cell/Proteinase Activated Receptor 2 (PAR2) Mediated Interactions in the Pathogenesis of Discogenic Back Pain.
Frontiers in cellular neuroscience
Richards J,Tang S,Gunsch G,Sul P,Wiet M,Flanigan DC,Khan SN,Moore S,Walter B,Purmessur D
Published figure using PAR2 monoclonal antibody (Product # 35-2300) in Immunofluorescence
Thu Oct 01 00:00:00 EDT 2020
Mast Cell/Proteinase Activated Receptor 2 (PAR2) Mediated Interactions in the Pathogenesis of Discogenic Back Pain.
Frontiers in cellular neuroscience
Richards J,Tang S,Gunsch G,Sul P,Wiet M,Flanigan DC,Khan SN,Moore S,Walter B,Purmessur D
Published figure using PAR2 monoclonal antibody (Product # 35-2300) in Immunofluorescence
Thu Oct 01 00:00:00 EDT 2020
Mast cells and mast cell tryptase enhance migration of human lung fibroblasts through protease-activated receptor 2.
Cell communication and signaling : CCS
Bagher M,Larsson-Callerfelt AK,Rosmark O,Hallgren O,Bjermer L,Westergren-Thorsson G
35-2300 was used in Immunocytochemistry-immunoflourescence to demonstrate roles for mast cells and the mast cell mediator tryptase in inducing lung fibroblast migration via PAR-2 activation.
Sat Sep 15 00:00:00 EDT 2018
Mast cells and mast cell tryptase enhance migration of human lung fibroblasts through protease-activated receptor 2.
Cell communication and signaling : CCS
Bagher M,Larsson-Callerfelt AK,Rosmark O,Hallgren O,Bjermer L,Westergren-Thorsson G
35-2300 was used in Immunocytochemistry-immunoflourescence to demonstrate roles for mast cells and the mast cell mediator tryptase in inducing lung fibroblast migration via PAR-2 activation.
Sat Sep 15 00:00:00 EDT 2018
Poly(I:C) Induces Human Lung Endothelial Barrier Dysfunction by Disrupting Tight Junction Expression of Claudin-5.
PloS one
Huang LY,Stuart C,Takeda K,D'Agnillo F,Golding B
35-2300 was used in immunocytochemistry assess the effect of polyinosinic-polycytidylic acid on the barrier function and tight junction integrity of primary human lung microvascular endothelial cells
Fri Aug 04 00:00:00 EDT 2017
Retinal neurons curb inflammation and enhance revascularization in ischemic retinopathies via proteinase-activated receptor-2.
The American journal of pathology
Sitaras N,Rivera JC,Noueihed B,Bien-Aimé M,Zaniolo K,Omri S,Hamel D,Zhu T,Hardy P,Sapieha P,Joyal JS,Chemtob S
35-2300 was used in immunohistochemistry and western blot to test if proteinase-activated receptor-2 modulates the deleterious effects of IL-1beta in ischemic retinopathies.
Sun Feb 01 00:00:00 EST 2015
Retinal neurons curb inflammation and enhance revascularization in ischemic retinopathies via proteinase-activated receptor-2.
The American journal of pathology
Sitaras N,Rivera JC,Noueihed B,Bien-Aimé M,Zaniolo K,Omri S,Hamel D,Zhu T,Hardy P,Sapieha P,Joyal JS,Chemtob S
35-2300 was used in immunohistochemistry and western blot to test if proteinase-activated receptor-2 modulates the deleterious effects of IL-1beta in ischemic retinopathies.
Sun Feb 01 00:00:00 EST 2015
Analysis of protease-activated receptor-1 and -2 in human scar formation.
The Journal of pathology
Materazzi S,Pellerito S,Di Serio C,Paglierani M,Naldini A,Ardinghi C,Carraro F,Geppetti P,Cirino G,Santucci M,Tarantini F,Massi D
35-2300 was used in immunohistochemistry (paraffin) to examine expression and biological activity of PAR-1 and PAR-2 in normal and pathological cutaneous scars.
Wed Aug 01 00:00:00 EDT 2007
Expression of protease-activated receptor-1 and -2 in orofacial granulomatosis.
Oral diseases
Ketabchi S,Massi D,Ficarra G,Rubino I,Franchi A,Paglierani M,Simoni A,Capodiferro S,Favia G,Maiorano E,Tarantini F,Cirino G,Santucci M
35-2300 was used in immunohistochemistry - paraffin section to determine the expression of PAR-1, PAR-2, MMP-2, MMP-9, COX-1, and COX-2 in patients with orofacial granulomatosis
Sun Jul 01 00:00:00 EDT 2007
The Bacteroides fragilis toxin binds to a specific intestinal epithelial cell receptor.
Infection and immunity
Wu S,Shin J,Zhang G,Cohen M,Franco A,Sears CL
Published figure using PAR2 monoclonal antibody (Product # 35-2300) in Immunofluorescence
Fri Sep 01 00:00:00 EDT 2006
Evidence that protease activated receptor 2 expression is enhanced in human coronary atherosclerotic lesions.
Journal of clinical pathology
Napoli C,de Nigris F,Wallace JL,Hollenberg MD,Tajana G,De Rosa G,Sica V,Cirino G
Published figure using PAR2 monoclonal antibody (Product # 35-2300) in Immunohistochemistry
Sat May 01 00:00:00 EDT 2004
Constitutive protease-activated receptor-2-mediated migration of MDA MB-231 breast cancer cells requires both beta-arrestin-1 and -2.
The Journal of biological chemistry
Ge L,Shenoy SK,Lefkowitz RJ,DeFea K
Fri Dec 31 00:00:00 EST 2004