Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 11
This antibody is specific for human K-Ras (K-Ras2, Ki-Ras, c-K-ras, GTPase KRas) protein (accession # NP_004976.2, P01116), which is 100% homologous with mouse, 95% with rat, and 94% with bovine, respectively. Reactivity has been confirmed on western blots with human HeLa and WI-38 cell lysates as well as rat KNRK and mouse NIH 3T3 cell lysates, and identifies the target band at~21 kDa. The reactivity has been also confirmed with rat KNRK cells using immunoprecipitation and with HeLa cells by immunofluorescence. Based on amino acid sequence homology, reactivity with bovine is also expected.
K-RAS (GTPase KRas) belongs to the Ras oncogene family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. It plays an important role in the regulation of cell proliferation and has a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes in colorectal cancer cells in a ZNF304-dependent manner. Mutations in the gene can result in leukemia acute myelogenous, leukemia juvenile myelomonocytic, Noonan Syndrome, gastric cancer, and cardiofaciocutaneous syndrome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-K-ras; c-K-ras2 protein; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; GTPase KRas; GTPase KRas, N-terminally processed; K RAS; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); oncogene KRAS2; p21 protein; PR310 c-K-ras oncogene; RASK; transforming protein p21; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog
Gene Aliases: AI929937; C-K-RAS; c-Ki-ras; CFC2; K-ras; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; Kras-2; KRAS1; KRAS2; NS; NS3; p21; p21B; RALD; ras; RASK2
UniProt ID: (Human) P01116, (Rat) P08644, (Mouse) P32883
Entrez Gene ID: (Human) 3845, (Rat) 24525, (Mouse) 16653
Molecular Function: small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support