Product References
Distinct niche structures and intrinsic programs of fallopian tube and ovarian surface epithelial cells.
iScience
Qin G,Park ES,Chen X,Han S,Xiang D,Ren F,Liu G,Chen H,Yuan GC,Li Z
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri Jan 20 00:00:00 EST 2023
The Wnt/TCF7L1 transcriptional repressor axis drives primitive endoderm formation by antagonizing naive and formative pluripotency.
Nature communications
Athanasouli P,Balli M,De Jaime-Soguero A,Boel A,Papanikolaou S,van der Veer BK,Janiszewski A,Vanhessche T,Francis A,El Laithy Y,Nigro AL,Aulicino F,Koh KP,Pasque V,Cosma MP,Verfaillie C,Zwijsen A,Heindryckx B,Nikolaou C,Lluis F
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2023
An estrogen-sensitive fibroblast population drives abdominal muscle fibrosis in an inguinal hernia mouse model.
JCI insight
Potluri T,Taylor MJ,Stulberg JJ,Lieber RL,Zhao H,Bulun SE
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Apr 19 00:00:00 EDT 2022
Hematopoietic-Mesenchymal Signals Regulate the Properties of Mesenchymal Stem Cells.
International journal of molecular sciences
Kanazawa S,Okada H,Riu D,Mabuchi Y,Akazawa C,Iwata J,Hoshi K,Hikita A
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Jul 26 00:00:00 EDT 2022
PEAR1 regulates expansion of activated fibroblasts and deposition of extracellular matrix in pulmonary fibrosis.
Nature communications
Geng Y,Li L,Yan J,Liu K,Yang A,Zhang L,Shen Y,Gao H,Wu X,Noth I,Huang Y,Liu J,Fan X
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Sat Nov 19 00:00:00 EST 2022
Lamin A/C-dependent chromatin architecture safeguards naïve pluripotency to prevent aberrant cardiovascular cell fate and function.
Nature communications
Wang Y,Elsherbiny A,Kessler L,Cordero J,Shi H,Serke H,Lityagina O,Trogisch FA,Mohammadi MM,El-Battrawy I,Backs J,Wieland T,Heineke J,Dobreva G
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri Nov 04 00:00:00 EDT 2022
Placental endocrine function shapes cerebellar development and social behavior.
Nature neuroscience
Vacher CM,Lacaille H,O'Reilly JJ,Salzbank J,Bakalar D,Sebaoui S,Liere P,Clarkson-Paredes C,Sasaki T,Sathyanesan A,Kratimenos P,Ellegood J,Lerch JP,Imamura Y,Popratiloff A,Hashimoto-Torii K,Gallo V,Schumacher M,Penn AA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Immunohistochemistry
Fri Oct 01 00:00:00 EDT 2021
IGF1R controls mechanosignaling in myofibroblasts required for pulmonary alveologenesis.
JCI insight
He H,Snowball J,Sun F,Na CL,Whitsett JA
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Mon Mar 22 00:00:00 EDT 2021
Interleukin-11-expressing fibroblasts have a unique gene signature correlated with poor prognosis of colorectal cancer.
Nature communications
Nishina T,Deguchi Y,Ohshima D,Takeda W,Ohtsuka M,Shichino S,Ueha S,Yamazaki S,Kawauchi M,Nakamura E,Nishiyama C,Kojima Y,Adachi-Akahane S,Hasegawa M,Nakayama M,Oshima M,Yagita H,Shibuya K,Mikami T,Inohara N,Matsushima K,Tada N,Nakano H
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Zbtb16 regulates social cognitive behaviors and neocortical development.
Translational psychiatry
Usui N,Berto S,Konishi A,Kondo M,Konopka G,Matsuzaki H,Shimada S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Immunohistochemistry
Sat Apr 24 00:00:00 EDT 2021
Gain-of-function p53 protein transferred via small extracellular vesicles promotes conversion of fibroblasts to a cancer-associated phenotype.
Cell reports
Ma S,McGuire MH,Mangala LS,Lee S,Stur E,Hu W,Bayraktar E,Villar-Prados A,Ivan C,Wu SY,Yokoi A,Dasari SK,Jennings NB,Liu J,Lopez-Berestein G,Ram P,Sood AK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2021
Extracellular vesicles synchronize cellular phenotypes of differentiating cells.
Journal of extracellular vesicles
Minakawa T,Matoba T,Ishidate F,Fujiwara TK,Takehana S,Tabata Y,Yamashita JK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 2021
Extracellular vesicles synchronize cellular phenotypes of differentiating cells.
Journal of extracellular vesicles
Minakawa T,Matoba T,Ishidate F,Fujiwara TK,Takehana S,Tabata Y,Yamashita JK
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 2021
Mesothelial cells are not a source of adipocytes in mice.
Cell reports
Westcott GP,Emont MP,Li J,Jacobs C,Tsai L,Rosen ED
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Jul 13 00:00:00 EDT 2021
Hyaluronan Degradation by Cemip Regulates Host Defense against Staphylococcus aureus Skin Infection.
Cell reports
Dokoshi T,Zhang LJ,Li F,Nakatsuji T,Butcher A,Yoshida H,Shimoda M,Okada Y,Gallo RL
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Jan 07 00:00:00 EST 2020
Tissue-Resident PDGFRα+ Progenitor Cells Contribute to Fibrosis versus Healing in a Context- and Spatiotemporally Dependent Manner.
Cell reports
Santini MP,Malide D,Hoffman G,Pandey G,D'Escamard V,Nomura-Kitabayashi A,Rovira I,Kataoka H,Ochando J,Harvey RP,Finkel T,Kovacic JC
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Loss of Fas signaling in fibroblasts impairs homeostatic fibrosis resolution and promotes persistent pulmonary fibrosis.
JCI insight
Redente EF,Chakraborty S,Sajuthi S,Black BP,Edelman BL,Seibold MA,Riches DW
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Adipocyte Piezo1 mediates obesogenic adipogenesis through the FGF1/FGFR1 signaling pathway in mice.
Nature communications
Wang S,Cao S,Arhatte M,Li D,Shi Y,Kurz S,Hu J,Wang L,Shao J,Atzberger A,Wang Z,Wang C,Zang W,Fleming I,Wettschureck N,Honoré E,Offermanns S
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri May 08 00:00:00 EDT 2020
Bim expression modulates the pro-inflammatory phenotype of retinal astroglial cells.
PloS one
Falero-Perez J,Sheibani N,Sorenson CM
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri Jul 31 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Interstitial cells of Cajal in Wsh/Wsh c-kit mutant mice.
Journal of smooth muscle research = Nihon Heikatsukin Gakkai kikanshi
Iino S,Horiguchi S,Horiguchi K,Hashimoto T
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Immunohistochemistry
Fri Jul 16 00:00:00 EDT 2021
Phosphatase SHP1 impedes mesenchymal stromal cell immunosuppressive capacity modulated by JAK1/STAT3 and P38 signals.
Cell & bioscience
Jiang M,Ye J,Wang X,Li N,Wang Y,Shi Y
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Thu Apr 14 00:00:00 EDT 2022
Dual lineage tracing identifies intermediate mesenchymal stage for endocardial contribution to fibroblasts, coronary mural cells, and adipocytes.
The Journal of biological chemistry
Huang X,Feng T,Jiang Z,Meng J,Kou S,Lu Z,Chen W,Lin CP,Zhou B,Zhang H
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Fri May 31 00:00:00 EDT 2019
Growth Factor Screening in Dystrophic Muscles Reveals PDGFB/PDGFRB-Mediated Migration of Interstitial Stem Cells.
International journal of molecular sciences
Camps J,Grosemans H,Gijsbers R,Maes C,Sampaolesi M
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Mar 05 00:00:00 EST 2019
A Brain-Melanocortin-Vagus Axis Mediates Adipose Tissue Expansion Independently of Energy Intake.
Cell reports
Holland J,Sorrell J,Yates E,Smith K,Arbabi S,Arnold M,Rivir M,Morano R,Chen J,Zhang X,Dimarchi R,Woods SC,Sanchez-Gurmaches J,Wohleb E,Perez-Tilve D
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue May 21 00:00:00 EDT 2019
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
The Isl1/Ldb1 Complex Orchestrates Genome-wide Chromatin Organization to Instruct Differentiation of Multipotent Cardiac Progenitors.
Cell stem cell
Caputo L,Witzel HR,Kolovos P,Cheedipudi S,Looso M,Mylona A,van IJcken WF,Laugwitz KL,Evans SM,Braun T,Soler E,Grosveld F,Dobreva G
Published figure using CD140a (PDGFRA) monoclonal antibody (Product # 46-1401-82) in Flow Cytometry
Thu Sep 03 00:00:00 EDT 2015