Product References
A bacterial autotransporter impairs innate immune responses by targeting the transcription factor TFE3.
Nature communications
Ta A,Ricci-Azevedo R,Vasudevan SO,Wright SS,Kumari P,Havira MS,Surendran Nair M,Rathinam VA,Vanaja SK
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Western Blot
Tue Apr 11 00:00:00 EDT 2023
P. gingivalis Infection Upregulates PD-L1 Expression on Dendritic Cells, Suppresses CD8+ T-cell Responses, and Aggravates Oral Cancer.
Cancer immunology research
Ren J,Han X,Lohner H,Hoyle RG,Li J,Liang S,Wang H
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
A small molecule binding HMGB1 inhibits caspase-11-mediated lethality in sepsis.
Cell death & disease
Wang X,Li Z,Bai Y,Zhang R,Meng R,Chen F,Wang H,Billiar TR,Xiao X,Lu B,Tang Y
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Western Blot
Wed Apr 14 00:00:00 EDT 2021
Macrophage mediated recognition and clearance of Borrelia burgdorferi elicits MyD88-dependent and -independent phagosomal signals that contribute to phagocytosis and inflammation.
BMC immunology
Benjamin SJ,Hawley KL,Vera-Licona P,La Vake CJ,Cervantes JL,Ruan Y,Radolf JD,Salazar JC
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunocytochemistry
Mon May 17 00:00:00 EDT 2021
Synaptojanin1 deficiency upregulates basal autophagosome formation in astrocytes.
The Journal of biological chemistry
Pan PY,Zhu J,Rizvi A,Zhu X,Tanaka H,Dreyfus CF
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunocytochemistry
Thu Jul 01 00:00:00 EDT 2021
An 8-Hydroxy-Quinoline Derivative Protects Against Lipopolysaccharide-Induced Lethality in Endotoxemia by Inhibiting HMGB1-Mediated Caspase-11 Signaling.
Frontiers in pharmacology
Wang X,Shi J,Li Z,Li L,Zhang R,Bai Y,Li J,Liang F,Tang Y
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Western Blot
Tue Jun 08 00:00:00 EDT 2021
Immunotherapeutic Blockade of CD47 Inhibitory Signaling Enhances Innate and Adaptive Immune Responses to Viral Infection.
Cell reports
Cham LB,Torrez Dulgeroff LB,Tal MC,Adomati T,Li F,Bhat H,Huang A,Lang PA,Moreno ME,Rivera JM,Galkina SA,Kosikova G,Stoddart CA,McCune JM,Myers LM,Weissman IL,Lang KS,Hasenkrug KJ
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Tue Apr 14 00:00:00 EDT 2020
Macropinocytosis drives T cell growth by sustaining the activation of mTORC1.
Nature communications
Charpentier JC,Chen D,Lapinski PE,Turner J,Grigorova I,Swanson JA,King PD
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunofluorescence
Fri Jan 10 00:00:00 EST 2020
25(OH) D3 alleviate liver NK cytotoxicity in acute but not in chronic fibrosis model of BALB/c mice due to modulations in vitamin D receptor.
BMC gastroenterology
Salhab A,Amer J,Yinying L,Safadi R
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Fri Apr 10 00:00:00 EDT 2020
Foam Cell Induction Activates AMPK But Uncouples Its Regulation of Autophagy and Lysosomal Homeostasis.
International journal of molecular sciences
LeBlond ND,Nunes JRC,Smith TKT,O'Dwyer C,Robichaud S,Gadde S,Côté M,Kemp BE,Ouimet M,Fullerton MD
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunocytochemistry
Fri Nov 27 00:00:00 EST 2020
Immunotherapy with engineered bacteria by targeting the STING pathway for anti-tumor immunity.
Nature communications
Leventhal DS,Sokolovska A,Li N,Plescia C,Kolodziej SA,Gallant CW,Christmas R,Gao JR,James MJ,Abin-Fuentes A,Momin M,Bergeron C,Fisher A,Miller PF,West KA,Lora JM
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunocytochemistry
Mon Jun 01 00:00:00 EDT 2020
Leishmania amazonensis hijacks host cell lysosomes involved in plasma membrane repair to induce invasion in fibroblasts.
Journal of cell science
Cavalcante-Costa VS,Costa-Reginaldo M,Queiroz-Oliveira T,Oliveira ACS,Couto NF,Dos Anjos DO,Lima-Santos J,Andrade LO,Horta MF,Castro-Gomes T
53-1071 was used in Immunocytochemistry-immunoflourescence to show that L. amazonensis can induce their own entry into fibroblasts independently of actin cytoskeleton activity, and, thus, through a mechanism that is distinct from phagocytosis.
Mon Mar 25 00:00:00 EDT 2019
CD1d highly expressed on DCs reduces lung tumor burden by enhancing antitumor immunity.
Oncology reports
Li Y,Zhao C,Liu J,Lu Z,Lu M,Gu J,Liu R
53-1071 was used in Flow cytometry/Cell sorting to investigate whether CD1d expression on dendritic cells is capable of enhancing anti-tumour immunity, particularly T-cell related immunity.
Wed May 01 00:00:00 EDT 2019
Bacteroides fragilis polysaccharide A induces IL-10 secreting B and T cells that prevent viral encephalitis.
Nature communications
Ramakrishna C,Kujawski M,Chu H,Li L,Mazmanian SK,Cantin EM
53-1071 was used in Flow cytometry/Cell sorting to study polysaccharide A as an immunomodulatory symbiosis factor whcih coordinates anti-inflammatory responses during viral infections.
Tue May 14 00:00:00 EDT 2019
Oncofetal gene SALL4 reactivation by hepatitis B virus counteracts miR-200c in PD-L1-induced T cell exhaustion.
Nature communications
Sun C,Lan P,Han Q,Huang M,Zhang Z,Xu G,Song J,Wang J,Wei H,Zhang J,Sun R,Zhang C,Tian Z
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Wed Mar 28 00:00:00 EDT 2018
A TLR7 agonist enhances the antitumor efficacy of obinutuzumab in murine lymphoma models via NK cells and CD4 T cells.
Leukemia
Cheadle EJ,Lipowska-Bhalla G,Dovedi SJ,Fagnano E,Klein C,Honeychurch J,Illidge TM
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
Cathepsin B modulates lysosomal biogenesis and host defense against Francisella novicida infection.
The Journal of experimental medicine
Qi X,Man SM,Malireddi RK,Karki R,Lupfer C,Gurung P,Neale G,Guy CS,Lamkanfi M,Kanneganti TD
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunofluorescence
Mon Sep 19 00:00:00 EDT 2016
Cathepsin B modulates lysosomal biogenesis and host defense against Francisella novicida infection.
The Journal of experimental medicine
Qi X,Man SM,Malireddi RK,Karki R,Lupfer C,Gurung P,Neale G,Guy CS,Lamkanfi M,Kanneganti TD
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunofluorescence
Mon Sep 19 00:00:00 EDT 2016
NK cell development requires Tsc1-dependent negative regulation of IL-15-triggered mTORC1 activation.
Nature communications
Yang M,Chen S,Du J,He J,Wang Y,Li Z,Liu G,Peng W,Zeng X,Li D,Xu P,Guo W,Chang Z,Wang S,Tian Z,Dong Z
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Wed Sep 07 00:00:00 EDT 2016
Intrahepatic innate lymphoid cells secrete IL-17A and IL-17F that are crucial for T cell priming in viral infection.
Journal of immunology (Baltimore, Md. : 1950)
Jie Z,Liang Y,Hou L,Dong C,Iwakura Y,Soong L,Cong Y,Sun J
53-1071 was used in Flow cytometry/Cell sorting to investigate the source and regulation of the IL-17 surge in the liver microenvironment that occurs early in the response to viral infection.
Tue Apr 01 00:00:00 EDT 2014
Tim-3 negatively mediates natural killer cell function in LPS-induced endotoxic shock.
PloS one
Hou H,Liu W,Wu S,Lu Y,Peng J,Zhu Y,Lu Y,Wang F,Sun Z
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2015
Role for E-cadherin as an inhibitory receptor on epidermal gammadelta T cells.
Journal of immunology (Baltimore, Md. : 1950)
Uchida Y,Kawai K,Ibusuki A,Kanekura T
53-1071 was used in Flow cytometry/Cell sorting to analyse functional differences between E-cadherin and α(E)β(7) integrin expressed on murine dendritic epidermal T cells.
Wed Jun 15 00:00:00 EDT 2011
The NALP3 inflammasome is involved in the innate immune response to amyloid-beta.
Nature immunology
Halle A,Hornung V,Petzold GC,Stewart CR,Monks BG,Reinheckel T,Fitzgerald KA,Latz E,Moore KJ,Golenbock DT
Published figure using CD107a (LAMP-1) monoclonal antibody (Product # 53-1071-82) in Immunofluorescence
Fri Aug 01 00:00:00 EDT 2008