Product References
Fluorescence-activated cell sorting and phenotypic characterization of human fibro-adipogenic progenitors.
STAR protocols
Billeskov TB,Jensen JB,Jessen N,Farup J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Mar 17 00:00:00 EDT 2023
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Mar 03 00:00:00 EST 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
LIM Mineralization Protein-1 Enhances the Committed Differentiation of Dental Pulp Stem Cells through the ERK1/2 and p38 MAPK Pathways and BMP Signaling.
International journal of medical sciences
Mu R,Chen B,Bi B,Yu H,Liu J,Li J,He M,Rong L,Liu B,Liu K,Zhu L,Shi X,Shuai Y,Jin L
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Aug 08 00:00:00 EDT 2022
Fused Cells between Human-Adipose-Derived Mesenchymal Stem Cells and Monocytes Keep Stemness Properties and Acquire High Mobility.
International journal of molecular sciences
Montalbán-Hernández K,Casado-Sánchez C,Avendaño-Ortiz J,Casalvilla-Dueñas JC,Bonel-Pérez GC,Prado-Montero J,Valentín-Quiroga J,Lozano-Rodríguez R,Terrón-Arcos V,de la Bastida FR,Córdoba L,Laso-García F,Diekhorst L,Del Fresno C,López-Collazo E
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Aug 26 00:00:00 EDT 2022
Cell-mimicking nanodecoys neutralize SARS-CoV-2 and mitigate lung injury in a non-human primate model of COVID-19.
Nature nanotechnology
Li Z,Wang Z,Dinh PC,Zhu D,Popowski KD,Lutz H,Hu S,Lewis MG,Cook A,Andersen H,Greenhouse J,Pessaint L,Lobo LJ,Cheng K
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Exosomes Regulate Interclonal Communication on Osteogenic Differentiation Among Heterogeneous Osteogenic Single-Cell Clones Through PINK1/Parkin-Mediated Mitophagy.
Frontiers in cell and developmental biology
Fei D,Xia Y,Zhai Q,Wang Y,Zhou F,Zhao W,He X,Wang Q,Jin Y,Li B
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Oct 05 00:00:00 EDT 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Aug 31 00:00:00 EDT 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Jul 15 00:00:00 EDT 2021
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Nov 19 00:00:00 EST 2020
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 EST 2019
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 EST 2019
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 EST 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2019
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
CD90 promotes cell migration, viability and sphere‑forming ability of hepatocellular carcinoma cells.
International journal of molecular medicine
Zhang K,Che S,Su Z,Zheng S,Zhang H,Yang S,Li W,Liu J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Feb 05 00:00:00 EST 2018
Endothelial and smooth muscle cells derived from human cardiac explants demonstrate angiogenic potential and suitable for design of cell-containing vascular grafts.
Journal of translational medicine
Zakharova IS,Zhiven' MK,Saaya SB,Shevchenko AI,Smirnova AM,Strunov A,Karpenko AA,Pokushalov EA,Ivanova LN,Makarevich PI,Parfyonova YV,Aboian E,Zakian SM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Fri Mar 03 00:00:00 EST 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2017
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Jan 19 00:00:00 EST 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Immunophenotypic comparison of heterogenous non-sorted versus sorted mononuclear cells from human umbilical cord blood: a novel cell enrichment approach.
Cytotechnology
Indumathi S,Harikrishnan R,Rajkumar JS,Dhanasekaran M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Thu Dec 11 00:00:00 EST 2014
PDGFR-β (+) perivascular cells from infantile hemangioma display the features of mesenchymal stem cells and show stronger adipogenic potential in vitro and in vivo.
International journal of clinical and experimental pathology
Yuan SM,Guo Y,Zhou XJ,Shen WM,Chen HN
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 62-0909-42) in Flow Cytometry
Tue May 26 00:00:00 EDT 2015