Product References
Cissus quadrangularis (Hadjod) Inhibits RANKL-Induced Osteoclastogenesis and Augments Bone Health in an Estrogen-Deficient Preclinical Model of Osteoporosis Via Modulating the Host Osteoimmune System.
Cells
Azam Z,Sapra L,Baghel K,Sinha N,Gupta RK,Soni V,Saini C,Mishra PK,Srivastava RK
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jan 04 00:00:00 EST 2023
Role of FGF10/FGFR2b Signaling in Homeostasis and Regeneration of Adult Lacrimal Gland and Corneal Epithelium Proliferation.
Investigative ophthalmology & visual science
Finburgh EN,Mauduit O,Noguchi T,Bu JJ,Abbas AA,Hakim DF,Bellusci S,Meech R,Makarenkova HP,Afshari NA
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Jan 03 00:00:00 EST 2023
High-yield enrichment of mouse small intestine intraepithelial lymphocytes by immunomagnetic depletion of EpCAM+ cells.
STAR protocols
Prakhar P,Gonzalez V,Park JH
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Fri Mar 18 00:00:00 EDT 2022
Single-cell transcriptomics reveals distinct cell response between acute and chronic pulmonary infection of Pseudomonas aeruginosa.
MedComm
Hu X,Wu M,Ma T,Zhang Y,Zou C,Wang R,Zhang Y,Ren Y,Li Q,Liu H,Li H,Wang T,Sun X,Yang Y,Tang M,Li X,Li J,Gao X,Li T,Zhou X
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Immunohistochemistry
Thu Dec 01 00:00:00 EST 2022
Valosin-containing protein (VCP/p97) inhibition reduces viral clearance and induces toxicity associated with muscular damage.
Cell death & disease
Del Rio Oliva M,Basler M
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
The ubiquitin ligase Peli1 inhibits ICOS and thereby Tfh-mediated immunity.
Cellular & molecular immunology
Huang X,Hao S,Liu J,Huang Y,Liu M,Xiao C,Wang Y,Pei S,Yu T,Xu J,Wang H,Dai D,Su X,Xiao Y
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Functional human IgA targets a conserved site on malaria sporozoites.
Science translational medicine
Tan J,Cho H,Pholcharee T,Pereira LS,Doumbo S,Doumtabe D,Flynn BJ,Schön A,Kanatani S,Aylor SO,Oyen D,Vistein R,Wang L,Dillon M,Skinner J,Peterson M,Li S,Idris AH,Molina-Cruz A,Zhao M,Olano LR,Lee PJ,Roth A,Sinnis P,Barillas-Mury C,Kayentao K,Ongoiba A,Francica JR,Traore B,Wilson IA,Seder RA,Crompton PD
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jun 23 00:00:00 EDT 2021
Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
The Journal of experimental medicine
Heindl S,Ricci A,Carofiglio O,Zhou Q,Arzberger T,Lenart N,Franzmeier N,Hortobagyi T,Nelson PT,Stowe AM,Denes A,Edbauer D,Liesz A
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
Aged hematopoietic stem cells are refractory to bloodborne systemic rejuvenation interventions.
The Journal of experimental medicine
Ho TT,Dellorusso PV,Verovskaya EV,Bakker ST,Flach J,Smith LK,Ventura PB,Lansinger OM,Hérault A,Zhang SY,Kang YA,Mitchell CA,Villeda SA,Passegué E
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
The gammaherpesvirus 68 viral cyclin facilitates expression of LANA.
PLoS pathogens
Niemeyer BF,Sanford B,Gibson JE,Berger JN,Oko LM,Medina E,Clambey ET,van Dyk LF
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Jan 24 00:00:00 EST 2022
Regulatory B Cells (Bregs) Inhibit Osteoclastogenesis and Play a Potential Role in Ameliorating Ovariectomy-Induced Bone Loss.
Frontiers in immunology
Sapra L,Bhardwaj A,Mishra PK,Garg B,Verma B,Mishra GC,Srivastava RK
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Nov 04 00:00:00 EDT 2021
Comprehensive mutagenesis identifies the peptide repertoire of a p53 T-cell receptor mimic antibody that displays no toxicity in mice transgenic for human HLA-A*0201.
PloS one
Trenevska I,Anderson AP,Bentley C,Hassanali T,Wiblin S,Maguire S,Pezzella F,Banham AH,Li D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Sep 29 00:00:00 EDT 2021
Calibration of cell-intrinsic interleukin-2 response thresholds guides design of a regulatory T cell biased agonist.
eLife
Glassman CR,Su L,Majri-Morrison SS,Winkelmann H,Mo F,Li P,Pérez-Cruz M,Ho PP,Koliesnik I,Nagy N,Hnizdilova T,Picton LK,Kovar M,Bollyky P,Steinman L,Meyer E,Piehler J,Leonard WJ,Garcia KC
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue May 18 00:00:00 EDT 2021
Excessive Costimulation Leads to Dysfunction of Adoptively Transferred T Cells.
Cancer immunology research
Wijewarnasuriya D,Bebernitz C,Lopez AV,Rafiq S,Brentjens RJ
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
Zdhhc2 Is Essential for Plasmacytoid Dendritic Cells Mediated Inflammatory Response in Psoriasis.
Frontiers in immunology
Zhou B,Yang W,Li W,He L,Lu L,Zhang L,Liu Z,Wang Y,Chao T,Huang R,Gu Y,Jia T,Liu Q,Tian S,Pierre P,Maeda T,Liang Y,Kong E
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jun 23 00:00:00 EDT 2021
Memory CD73+IgM+ B cells protect against Plasmodium yoelii infection and express Granzyme B.
PloS one
Parra M,Weitner M,Yang A,Akue A,Liu X,Schmidt T,Allman WR,Akkoyunlu M,Derrick SC
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Oct 27 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Tregitopes regulate the tolerogenic immune response and decrease the foetal death rate in abortion-prone mouse matings.
Scientific reports
Kedzierska AE,Lorek D,Slawek A,Chelmonska-Soyta A
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Jun 29 00:00:00 EDT 2020
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Jun 17 00:00:00 EDT 2021
Distinct Transcriptional Responses across Tissue-Resident Macrophages to Short-Term and Long-Term Metabolic Challenge.
Cell reports
Brykczynska U,Geigges M,Wiedemann SJ,Dror E,Böni-Schnetzler M,Hess C,Donath MY,Paro R
63-0193 was used in Flow cytometry/Cell sorting to provide a resource for assessing the impact of nutrition and over-nutrition on the spectrum of macrophages across tissues with a potential for identification of systemic responses.
Tue Feb 04 00:00:00 EST 2020
NLRX1 inhibits the early stages of CNS inflammation and prevents the onset of spontaneous autoimmunity.
PLoS biology
Gharagozloo M,Mahmoud S,Simard C,Yamamoto K,Bobbala D,Ilangumaran S,Smith MD,Lamontagne A,Jarjoura S,Denault JB,Blais V,Gendron L,Vilariño-Güell C,Sadovnick AD,Ting JP,Calabresi PA,Amrani A,Gris D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Peritoneal Cells Mediate Immune Responses and Cross-Protection Against Influenza A Virus.
Frontiers in immunology
Gautam A,Park BK,Kim TH,Akauliya M,Kim D,Maharjan S,Park J,Kim J,Lee H,Park MS,Lee Y,Kwon HJ
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Aug 17 00:00:00 EDT 2020
Variation in zygotic CRISPR/Cas9 gene editing outcomes generates novel reporter and deletion alleles at the Gdf11 locus.
Scientific reports
Goldstein JM,Valido A,Lewandowski JP,Walker RG,Mills MJ,Messemer KA,Besseling P,Lee KH,Wattrus SJ,Cho M,Lee RT,Wagers AJ
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Dec 09 00:00:00 EST 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
TET2 Deficiency Causes Germinal Center Hyperplasia, Impairs Plasma Cell Differentiation, and Promotes B-cell Lymphomagenesis.
Cancer discovery
Dominguez PM,Ghamlouch H,Rosikiewicz W,Kumar P,Béguelin W,Fontán L,Rivas MA,Pawlikowska P,Armand M,Mouly E,Torres-Martin M,Doane AS,Calvo Fernandez MT,Durant M,Della-Valle V,Teater M,Cimmino L,Droin N,Tadros S,Motanagh S,Shih AH,Rubin MA,Tam W,Aifantis I,Levine RL,Elemento O,Inghirami G,Green MR,Figueroa ME,Bernard OA,Aoufouchi S,Li S,Shaknovich R,Melnick AM
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
11 Beta-hydroxysteroid dehydrogenase type 1 regulates synovitis, joint destruction, and systemic bone loss in chronic polyarthritis.
Journal of autoimmunity
Hardy RS,Fenton C,Croft AP,Naylor AJ,Begum R,Desanti G,Buckley CD,Lavery G,Cooper MS,Raza K
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
The Role of Allograft Inflammatory Factor-1 in the Effects of Experimental Diabetes on B Cell Functions in the Heart.
Frontiers in cardiovascular medicine
Sarkar A,Shukla SK,Alqatawni A,Kumar A,Addya S,Tsygankov AY,Rafiq K
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Dec 06 00:00:00 EST 2021
In Vitro Differentiation of Gata2 and Ly6a Reporter Embryonic Stem Cells Corresponds to In Vivo Waves of Hematopoietic Cell Generation.
Stem cell reports
Kauts ML,Rodriguez-Seoane C,Kaimakis P,Mendes SC,Cortés-Lavaud X,Hill U,Dzierzak E
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Jan 09 00:00:00 EST 2018
Peli1 negatively regulates noncanonical NF-κB signaling to restrain systemic lupus erythematosus.
Nature communications
Liu J,Huang X,Hao S,Wang Y,Liu M,Xu J,Zhang X,Yu T,Gan S,Dai D,Luo X,Lu Q,Mao C,Zhang Y,Shen N,Li B,Huang M,Zhu X,Jin J,Cheng X,Sun SC,Xiao Y
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Mon Mar 19 00:00:00 EDT 2018
Differential requirement of kindlin-3 for T cell progenitor homing to the non-vascularized and vascularized thymus.
eLife
Moretti FA,Klapproth S,Ruppert R,Margraf A,Weber J,Pick R,Scheiermann C,Sperandio M,Fässler R,Moser M
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Sep 06 00:00:00 EDT 2018
A protocol for generating induced T cells by reprogramming B cells in vivo.
Cell regeneration (London, England)
Weng Q,Hu F,Zhang M,Dong Y,Lv C,Wang Y,Liu X,Wang J
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
Direct conversion of injury-site myeloid cells to fibroblast-like cells of granulation tissue.
Nature communications
Sinha M,Sen CK,Singh K,Das A,Ghatak S,Rhea B,Blackstone B,Powell HM,Khanna S,Roy S
63-0193 was used in Flow cytometry/Cell sorting to introduce a paradigm wherein myeloid cells are recognized as a major source of fibroblast-like cells in granulation tissue.
Mon Mar 05 00:00:00 EST 2018
A therapeutic T cell receptor mimic antibody targets tumor-associated PRAME peptide/HLA-I antigens.
The Journal of clinical investigation
Chang AY,Dao T,Gejman RS,Jarvis CA,Scott A,Dubrovsky L,Mathias MD,Korontsvit T,Zakhaleva V,Curcio M,Hendrickson RC,Liu C,Scheinberg DA
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Fri Jun 30 00:00:00 EDT 2017
Prolonged Deleterious Influences of Chemotherapeutic Agent CPT-11 on Resident Peritoneal Macrophages and B1 Cells.
Frontiers in immunology
Bai WJ,Li CG,Zhang CC,Xu LH,Zeng QZ,Hu B,Hong Z,He XH,Ouyang DY
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
ADAR1 deletion induces NFκB and interferon signaling dependent liver inflammation and fibrosis.
RNA biology
Ben-Shoshan SO,Kagan P,Sultan M,Barabash Z,Dor C,Jacob-Hirsch J,Harmelin A,Pappo O,Marcu-Malina V,Ben-Ari Z,Amariglio N,Rechavi G,Goldstein I,Safran M
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu May 04 00:00:00 EDT 2017
Diminished bone regeneration after debridement of posttraumatic osteomyelitis is accompanied by altered cytokine levels, elevated B cell activity, and increased osteoclast activity.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Wagner JM,Jaurich H,Wallner C,Abraham S,Becerikli M,Dadras M,Harati K,Duhan V,Khairnar V,Lehnhardt M,Behr B
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Intravenously usable fully serotype 3 oncolytic adenovirus coding for CD40L as an enabler of dendritic cell therapy.
Oncoimmunology
Zafar S,Parviainen S,Siurala M,Hemminki O,Havunen R,Tähtinen S,Bramante S,Vassilev L,Wang H,Lieber A,Hemmi S,de Gruijl T,Kanerva A,Hemminki A
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
IL35-Producing B Cells Promote the Development of Pancreatic Neoplasia.
Cancer discovery
Pylayeva-Gupta Y,Das S,Handler JS,Hajdu CH,Coffre M,Koralov SB,Bar-Sagi D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Medial HOXA genes demarcate haematopoietic stem cell fate during human development.
Nature cell biology
Dou DR,Calvanese V,Sierra MI,Nguyen AT,Minasian A,Saarikoski P,Sasidharan R,Ramirez CM,Zack JA,Crooks GM,Galic Z,Mikkola HK
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Antigen Targeting to Human HLA Class II Molecules Increases Efficacy of DNA Vaccination.
Journal of immunology (Baltimore, Md. : 1950)
Grodeland G,Fredriksen AB,Løset GÅ,Vikse E,Fugger L,Bogen B
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2016
CD19 CAR immune pressure induces B-precursor acute lymphoblastic leukaemia lineage switch exposing inherent leukaemic plasticity.
Nature communications
Jacoby E,Nguyen SM,Fountaine TJ,Welp K,Gryder B,Qin H,Yang Y,Chien CD,Seif AE,Lei H,Song YK,Khan J,Lee DW,Mackall CL,Gardner RA,Jensen MC,Shern JF,Fry TJ
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jul 27 00:00:00 EDT 2016
Novel CXCL13 transgenic mouse: inflammation drives pathogenic effect of CXCL13 in experimental myasthenia gravis.
Oncotarget
Weiss JM,Robinet M,Aricha R,Cufi P,Villeret B,Lantner F,Shachar I,Fuchs S,Souroujon MC,Berrih-Aknin S,Le Panse R
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Feb 16 00:00:00 EST 2016
Protein Kinase D2 Protects against Acute Colitis Induced by Dextran Sulfate Sodium in Mice.
Scientific reports
Xiong J,Zhou MF,Wang YD,Chen LP,Xu WF,Wang YD,Deng F,Liu SD
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Fri Sep 23 00:00:00 EDT 2016
Cis-regulatory mechanisms governing stem and progenitor cell transitions.
Science advances
Johnson KD,Kong G,Gao X,Chang YI,Hewitt KJ,Sanalkumar R,Prathibha R,Ranheim EA,Dewey CN,Zhang J,Bresnick EH
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Role of caveolin-3 in lymphocyte activation.
Life sciences
Tran C,Stary CM,Schilling JM,Bentley B,Patel HH,Roth DM
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Thu Jan 15 00:00:00 EST 2015
B cell Rab7 mediates induction of activation-induced cytidine deaminase expression and class-switching in T-dependent and T-independent antibody responses.
Journal of immunology (Baltimore, Md. : 1950)
Pone EJ,Lam T,Lou Z,Wang R,Chen Y,Liu D,Edinger AL,Xu Z,Casali P
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
A novel method for autophagy detection in primary cells: impaired levels of macroautophagy in immunosenescent T cells.
Autophagy
Phadwal K,Alegre-Abarrategui J,Watson AS,Pike L,Anbalagan S,Hammond EM,Wade-Martins R,McMichael A,Klenerman P,Simon AK
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2012
Critical role of B cell lymphoma 10 in BAFF-regulated NF-κB activation and survival of anergic B cells.
Journal of immunology (Baltimore, Md. : 1950)
Yu M,Chen Y,He Y,Podd A,Fu G,Wright JA,Kleiman E,Khan WN,Wen R,Wang D
Published figure using CD19 monoclonal antibody (Product # 63-0193-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2012