Product References
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Thu Mar 03 00:00:00 EST 2022
Production enhancement of human adipose-derived mesenchymal stem cells by low-intensity ultrasound stimulation.
Scientific reports
Min S,Byeon Y,Kim M,Lee Y,Lee SH,Lee Y,Farooqi HMU,Lee HK,Paeng DG
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2022
Isolation and characterization mesenchymal stem cells from red panda (Ailurus fulgens styani) endometrium.
Conservation physiology
Wang DH,Wu XM,Chen JS,Cai ZG,An JH,Zhang MY,Li Y,Li FP,Hou R,Liu YL
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Sat Jan 01 00:00:00 EST 2022
Induced Neurodifferentiation of hBM-MSCs through Activation of the ERK/CREB Pathway via Pulsed Electromagnetic Fields and Physical Stimulation Promotes Neurogenesis in Cerebral Ischemic Models.
International journal of molecular sciences
Park HJ,Choi JH,Nam MH,Seo YK
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Jan 21 00:00:00 EST 2022
Mesenchymal stem cells derived from patients with premature aging syndromes display hallmarks of physiological aging.
Life science alliance
Trani JP,Chevalier R,Caron L,El Yazidi C,Broucqsault N,Toury L,Thomas M,Annab K,Binetruy B,De Sandre-Giovannoli A,Levy N,Magdinier F,Robin JD
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Sep 14 00:00:00 EDT 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2022
The Effect of Different Doses of Melatonin on in Vitro Maturation of Human Follicular Fluid-Derived Oocyte-Like Cells.
JBRA assisted reproduction
Azandeh S,Moghadam MT,Rashno M,Zargar M,Zadeh PA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Immunocytochemistry
Sun Apr 17 00:00:00 EDT 2022
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Tue Aug 31 00:00:00 EDT 2021
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy
Güney-Esken G,Erol ÖD,Pervin B,Gürhan Sevinç G,Önder T,Bilgiç E,Korkusuz P,Günel-Özcan A,Uçkan-Çetinkaya D,Aerts-Kaya F
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Thu May 13 00:00:00 EDT 2021
Human Amniotic Mesenchymal Stem Cells Inhibit aGVHD by Regulating Balance of Treg and T Effector Cells.
Journal of inflammation research
Gao Y,Li W,Bu X,Xu Y,Cai S,Zhong J,Du M,Sun H,Huang L,He Y,Hu X,Liu Q,Jin H,Wang Q,Ping B
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Tue Apr 26 00:00:00 EDT 2022
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Thu Jul 15 00:00:00 EDT 2021
ZKSCAN3 counteracts cellular senescence by stabilizing heterochromatin.
Nucleic acids research
Hu H,Ji Q,Song M,Ren J,Liu Z,Wang Z,Liu X,Yan K,Hu J,Jing Y,Wang S,Zhang W,Liu GH,Qu J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Rescue of premature aging defects in Cockayne syndrome stem cells by CRISPR/Cas9-mediated gene correction.
Protein & cell
Wang S,Min Z,Ji Q,Geng L,Su Y,Liu Z,Hu H,Wang L,Zhang W,Suzuiki K,Huang Y,Zhang P,Tang TS,Qu J,Yu Y,Liu GH,Qiao J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Thu Nov 19 00:00:00 EST 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Nov 27 00:00:00 EST 2020
Up-regulation of FOXD1 by YAP alleviates senescence and osteoarthritis.
PLoS biology
Fu L,Hu Y,Song M,Liu Z,Zhang W,Yu FX,Wu J,Wang S,Izpisua Belmonte JC,Chan P,Qu J,Tang F,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2019
Differential stem cell aging kinetics in Hutchinson-Gilford progeria syndrome and Werner syndrome.
Protein & cell
Wu Z,Zhang W,Song M,Wang W,Wei G,Li W,Lei J,Huang Y,Sang Y,Chan P,Chen C,Qu J,Suzuki K,Belmonte JCI,Liu GH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Antagonizing CD105 enhances radiation sensitivity in prostate cancer.
Oncogene
Madhav A,Andres A,Duong F,Mishra R,Haldar S,Liu Z,Angara B,Gottlieb R,Zumsteg ZS,Bhowmick NA
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Utilizing Confocal Microscopy to Characterize Human and Mouse Adipose Tissue.
Tissue engineering. Part C, Methods
Blackshear CP,Borrelli MR,Shen EZ,Ransom RC,Chung NN,Vistnes SM,Irizarry D,Nazerali R,Momeni A,Longaker MT,Wan DC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Immunohistochemistry
Mon Oct 01 00:00:00 EDT 2018
Utilizing Confocal Microscopy to Characterize Human and Mouse Adipose Tissue.
Tissue engineering. Part C, Methods
Blackshear CP,Borrelli MR,Shen EZ,Ransom RC,Chung NN,Vistnes SM,Irizarry D,Nazerali R,Momeni A,Longaker MT,Wan DC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Immunohistochemistry
Mon Oct 01 00:00:00 EDT 2018
The Improvement of Respiratory Performance After Phototherapy-Induced EPC Mobilization in Preterm Infants With RDS.
Journal of cellular biochemistry
Siavashi V,Asadian S,Taheri-Asl M,Babaei H,Keshavarz S,Bazaei M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Circulation Enrichment of Functional Endothelial Progenitor Cells by Infantile Phototherapy.
Journal of cellular biochemistry
Siavashi V,Asadian S,Sharifi A,Esmaeilivand M,Norouzinia R,Azadbakht M,Nassiri SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 63-1057-42) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020