Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arsenical pump-driving ATPase; Arsenite-stimulated ATPase; arsenite-translocating ATPase; ATPase ASNA1; ATPase GET3; Guided entry of tail-anchored proteins factor 3, ATPase; hARSA-I; hASNA-I; MGC3821; Transmembrane domain recognition complex 40 kDa ATPase subunit
Gene Aliases: 1810048H22Rik; ARSA; ARSA-I; ARSA1; ASNA-I; ASNA1; GET3; TRC40
UniProt ID: (Human) O43681, (Mouse) O54984, (Rat) G3V9T7
Entrez Gene ID: (Human) 439, (Mouse) 56495, (Rat) 288919
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support