Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-06304 detects E-cadherin in human samples.
MA1-06304 has sucessfully been used in Western blotting. By Western blotting, it detects a ~120 kDa protein representing E-cadherin.
The MA1-06304 immunogen is a synthetic peptide corresponding to residues P(30) G F D A E S Y T F T V P R(43) of mature human E-cadherin.
Store at 4°C or in small aliquots at -20°C.
E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cadherin 1, E-cadherin (epithelial); cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; Cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; cell-CAM 120/80; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; E-Cadherin; E-cadherin 1; E-Cadherin phospho; Ecadherin; Epithelial cadherin; phospho cadherin 1; phospho E-Cadherin; phospho LCAM; phospho S838; phospho UVO; S840; S840 E-Cadherin; Ser840; Ser840 E-Cadherin; Uvomorulin
Gene Aliases: CDH1; CDHE; UVO
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support