Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 13
This antibody recognizes CREB protein phosphorylated at serine 133 and ATF1 phosphorylated at serine 63. The antibody was purified from serum-free cell culture supernatant by subsequent thiophilic adsorption and size exclusion chromatography.
Aliquot and freeze in liquid nitrogen. Antibody can be stored frozen at -80ºC for up to 1 year. Thaw aliquots at 37ºC; thawed aliquots may be stored at 4ºC for up to 3 months. Avoid repeated freeze/thaw cycles.
This antibody was originally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Cyclic AMP response element binding protein (CREB) is a basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation occurs via p44/42 MAP kinase and p90RSK and also via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB appears to play an important role in learning and memory.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating transcription factor 1; active transcription factor CREB; cAMP-dependent transcription factor ATF-1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; Cyclic AMP-dependent transcription factor ATF-1; Cyclic AMP-responsive element-binding protein 1; Protein TREB36; Ser133; TCR-ATF1; transactivator protein
Gene Aliases: 2310001E10Rik; 3526402H21Rik; ATF1; AV083133; CREB; CREB-1; CREB1; EWS-ATF1; FUS/ATF-1; TREB36
UniProt ID: (Human) P18846, (Mouse) P81269, (Human) P21934, (Mouse) Q01147
Entrez Gene ID: (Dog) 486545, (Human) 466, (Mouse) 11908, (Dog) 607922, (Human) 1385, (Mouse) 12912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support