Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-33833 detects CD79b from mouse samples.
MA1-33833 has been successfully used in immunohistochemistry (frozen tissue), immunoprecipitation, and Western blot applications. Use 10 ul of the suggested working dilution to label 10^6 cells or 100 ul.
The MA1-33833 immunogen is CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells.
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; Ig beta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7
Gene Aliases: Cd79b; Igb
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support