Product References
A nanofiber-hydrogel composite improves tissue repair in a rat model of Crohn's disease perianal fistulas.
Science advances
Li L,Yao ZC,Parian A,Yang YH,Chao J,Yin J,Salimian KJ,Reddy SK,Zaheer A,Gearhart SL,Mao HQ,Selaru FM
MA5-11532 was used in Immunohistochemistry-immunofluorescence to demonstrate that ADSC-loaded mfNHC is a promising therapy for CD-PAF, and warrants further studies to advance mfNHC toward clinical translation.
Wed Jan 04 00:00:00 EST 2023
Lung Inflammation in alpha-1-antitrypsin deficient individuals with normal lung function.
Respiratory research
Kokturk N,Khodayari N,Lascano J,Riley EL,Brantly ML
MA5-11532 was used in Immunohistochemistry to investigate the characteristics of lower respiratory tract of AATD individuals prior to the onset of clinically significant COPD.
Thu Feb 02 00:00:00 EST 2023
PBRM1 loss defines a nonimmunogenic tumor phenotype associated with checkpoint inhibitor resistance in renal carcinoma.
Nature communications
Liu XD,Kong W,Peterson CB,McGrail DJ,Hoang A,Zhang X,Lam T,Pilie PG,Zhu H,Beckermann KE,Haake SM,Isgandrova S,Martinez-Moczygemba M,Sahni N,Tannir NM,Lin SY,Rathmell WK,Jonasch E
Published figure using CD45RO monoclonal antibody (Product # MA5-11532) in Immunohistochemistry
Fri May 01 00:00:00 EDT 2020
PBRM1 loss defines a nonimmunogenic tumor phenotype associated with checkpoint inhibitor resistance in renal carcinoma.
Nature communications
Liu XD,Kong W,Peterson CB,McGrail DJ,Hoang A,Zhang X,Lam T,Pilie PG,Zhu H,Beckermann KE,Haake SM,Isgandrova S,Martinez-Moczygemba M,Sahni N,Tannir NM,Lin SY,Rathmell WK,Jonasch E
Published figure using CD45RO monoclonal antibody (Product # MA5-11532) in Immunohistochemistry
Fri May 01 00:00:00 EDT 2020
A case report of clonal EBV-like memory CD4+ T cell activation in fatal checkpoint inhibitor-induced encephalitis.
Nature medicine
Johnson DB,McDonnell WJ,Gonzalez-Ericsson PI,Al-Rohil RN,Mobley BC,Salem JE,Wang DY,Sanchez V,Wang Y,Chastain CA,Barker K,Liang Y,Warren S,Beechem JM,Menzies AM,Tio M,Long GV,Cohen JV,Guidon AC,O'Hare M,Chandra S,Chowdhary A,Lebrun-Vignes B,Goldinger SM,Rushing EJ,Buchbinder EI,Mallal SA,Shi C,Xu Y,Moslehi JJ,Sanders ME,Sosman JA,Balko JM
MA5-11532 was used in Immunohistochemistry to identify if CD4+ and CD8+ T cells are potential culprits of checkpoint inhibitor-associated immune encephalitis.
Thu Aug 01 00:00:00 EDT 2019
Nephrogenic Systemic Fibrosis Is Mediated by Myeloid C-C Chemokine Receptor 2.
The Journal of investigative dermatology
Do C,Drel V,Tan C,Lee D,Wagner B
MA5-11532 was used in Immunocytochemistry-immunoflourescence to establish a mouse model of nephrogenic systemic fibrosis.
Tue Oct 01 00:00:00 EDT 2019
Gadolinium-based contrast agents: Stimulators of myeloid-induced renal fibrosis and major metabolic disruptors.
Toxicology and applied pharmacology
Do C,Ford B,Lee DY,Tan C,Escobar P,Wagner B
MA5-11532 was used in Immunohistochemistry to examine the consequences of GBCA treatment in the kidneys from mice with normal kidney function and the potential interplay between obesity and gadolinium exposure.
Mon Jul 15 00:00:00 EDT 2019
The absence of HLA class I expression in non-small cell lung cancer correlates with the tumor tissue structure and the pattern of T cell infiltration.
International journal of cancer
Perea F,Bernal M,Sánchez-Palencia A,Carretero J,Torres C,Bayarri C,Gómez-Morales M,Garrido F,Ruiz-Cabello F
MA511532 was used in immunohistochemistry - frozen section to characterize HLA class I-positive and negative tumors
Wed Feb 15 00:00:00 EST 2017
Trichinella Spiralis Impact on Mesenchymal Stem Cells: Immunohistochemical Study by Image Analyzer in Murine Model.
Experimental and molecular pathology
Hasby Saad MA,Hasby EA
MA511532 was used in immunohistochemistry - paraffin section to test if Trichinella spiralis infection or its derivative stimulate recruitment of CD105+/CD45- cells
Thu Jun 01 00:00:00 EDT 2017
CD103+ intraepithelial T cells in high-grade serous ovarian cancer are phenotypically diverse TCRαβ+ CD8αβ+ T cells that can be targeted for cancer immunotherapy.
Oncotarget
Komdeur FL,Wouters MC,Workel HH,Tijans AM,Terwindt AL,Brunekreeft KL,Plat A,Klip HG,Eggink FA,Leffers N,Helfrich W,Samplonius DF,Bremer E,Wisman GB,Daemen T,Duiker EW,Hollema H,Nijman HW,de Bruyn M
MA511532 was used in flow cytometry to define the contribution of CD103+ tumor-infiltrating lymphocytes to high-grade serous epithelial ovarian cancer
Tue Nov 15 00:00:00 EST 2016
Resistance to Antiangiogenic Therapy Is Associated with an Immunosuppressive Tumor Microenvironment in Metastatic Renal Cell Carcinoma.
Cancer immunology research
Liu XD,Hoang A,Zhou L,Kalra S,Yetil A,Sun M,Ding Z,Zhang X,Bai S,German P,Tamboli P,Rao P,Karam JA,Wood C,Matin S,Zurita A,Bex A,Griffioen AW,Gao J,Sharma P,Tannir N,Sircar K,Jonasch E
Published figure using CD45RO monoclonal antibody (Product # MA5-11532) in Immunohistochemistry
Tue Sep 01 00:00:00 EDT 2015
High frequency of CD8 positive lymphocyte infiltration correlates with lack of lymph node involvement in early rectal cancer.
Disease markers
Däster S,Eppenberger-Castori S,Hirt C,Zlobec I,Delko T,Nebiker CA,Soysal SD,Amicarella F,Iezzi G,Sconocchia G,Heberer M,Lugli A,Spagnoli GC,Kettelhack C,Terracciano L,Oertli D,von Holzen U,Tornillo L,Droeser RA
MA5-11532 was used in Immunohistochemistry to determine the efficacy of immunoprofiling in predicting lymph node involvement in early rectal cancer.
Fri Jul 31 00:00:00 EDT 2015
CD30+ extranodal natural killer/T-cell lymphoma mimicking phlegmonous myositis: A case report.
Oncology letters
Yang YJ,Li YX,Liu YB,Yang M,Liu K
MA5-11532 was used in immunohistochemistry (paraffin) to report on a case of CD30-positive extranodal natural killer/T-cell lymphoma.
Thu May 01 00:00:00 EDT 2014
A giant primary sclerosing lipogranuloma of the scrotum.
Turk patoloji dergisi
Mungan S,Karagüzel E,Turan C,Reıs A
MA5-11532 was used in immunohistochemistry to report on a case of giant primary sclerosing lipogranuloma of the scrotum
Tue Jul 08 00:00:00 EDT 2014
IL-21 can supplement suboptimal Lck-independent MAPK activation in a STAT-3-dependent manner in human CD8(+) T cells.
Journal of immunology (Baltimore, Md. : 1950)
Imataki O,Ansén S,Tanaka M,Butler MO,Berezovskaya A,Milstein MI,Kuzushima K,Nadler LM,Hirano N
MA5-11532 was used in flow cytometry to investigate the role of CD8 and antigen in proliferation of CTLs.
Wed Feb 15 00:00:00 EST 2012
The significance of multiplex PCR/heteroduplex analysis-based TCR-γ gene rearrangement combined with laser-capture microdissection in the diagnosis of early mycosis fungoides.
Journal of cutaneous pathology
Yang H,Xu C,Tang Y,Wan C,Liu W,Wang L
MA5-11532 was used in immunohistochemistry (paraffin) to study the diagnostic value for detecting early mycosis fungoides of a combination of multiplex PCR analyis of TCRgamma gene rearrangements and laser-capture microdissection.
Thu Mar 01 00:00:00 EST 2012
Serous effusion cytology of extranodal natural killer/T-cell lymphoma.
Cytopathology : official journal of the British Society for Clinical Cytology
Su XY,Huang J,Jiang Y,Tang Y,Li GD,Liu WP
MA5-11532 was used in flow cytometry to characterize extranodal natural killer/T-cell lymphoma, nasal type using patient samples.
Sun Apr 01 00:00:00 EDT 2012
Effector Memory T cells Are Associated With Atherosclerosis in Humans and Animal Models.
Journal of the American Heart Association
Ammirati E,Cianflone D,Vecchio V,Banfi M,Vermi AC,De Metrio M,Grigore L,Pellegatta F,Pirillo A,Garlaschelli K,Manfredi AA,Catapano AL,Maseri A,Palini AG,Norata GD
MA5-11532 was used in flow cytometry to study the relationship between circulating CD4+ T-cell subsets and atherosclerosis
Wed Feb 01 00:00:00 EST 2012
Axonal projections of mechanoreceptive dorsal root ganglion neurons depend on Ret.
Development (Cambridge, England)
Honma Y,Kawano M,Kohsaka S,Ogawa M
MA5-11532 was used in immunohistochemistry to study the role of Ret in the neurturin-mediated formation of precise axonal projections in the central nervous system
Thu Jul 01 00:00:00 EDT 2010
A patient with clinicopathologic features of small plaque parapsoriasis presenting later with plaque-stage mycosis fungoides: report of a case and comparative retrospective study of 27 cases of "nonprogressive" small plaque parapsoriasis.
Journal of the American Academy of Dermatology
Belousova IE,Vanecek T,Samtsov AV,Michal M,Kazakov DV
MA5-11532 was used in immunohistochemistry to report on a case of small plaque parapsoriasis with plaque-stage mycosis fungoides
Mon Sep 01 00:00:00 EDT 2008
Hyperplasia of hair follicles and other adnexal structures in cutaneous lymphoproliferative disorders: a study of 53 cases, including so-called pseudolymphomatous folliculitis and overt lymphomas.
The American journal of surgical pathology
Kazakov DV,Belousova IE,Kacerovska D,Sima R,Vanecek T,Vazmitel M,Pizinger K,Michal M
MA5-11532 was used in immunohistochemistry to study 53 cases of cutaneous lymphoproliferative disorders
Wed Oct 01 00:00:00 EDT 2008
Natural regulatory T cells: number and function are normal in the majority of patients with lupus nephritis.
Clinical and experimental immunology
Yates J,Whittington A,Mitchell P,Lechler RI,Lightstone L,Lombardi G
MA5-11532 was used in flow cytometry to examine the frequency, phenotype, and function of T regulatory cells in patients with systemic lupus erythematosus and inactive disease
Tue Jul 01 00:00:00 EDT 2008
Up-regulation of expression of the ubiquitin carboxyl-terminal hydrolase L1 gene in human airway epithelium of cigarette smokers.
Cancer research
Carolan BJ,Heguy A,Harvey BG,Leopold PL,Ferris B,Crystal RG
MA5-11532 was used in immunocytochemistry to investigate the expression of carboxyl-terminal hydrolyase L1 gene in human airway epithelium of cigarette smokers
Wed Nov 15 00:00:00 EST 2006
Monocyte-derived human macrophages mediate anergy in allogeneic T cells and induce regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Hoves S,Krause SW,Schütz C,Halbritter D,Schölmerich J,Herfarth H,Fleck M
MA5-11532 was used in flow cytometry to assess the T cell-stimulatory capacity of mature dendritic cells and macrophages using a model of allogeneic MLR
Tue Aug 15 00:00:00 EDT 2006
Primary cutaneous histiocyte and neutrophil-rich CD30+ and CD56+ anaplastic large-cell lymphoma with prominent angioinvasion and nerve involvement in the forehead and scalp of an immunocompetent woman.
Journal of cutaneous pathology
Boudova L,Kazakov DV,Jindra P,Sima R,Vanecek T,Kuntscher V,Vera V,Bouda J,Michal M
MA5-11532 was used in immunohistochemistry to report on a case of utaneous lymphoma of the head co-expressing CD56 and CD30
Tue Aug 01 00:00:00 EDT 2006
CIK cells from patients with HCC possess strong cytotoxicity to multidrug-resistant cell line Bel-7402/R.
World journal of gastroenterology
Zhang YS,Yuan FJ,Jia GF,Zhang JF,Hu LY,Huang L,Wang J,Dai ZQ
MA5-11532 was used in immunohistochemistry to investigate the effectiveness of cytokine-induced killer cell treatment in patient with hepatocellular carcinoma
Tue Jun 14 00:00:00 EDT 2005
Cutaneous lymphoid hyperplasia and other lymphoid infiltrates of the breast nipple: a retrospective clinicopathologic study of fifty-six patients.
The American Journal of dermatopathology
Boudova L,Kazakov DV,Sima R,Vanecek T,Torlakovic E,Lamovec J,Kutzner H,Szepe P,Plank L,Bouda J,Hes O,Mukensnabl P,Michal M
MA5-11532 was used in immunohistochemistry to study the clinicopathological spectrum of lymphoproliferations and other lymphoid infiltration of the breast nipple
Sat Oct 01 00:00:00 EDT 2005
T-cell-rich B-cell lymphoma: a clinicopathologic study of 21 cases and comparison with 43 cases of diffuse large B-cell lymphoma.
Leukemia research
Aki H,Tuzuner N,Ongoren S,Baslar Z,Soysal T,Ferhanoglu B,Sahinler I,Aydin Y,Ulku B,Aktuglu G
MA5-11532 was used in immunohistochemistry to compare the clinocopathology of T-cell rich B-cell lymphoma and diffuse large B-cell lymphoma
Mon Mar 01 00:00:00 EST 2004
Chemokine receptor CCR4 on CD4+ T cells in juvenile rheumatoid arthritis synovial fluid defines a subset of cells with increased IL-4:IFN-gamma mRNA ratios.
Journal of immunology (Baltimore, Md. : 1950)
Thompson SD,Luyrink LK,Graham TB,Tsoras M,Ryan M,Passo MH,Glass DN
MA5-11532 was used in flow cytometry to elucidate the mechanisms that promote recruitment and survival of T cells within the pediatric inflamed joint.
Fri Jun 01 00:00:00 EDT 2001
Quantification of CD4, CCR5, and CXCR4 levels on lymphocyte subsets, dendritic cells, and differentially conditioned monocyte-derived macrophages.
Proceedings of the National Academy of Sciences of the United States of America
Lee B,Sharron M,Montaner LJ,Weissman D,Doms RW
MA5-11532 was used in flow cytometry to determine the number of CD4, CCR5, and CXCR4 antibody-binding sites on various T cells and macrophages.
Tue Apr 27 00:00:00 EDT 1999
CK beta-11/macrophage inflammatory protein-3 beta/EBI1-ligand chemokine is an efficacious chemoattractant for T and B cells.
Journal of immunology (Baltimore, Md. : 1950)
Kim CH,Pelus LM,White JR,Applebaum E,Johanson K,Broxmeyer HE
MA5-11532 was used in flow cytometry to determine the functional properties of CK beta-11/MIP-3 beta/ELC.
Sun Mar 01 00:00:00 EST 1998
Functional subsets of human helper-inducer cells defined by a new monoclonal antibody, UCHL1.
Immunology
Smith SH,Brown MH,Rowe D,Callard RE,Beverley PC
MA5-11532 was used in flow cytometry, immunohistochemistry - frozen section, and immunoprecipitation to characterize the monoclonal antibody UCHL1.
Thu May 01 00:00:00 EDT 1986