Product References
Targeting L1 cell adhesion molecule expression using liposome-encapsulated siRNA suppresses prostate cancer bone metastasis and growth.
Oncotarget
Sung SY,Wu IH,Chuang PH,Petros JA,Wu HC,Zeng HJ,Huang WC,Chung LW,Hsieh CL
MA5-14137 was used in ELISA to determine how using liposome-encapsulated siRNA to target L1 cell adhesion molecule expression suppresses prostate cancer bone metastasis and growth
Thu Oct 30 00:00:00 EDT 2014
Adhesion molecule L1 is down-regulated in malignant peripheral nerve sheath tumors versus benign neurofibromatosis type 1-associated tumors.
Oral surgery, oral medicine, oral pathology and oral radiology
Blessmann M,Gröbe A,Quaas A,Kaifi JT,Mistakidis G,Bernreuther C,Sauter G,Gros S,Rawnaq T,Friedrich R,Mautner VF,Smeets R,Heiland M,Schachner M,Izbicki JR
MA5-14137 was used in immunohistochemistry to study the expression of the L1 adhesion molecule in malignant peripheral nerve sheath tumors and benign neurofibromatosis type 1-associated tumors
Wed Feb 01 00:00:00 EST 2012
Two alcohol binding residues interact across a domain interface of the L1 neural cell adhesion molecule and regulate cell adhesion.
The Journal of biological chemistry
Dou X,Menkari CE,Shanmugasundararaj S,Miller KW,Charness ME
MA5-14137 was used in flow cytometry to characterize two alcohol binding residues in L1 neural cell adhesion molecule and their involvement in cell adhesion
Fri May 06 00:00:00 EDT 2011
DOG1 and CD117 are the antibodies of choice in the diagnosis of gastrointestinal stromal tumours.
Histopathology
Novelli M,Rossi S,Rodriguez-Justo M,Taniere P,Seddon B,Toffolatti L,Sartor C,Hogendoorn PC,Sciot R,Van Glabbeke M,Verweij J,Blay JY,Hohenberger P,Flanagan A,Dei Tos AP
MA5-14137 was used in immunohistochemistry to study the value of COG1 and CD117 immunohistochemistry in the diagnosis of gastrointestinal stromal tumors
Sun Aug 01 00:00:00 EDT 2010
Subset of esophageal adenocarcinoma expresses adhesion molecule l1 in contrast to squamous cell carcinoma.
Anticancer research
Rawnaq T,Kleinhans H,Uto M,Schurr PG,Reichelt U,Cataldegirmen G,Gawad KA,Yekebas EF,Schachner M,Izbicki JR,Kaifi JT
MA5-14137 was used in immunohistochemistry to study neural cell adhesion molecule L1 overexpression in a subgroup of esophageal cancer
Wed Apr 01 00:00:00 EDT 2009
L1 is associated with micrometastatic spread and poor outcome in colorectal cancer.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Kaifi JT,Reichelt U,Quaas A,Schurr PG,Wachowiak R,Yekebas EF,Strate T,Schneider C,Pantel K,Schachner M,Sauter G,Izbicki JR
MA5-14137 was used in immunohistochemistry to study the role of the L1 adhesion molecule in micrometastatic spread and poor outcome in colorectal cancer
Thu Nov 01 00:00:00 EDT 2007
L1 is associated with favorable outcome in neuroblastomas in contrast to adult tumors.
Annals of surgical oncology
Wachowiak R,Fiegel HC,Kaifi JT,Quaas A,Krickhahn A,Schurr PG,Erttmann R,Schachner M,Kluth D,Sauter G,Izbicki JR
MA5-14137 was used in immunohistochemistry to study the association of the L1 neuronal adhesion molecule with favorable outcome in childhood neuroblastomas
Sat Dec 01 00:00:00 EST 2007
L1 (CD171) is highly expressed in gastrointestinal stromal tumors.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Kaifi JT,Strelow A,Schurr PG,Reichelt U,Yekebas EF,Wachowiak R,Quaas A,Strate T,Schaefer H,Sauter G,Schachner M,Izbicki JR
MA5-14137 was used in immunohistochemistry to study the expression of L1 (CD171) in gastrointestinal stromal tumors
Wed Mar 01 00:00:00 EST 2006
Characterization of gene expression in mucinous cystic neoplasms of the pancreas using oligonucleotide microarrays.
Oncogene
Fukushima N,Sato N,Prasad N,Leach SD,Hruban RH,Goggins M
MA5-14137 was used in immunohistochemistry to investigate the changes of gene expression in pancreatic mucinous cystic neoplasms
Thu Dec 02 00:00:00 EST 2004
Identification of cell surface and secreted proteins essential for tumor cell survival using a genetic suppressor element screen.
Oncogene
Gelman MS,Ye XK,Stull R,Suhy D,Jin L,Ng D,Than B,Ji M,Pan A,Perez P,Sun Y,Yeung P,Garcia LM,Harte R,Lu Y,Lamar E,Tavassoli R,Kennedy S,Osborn S,Chin DJ,Meshaw K,Holzmayer TA,Axenovich SA,Abo A
MA5-14137 was used in flow cytometry to identify molecular markers and therapeutic targets for tumors
Thu Oct 21 00:00:00 EDT 2004
Patterning axonal guidance molecules using a novel strategy for microcontact printing.
Neurochemical research
Oliva AA,James CD,Kingman CE,Craighead HG,Banker GA
MA5-14137 was used in western blot to develop a protocol for microprinting proteins on a substrate in order to control neurite growth in culture
Sat Nov 01 00:00:00 EST 2003