Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17047 targets CD6 in IHC, indirect ELISA, and WB applications and shows reactivity with Human samples.
The MA5-17047 immunogen is purified recombinant fragment of human CD6 (amino acids: 472-668) expressed in E. Coli.
MA5-17047 detects CD6 which has a predicted molecular weight of approximately 71.8kDa.
CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD6; CD6 antigen; FLJ44171; Soluble CD6; T-cell differentiation antigen CD6; T12; TP120
Gene Aliases: CD6; TP120
UniProt ID: (Human) P30203
Entrez Gene ID: (Human) 923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support