Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17931 targets CD147/BSG in FACS applications and shows reactivity with Mouse samples.
The MA5-17931 immunogen is recombinant md0-2rCD4d3+4.
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and malignant tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: 5A11/Basigin; Basic immunoglobulin superfamily; Basigin; CD147; gp 42; HT7 antigen; Membrane glycoprotein gp42; neurothelin
Gene Aliases: AI115436; AI325119; Bsg; CD147; EMMPRIN; HT-7
UniProt ID: (Mouse) P18572
Entrez Gene ID: (Mouse) 12215
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support