Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes the p28 subunit either as part of a p28/EBI-3 heterodimer or as a free subunit.
IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epstein-Barr virus-induced gene 3 protein homolog; IL-27 p28 subunit; IL-27 subunit alpha; IL-27 subunit beta; IL-27-A; IL-27B; IL27 p28; IL27-A; ILN; Interleukin; interleukin 30; Interleukin-27 subunit alpha; Interleukin-27 subunit beta; M-IL-27 (p28); MGC71873; p28
Gene Aliases: Ebi3; Il27; Il27a; Il27b
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support