Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mouse anti Horse MHC Class I Monomorphic antibody, clone CVS22 recognizes monomorphic equine MHC Class I and was classified at the International Equine Leucocyte Antigen Workshop.
MHC I Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MHC class I antigen 10.b; MHC class I antigen 2.5; MHC class I antigen 3.1; MHC class I antigen 3.6; MHC class I antigen 9.5; MHC class I antigen 9.6
Gene Aliases: Eqca-1
Entrez Gene ID: (Horse) 100034210, (Horse) 100034211, (Horse) 100034209, (Horse) 100056391
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support