Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-25721 detects IKK alpha from human, mouse, and rat samples.
PA1-25721 has been successfully used in Western blot procedures.
The PA1-25721 immunogen is a synthetic peptide corresponding to residues 311-377 of human IKK alpha.
Inhibitor of Nuclear factor Kappa-B kinase subunit alpha (IKK alpha) is encoded by the CHUK gene and located on chromosome 10 in humans. It is also known as IKK1 or CHUK (Conserved helix-loop-helix ubiquitous kinase). IKK alpha belongs to the family of serine threonine protein kinase and forms a part of IKK complex. The IKK complex is required for the activation of NF-kB with IKK alpha specifically known to be involved in the non-canocial pathway for NF-kB activation. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; FLJ40509; I kappa B kinase 1; I-kappa-B kinase 1; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; ikappaB kinase; IkappaB kinase alpha; IkB kinase alpha subunit; IKBKB; IKK alpha; IKK-A; IKK-a kinase; IKK-B; IKK-beta; IKK1; IKKB; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NFkappaB inhibitor kinase alpha; TCF-16; Transcription factor 16
Gene Aliases: CHUK; IKKA; TCF16
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support