Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
PA1-26678 detects ERK1 + ERK2 (phospho T185 + T202) from human samples. PA1-26678 is expected to cross react with a wide range of species due to sequence homology.
PA1-26678 has been successfully used in Western blot procedures.
The PA1-26678 immunogen is a synthetic phosphopeptide corresponding to a portion of human ERK 1 + 2 containing threonine 202/185 and tyrosine 204/187. This portion is conserved among many species including rat, mouse, cow, frog, snail, nematode, and fruit
ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERK-1; ERK-2; erk1 erk2; ERK1b; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-related kinase 1; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; Microtubule-associated protein 2 kinase; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; p42 MAP Kinase; p42-MAPK; p44-ERK1; p44-MAPK; protein tyrosine kinase ERK2
Gene Aliases: ERK1; ERK2; MAPK1; MAPK3; PRKM1; PRKM2; PRKM3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support