Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX25074-PEP)..
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FKHL20; Forkhead box protein A2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3, beta; Hepatocyte nuclear factor 3-beta; HNF-3-beta; HNF-3B; MGC19807; RONU; TCF-3B; Transcription factor 3B; WHN
Gene Aliases: FOXA2; HNF3B; Tcf-3b; TCF3B
UniProt ID: (Human) Q9Y261, (Rat) P32182
Entrez Gene ID: (Human) 3170, (Rat) 25099
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support