Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with chicken, mouse, rat and Xenopus and zebrafish based on sequence homology.
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; Homeobox protein PITX2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S α 1 glycoprotein; S α1glycoprotein; Solurshin
Gene Aliases: ARP1; PITX2; RGS; RIEG; RIEG1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support