Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of total TUTase.
TUT1 is a nucleotidyl transferase that functions as both a terminal uridylyltransferase and a nuclear poly(A) polymerase. TUT1 specifically adds and removes nucleotides from the 3' end of small nuclear RNAs and select mRNAs and may function in controlling gene expression and cell proliferation. TUT1 specifically catalyzes uridylylation of U6 snRNA (RNU6; MIM 180692) and is essential for cell proliferation (Trippe et al., 2006 [PubMed 16790842]).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21850; FLJ22267; FLJ22347; MGC131987; MGC149809; nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpha regulated-poly(A) polymerase; nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase; PAP-associated domain-containing 2; poly(A) polymerase associated domain containing 2; RNA binding motif protein 21; RNA uridylyltransferase; RNA-binding motif protein 21; RNA-binding protein 21; Speckle targeted PIP5K1A-regulated poly(A) polymerase; Star-PAP; terminal uridylyl transferase 1 U6 snRNA-specific; TUTase 6; U6 snRNA-specific terminal uridylyltransferase 1; U6 TUTase; U6-TUTase; up-regulated in lung cancer 6
Gene Aliases: 2700038E08Rik; PAPD2; RBM21; STARPAP; TUT1; TUTase; TUTase6; URLC6
UniProt ID: (Human) Q9H6E5, (Rat) Q3MHT4, (Mouse) Q8R3F9
Entrez Gene ID: (Human) 64852, (Rat) 499314, (Mouse) 70044
Molecular Function: nucleotidyltransferase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support