Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dermal papilla-derived protein 2; DKFZp566C0746; FLJ26584; Growth hormone-inducible transmembrane protein; MICS1; Mitochondrial morphology and cristae structure 1; transmembrane BAX inhibitor motif containing 5; Transmembrane BAX inhibitor motif-containing protein 5
Gene Aliases: DERP2; GHITM; HSPC282; MICS1; My021; PTD010; TMBIM5; UNQ244/PRO281
UniProt ID: (Human) Q9H3K2
Entrez Gene ID: (Human) 27069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support