Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, canine, mouse, porcine, rabbit and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IGF-1; IGF; IGF-I; IGF-IA; IGF-IB; IGF1A; IGFIa; Insulin like growth factor; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; M-IGF-1; Mechano growth factor; MGF; OTTHUMP00000195084; R-IGF-1; Somatomedin C; Somatomedin-C
Gene Aliases: IBP1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019
Entrez Gene ID: (Human) 3479
Molecular Function: growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support