Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IKK2 binding protein; NIK and IKK(beta) binding protein; NIK and IKK-beta binding protein; NIK and IKK{beta} binding protein; NIK- and IKBKB-binding protein; Trafficking protein particle complex subunit 9; TRAPP 120 kDa subunit; Tularik gene 1 protein
Gene Aliases: 1810044A24Rik; 2900005P22Rik; 4632408O18Rik; IBP; IKBKBBP; KIAA1882; mKIAA1882; MRT13; NIBP; T1; TRAPPC9; TRS120; TRS130
UniProt ID: (Human) Q96Q05, (Bovine) Q32PH0, (Mouse) Q3U0M1
Entrez Gene ID: (Dog) 482058, (Chicken) 420312, (Human) 83696, (Bovine) 533451, (Mouse) 76510
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support