Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody detects endogenous protein at a molecular weight of 55 and 60 kDa.
Purity is >95% by SDS-PAGE.
Smad proteins, the mammalian homologs of the Drosophila mothers against decapentaplegic (Mad), have been implicated as downstream effectors of TGFβ/BMP signaling. Smad1 (also designated Madr1 or JV4-1) and Smad5 are effectors of BMP-2 and BMP-4 function, while Smad2 (also designated Madr2 or JV18-1) and Smad3 are involved in TGFβ and Activin-mediated growth modulation. Smad4 (also designated DPC4) has been shown to mediate all of the above activities through interaction with various Smad family members. Smad6 and Smad7 regulate the response to Activin/TGFβ signaling by interfering with TGFβ-mediated phosphorylation of other Smad proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMAD-2; hMAD-3; hSMAD3; JV15-2; JV18-1; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 2; MAD homolog 3; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; Mad-related protein 2; mad3; mMad2; mMad3; mother against DPP homolog 2; Mothers against decapentaplegic homolog 2; Mothers against decapentaplegic homolog 3; mothers against DPP homolog 2; mothers against DPP homolog 3; pSMAD2; Sma- and Mad-related protein 2; SMA- and MAD-related protein 3; SMAD; SMAD 2; SMAD 3; SMAD family member 2; SMAD family member 3; SMAD, mothers against DPP homolog 2; SMAD, mothers against DPP homolog 3
Gene Aliases: 7120426M23Rik; AU022421; hMAD-2; hSMAD2; HSPC193; HsT17436; JV15-2; JV18; JV18-1; LDS1C; LDS3; mad3; MADH2; MADH3; MADR2; mMad2; Smad 3; Smad-2; SMAD2; SMAD3
UniProt ID: (Human) Q15796, (Human) P84022, (Mouse) Q62432, (Rat) O70436, (Mouse) Q8BUN5, (Rat) P84025
Entrez Gene ID: (Human) 4087, (Human) 4088, (Mouse) 17126, (Rat) 29357, (Mouse) 17127, (Rat) 25631
Molecular Function: DNA-binding transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support