Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody detects endogenous protein at a molecular weight of 70-82 kDa.
Purity is >95% by SDS-PAGE.
FOXO1 + FOX3 + FOX4 belong to the forkhead family of transcription factors. FOXO1 (FKHR or ForkHead in Rhabdomyosarcoma) is a 70 kDa protein. Recent studies have shown that this protein can act as either a coactivator or a corepressor of nuclear receptor activity. This action is mediated through the LXXLL motif found in the C terminus of the FOXO1A protein. The specific function of the gene has not yet been determined.FOX3 probably functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of the FOXO3 gene with the MLL gene is associated with secondary acute leukemia.FOX4 is a forkhead transcription factor involved in the regulation of the insulin signaling pathway. It binds to insulin-response elements (IREs) and can activate transcription of IGFBP1. FOXO4 down-regulates expression of HIF1A and suppresses hypoxia-induced transcriptional activation of HIF1A-modulated genes. It is also involved in negative regulation of the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF6q21 protein; Afxh; Fork head domain transcription factor AFX1; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box O3A; Forkhead box protein O1; Forkhead box protein O1A; Forkhead box protein O3; Forkhead box protein O4; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma; Forkhead in rhabdomyosarcoma-like 1; forkhead protein; forkhead protein 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; MGC120490; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7
Gene Aliases: 1110048B16Rik; 2010203A17Rik; AF6q21; AFX; AFX1; Afxh; AI876417; C76856; FKH1; FKHR; Fkhr1; Fkhr2; Fkhr3; FKHRL1; FKHRL1P2; FOXO1; FOXO1A; FOXO2; FOXO3; FOXO3A; FOXO4; MLLT7; RGD1561201
UniProt ID: (Human) Q12778, (Human) O43524, (Human) P98177, (Rat) G3V7R4, (Mouse) Q9R1E0, (Mouse) Q9WVH4, (Mouse) Q9WVH3
Entrez Gene ID: (Human) 2308, (Human) 2309, (Human) 4303, (Rat) 84482, (Mouse) 56458, (Mouse) 56484, (Rat) 294515, (Rat) 302415, (Mouse) 54601
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support