Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with human, rat, dog, pig and cow based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761G1421; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; Ras GTPase-activating protein SynGAP; Ras/Rap GTPase-activating protein SynGAP; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein, 135kDa; Synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1
Gene Aliases: Gm1963; KIAA1938; MRD5; RASA1; RASA5; SYNGAP; SYNGAP1
UniProt ID: (Human) Q96PV0, (Mouse) F6SEU4
Entrez Gene ID: (Human) 8831, (Mouse) 240057
Molecular Function: GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support