Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of choline/ethanolamine kinase family which catalyzes the first step of phosphatidylethanolamine (PtdEtn) biosynthesis via the cytidine diphosphate (CDP) ethanolamine pathway. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2014].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EKI 2; Ethanolamine kinase 2; Ethanolamine kinase-like protein; putative ethanolamine kinase
Gene Aliases: EKI2; ETNK2; HMFT1716; Tuc1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support