Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IPVAAAKETV AIGRTDIEDL DLYATSRERR FRLFASIECE GQLFMTPYDF ILAVTTDEPK VAKTWKSLSK QELNQMLAET PPVWKGSSKL FRNLK
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
May play a role in mitochondrial calcium uptake. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium uptake protein 3, mitochondrial; EF hand domain family A2; EF hand domain family, member A2; EF-hand domain family, member A2; EF-hand domain-containing family member A2; mitochondrial calcium uptake family, member 3; mitochondrial uptake family, member 3
Gene Aliases: EFHA2; MICU3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support