Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NWSVRRSDFR FSEVHGAENF IQHNLQTLKS KIFDNESA
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene aligns with an internal intron of the retinoblastoma susceptibility gene in the reverse orientation. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled purinergic receptor P2Y5; LPA receptor 6; LPA-6; LPA6 Receptor; Lysophosphatidic acid receptor 6; Oleoyl-L-alpha-lysophosphatidic acid receptor; P2Y purinoceptor 5; P2Y5; Purinergic receptor 5; purinergic receptor P2Y G protein-coupled protein 5; purinergic receptor P2Y, G-protein coupled, 5; RB intron encoded G-protein coupled receptor
Gene Aliases: LPAR6; P2RY5
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support