Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YPEDNFPDEE KASTIKHCPD SESRAPQILR RQSSPSCGPV AETSSIGNGD GISKLMSENT EGSAQAPQLP RKKDKRDFPK PA
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 82%.
MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases and has been implicated as an upstream regulator of MAP kinase signaling pathways. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for MAP4K5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: germinal center kinase-related; KHS; Kinase homologous to SPS1/STE20; MAPK/ERK kinase kinase kinase 5; MEK kinase kinase 5; MEKKK 5; Mitogen-activated protein kinase kinase kinase kinase 5
Gene Aliases: GCKR; KHS; KHS1; MAP4K5; MAPKKKK5
UniProt ID: (Human) Q9Y4K4
Entrez Gene ID: (Human) 11183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support