Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SVPALYSDAY DISQIIELYN LPDENEISSS SFHCLYQYLS LTKYQVGGQG SSLSIFAGPA VGLSPAQTWP NM
Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 26%.
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the ARGFX homeobox gene family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arginine-fifty homeobox
Gene Aliases: ARGFX
UniProt ID: (Human) A6NJG6
Entrez Gene ID: (Human) 503582
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support