Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RAILCYRNTS SICSNEGLIF KLKRGKEACA LDTVGWVQRH RKMLRHCPS
Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 37%.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 1; CCL 1; chemokine (C-C motif) ligand 1; inflammatory cytokine I-309; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); Small-inducible cytokine A1; T lymphocyte-secreted protein I-309
Gene Aliases: CCL1; I-309; P500; SCYA1; SISe; TCA3
UniProt ID: (Human) P22362
Entrez Gene ID: (Human) 6346
Molecular Function: cytokine intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support