Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPFRAFPGGDK FGTTFLSAAA KAQGFGDAKS RARYGAGQQD LATPLESGAG ARGSFNKFQP QPSTPQPQPS PQPQPQQQQP QPQPPAQPHL YLQRGACKTP PDGSLKLQEG S
Highest antigen sequence identity to the following orthologs - mouse 80%, rat 81%.
This gene encodes a paired-like homeodomain transcription factor expressed in the mesenchyme of developing bones, limbs, hair, teeth, and mammary tissue. Mutations in this gene cause parietal foramina 2 (PFM2); an autosomal dominant disease characterized by deficient ossification of the parietal bones. Mutations in this gene also cause a form of frontonasal dysplasia with alopecia and hypogonadism; suggesting a role for this gene in craniofacial development, mesenchymal-epithelial communication, and hair follicle development. Deletion of a segment of chromosome 11 containing this gene, del(11)(p11p12), causes Potocki-Shaffer syndrome (PSS); a syndrome characterized by craniofacial anomalies, mental retardation, multiple exostoses, and genital abnormalities in males. In mouse, this gene has been shown to use dual translation initiation sites located 16 codons apart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aristaless-like homeobox 4; Homeobox protein aristaless-like 4; homeodomain transcription factor ALX4
Gene Aliases: ALX4; CRS5; FND2; KIAA1788
UniProt ID: (Human) Q9H161
Entrez Gene ID: (Human) 60529
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support